Sigma-Aldrich
Fluka & Supelco - Analytical Logo
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Analytical / Chromatography > Air Monitoring > Learning Center > Thermal Desorption Tubes
Air Monitoring
Thermal Desorption Tubes
Overview
Tubes by Instrument Manufacturer
VOST Stack Sampling Tubes
Tubes for Low Concentration Odors - NEW!
Selection Guide
Custom Tubes
Overview back to top
Thermal Desorption Introduction
Supelco produces a comprehensive selection of adsorbent tubes – Carbotrap™ tubes – offering superior performance for trapping and thermally desorbing organic compounds. Selection of the proper adsorbent(s) is critical to achieving the best results for a thermal desorption application. An ideal adsorbent tube will trap and retain compounds of interest for the entire sampling period, then allow total analyte desorption without thermal decomposition. The rate of release should be as rapid as possible, to minimize analysis time and provide the most efficient separation. Stable adsorbents ensure the best detection limits by minimizing the possibility of breakdown products interfering with quantification.
Multi-Bed Adsorbent Tubes
Because no single adsorbent is capable of trapping and efficiently releasing all compounds, many Carbotrap™ thermal desorption tubes contain more than one adsorbent. Multiple beds of adsorbents enable you to analyze a wider range of compounds in a single sampling.
In multi-bed adsorbent tubes, the adsorbents are arranged in order of increasing adsorbent strength, from sample inlet to sample outlet. The largest molecules in the sample are trapped by the first bed of adsorbent. Smaller molecules are trapped by the succeeding, stronger beds. To avoid forcing analytes through an adsorbent that is too strong, desorption flow is always in the direction opposite of sample collection flow.
Tube Construction
The design of a thermal desorption tube and its contents are critical to a well-performing air monitoring system. Glass tube blanks are manufactured to close tolerances, to ensure consistent lot-to-lot performance. For identification, we mark each of our glass tubes with a unique number. Thermal desorption tubes are most commonly made of glass or stainless steel, with glass wool, wire gauze, or glass frits to hold the adsorbents in place. Tube lengths and outside diameters are dictated by the instrument of choice. Tubes having a large internal volume (4mm I.D. or greater) are typically used for sampling because they hold relatively large size beds of adsorbents. Analytical results can be improved by transferring the analytes from the large I.D. sampling tube to a tube of smaller I.D. (1-2mm), thus “focusing” the sample into a smaller volume. Subsequent desorption from the smaller I.D. tube to the chromatography column gives a higher linear velocity of desorption flow through the tube, delivering the sample to the column more rapidly and in a much narrower band.
Empty Tubes
We mark these specially cleaned and silanized glass tubes with an arrow to indicate sampling flow direction. Fill them in this direction with successively stronger adsorbents. You can fill an empty 4mm I.D. tube with adsorbents for air sampling, or with a solid sample (soil, plastic, etc.) for a thermal extraction analysis. Use our empty 1mm I.D. and 2mm I.D. tubes to create your own focusing tubes, for better chromatography.
To trap highly sensitive compounds, use an adsorbent tube with a frit in the inlet rather than a glass wool plug. The frit will hold the adsorbent in place without adsorbing or decomposing the analytes; glass wool plugs can adsorb or degrade sensitive compounds.
Tubes by Instrument Manufacturer back to top
Tubes for PerkinElmer® Instruments (ordering information)
Supelco offers both Stainless Steel and Glass pre-packed sampling tubes that are fully compatible with the ATD-50, the ATD-400, and the new TurboMatrix™ instrument. Each tube has a unique number. The tubes are available either pre-conditioned and ready for use with brass Swagelok® end caps or unconditioned with PTFE caps.
Tubes for Teledyne Tekmar Instruments (ordering information)
Supelco offers both glass and stainless steel tubes for Teledyne Tekmar instruments. Our prepacked tubes are thermally conditioned and tested for background levels and backpressure. The ¼ in. O.D. tubes are sealed in our exclusive TDS3™ storage container and the ½ in. O.D. tubes are sealed in a glass storage container. Fits AEROTrap 6000 instrument.
Tubes for Gerstel ThermoDesorption System (ordering information)
Supelco offers both stainless steel and glass pre-packed sampling tubes that are fully compatible with the Gerstel instrument. Each tube is thermally conditioned, and batch tested for backpressure and background. The pre-packed tubes are sealed in our exclusive TDS3™ storage containers. All of the pre-packed glass sampling tubes incorporate a glass frit at the inlet, which increases their performance, and are individually numbered. The stainless steel tubes use a SS screen at both inlet and outlet to keep the adsorbent beds secure during use. Each stainless steel tube includes two stainless steel screens.
Tubes for CDS/DYNATHERM Instruments (ordering information)
Supelco offers a complete line of both pre-packed and empty tubes for the DYNATHERM Thermal Desorbers. Each tube is individually numbered, thermally conditioned, and batch tested for backpressure and background. The pre-packed Standard Sampling Tubes are sealed in our exclusive TDS3™ storage containers and the Fast-Flow Tubes are sealed with Swagelok® fittings. All of the pre-packed sampling tubes incorporate a glass frit at the inlet, which increases their performance. The Focusing tubes are used to refocus the sample for better chromatography of the early eluting compounds.
VOST Stack Sampling Tubes (ordering information) back to top
VOST (Volatile Organic Sampling Train) Tubes are designed to meet specifications in US EPA SW-846, Method 0030. Each tube is individually numbered, pre-conditioned, and sealed with stainless steel Swagelok® fittings before stored in a glass storage container. Each lot is tested for background and backpressure.
Thermal Desorption Tubes for Low Concentration Odors - New!
(ordering information)
back to top
Olfactory Measurement by Triangle Odor Bag Method
This product is designed specifically for collecting odors in low concentrations. The tube incorporates a glass frit at the inlet to prevent the adsorbent from exiting during use. Each tube includes a unique number for easy identification. Maximum temperature: 320°C.
This tube is comprised of the following adsorbents: (packed by volume)
Adsorbent Length (approx. weight)
Tenax® TA (35/60) 54mm (0.84 grams)
Carbotrap™ B (20/40) 22mm (0.44 grams)
Carbosieve™-SIII (60/80) 12mm (0.44 grams)
Tubes are conditioned in an oven at 250°C with 90-100mL/min of nitrogen flow for 8-hrs before placement in its storage container. Because the product is designed to collect odors at low concentrations, it is important for the product to remain in the storage container until use and to be analyzed immediately (within hours of sampling) for best results. If samples cannot be analyzed within hours, store refrigerated at 4°C.