|
AQUANAL®
AQUANAL SPECTRO 3 Photometer
|
A sophisticated solution for your water analysis. AQUANAL®-SPECTRO 3 Photometer can be used with both AQUANAL®-plus and AQUANAL®-professional reagents. Operations in UV and infrared spectral range are possible.
Features of SPECTRO 3 Photometer
- Built-in calibration curves for 100 determinations
- Combined LED and fibre-optic technology
- Spectral range: 380 / 430 / 470 / 500 / 520 / 560 / 610 / 700 / 810 nm
- Electro-optical accuracy: ±1.5 %
- Absorbance range: –0.500 to 3.500 Abs
- Test chamber for 16 mm tubes
- Software: operates with Windows 98 and higher versions
- Data memory for 1000 results, with data protection
- History view – list of the latest 50 results
- GLP compliant (results given with time / date / place)
- Automatic blank setting
- Comprehensive recognition of measurement faults
- Power-saving function, automatic power off
- Test result displayed in mg/L or in extinction mode
- Automatic selection of the wavelength
For your convenience
Built-in user prompt in different languages (10 steps instructions) favourites – selection of the most frequently used curves language-selection (choice of six languages)
The SPECTRO 3 Photometer comes in this package:
- Spectro 3 photometer with software
- External power pack
- Four rechargeable batteries (1.5 V)
- Infrared interface
- Instruction manual
- Small funnel
- Six 16 mm diameter tubes
- Four 20 ml sample tubes
- Packed in one case
- USB-To-IrDA-Adapter
| Product # |
Product Name |
Add to Cart |
| 70403 |
AQUANAL® Spectro 3 Photometer |
|
| 37561 |
Refill 16 mm tubes, 5 pcs. |