Analytical / Chromatography
Air Monitoring
Analytical Reagents
AQUANAL
Compact Labs
Dealership
LabInfos
Literature Request
Photometers
Prof. Reagent Tests
Test Kits
Test Sticks
Thermoblock
Tube Tests
Vario H Powder Packs
Waste Water Analysis
Analytic basic reagents
Derivatization Reagents
Electrophoresis
Sensoric Applications
Solvents
Trace Analysis
Water Analysis
Analytical Standards
Gas Chromatography
HPLC
Microbiology
Microscopy
Sample Preparation
Spectroscopy
Syringes
Titration
TLC
Vials
Videos
Events
Literature
Analytix
The Reporter
Reporter Europe
 Professional Spectro 1000
AQUANAL®
Introduction
Functional Description
Technical Data
Product List
Further Details:
Introduction
AQUANAL®-PROFESSIONAL SPECTRO 1000 is a spectrophotometer for water and waste water analysis with an wide range of applications. It is a modern single-beam spectrophotometer with an excellent price/performance ratio specifically designed for water testing.
The unit is equipped with a wide spectrum of pre-programmed methods based on the proven range of Vario H Powder Pack tests, tablet reagents and liquid reagents.
Functional Description
  • Approx. 60 pre-installed methods:
    -results can be stored (up to 760)
    -count down of colour development time
    -differentiation of results e.g. free, combined and total chlorine
  • Data transfer to a PC:
    -delivery includes CD-ROM with software
    -transfer of results and wavelength scans (nm/Abs)
  • Updates-delivery includes CD-ROM with software for:
    -download methods
    -download languages
  • Files necessary for updates are available on our website:
    -"parameters Vx" for new methods
    -"language option" for optional language
    -please note:
    During the process of method download all stored test results will be deleted. Stored customer-specific methods (polynomials or concentrations and the calibration of fluoride) remain stored. The file "parameters Vx" contains all methods (old and new). Old methods are overwritten.
  • Abs / Trans:
    -measurement at selected wavelength (330-900 nm)
    -reading in absorption-units and transmission simultaneously
    -photometric range from - 0.3 Abs to 2.5 Abs (0.3 - 200 % T)
  • Multi WL:
    -measurement at two different wavelengths (330-900 nm)
    -reading in absorption-units
    -photometric range from - 0.3 Abs to 2.5 Abs
  • Concentration:
    -storage of 4 user defined methods (linear relationship conc. / abs)
    -name, range and unit can be selected
    -measurement of min. 2 to max. 8 standards
    -linear relationship is calculated by the instrument (y = A + B x)
  • Polynome:
    -storage of 10 user defined methods
    -10 polynomials up to third order can be programmed (y = A + B x + C x2 + D x3)
    -name, range and unit can be stored
    -results can be stored
  • Spectrum:
    -wavelength scan from 330 to 900 nm
    -measurement range can be selected
    -graphic on the display
    -automatic calculation of peaks and valleys
    -transfer of data to a PC in 1 nm steps
  • Kinetic:
    -selectable delay time to start the measurement (0-240 sec)
    -selectable number of measurements per test (max. 25)
    -selectable interval between two readings
    -correlation to a defined standard or calculation using a factory
Technical Data
  • Wavelength range: 330 to 900 nm
  • Wavelength accuracy: ± 2 nm
  • Wavelength reproducibility: ± 1 nm
  • Spectral band width: 10 nm
  • Photometric range: -0.3 to 2.5 Abs (0.3 to 200 % T)
  • Photometric accuracy: ± 2 %
  • Photometric linearity: ± 0.005 Abs
  • Photometric noise level: max. of 0.5 x 10-3 at 330 nm
  • Stray light: 0.5 % bei 340 nm and 400 nm
  • Drift: 0.005 Abs/h at 500 nm
  • Drift correction: automatic on each self test / zero reading
  • Optical system: single beam spectrophotometer; verification / readjustment of the optical system after switching on (self test); didymium-filter (at 442, 476, 529, 584, 685, 744 nm)
  • Light source: pre-adjusted tungsten-halogen lamp (6 V, 10 W)
  • Lamp life: permanent light: 1000 h; flash light: about 200 000 measurements; no warming-up time necessary
  • Monochromator: holographic grating from Jobin Yvon
  • Detektor: silicon diode
  • Date and time: real time clock in the photometer
  • Operating conditions: 10 to 40°C (up to 90% relative humidity, without condensation)
  • Display: graphical display (9 lines / 21 signs); background illuminated
  • Dimensions: 275 x 340 x 165 mm (L x W x H)
  • Weight: approx. 3.2 kg
Back to top
Product List
Product No.
Product Name
Description
Add to Cart
70012
Spectro 1000 single-beam spectrometer (330-900 nm) suitable for all QUANAL®-PROFESSIONAL tests 16 and 24 mm cyclic tubes
10 to 50 mm rectangular tubes
70016
Light source replacement for Spectro 1000 preadjusted light source
70024
Cable connection (printer) RS232 C, D9F-D25M for Spectro 1000
70026
Cable connection (PC) RS232 C, D9F-D25F for Spectro 1000
70027
Cylindric cells 12 cylindric tubes diameter 24 mm
70028
Cylindric cells 5 cylindric tubes diameter 16 mm
70029
Rectangular cell 10 mm optic glas for Spectro 1000
70032
Rectangular cell 50 mm optic glas for Spectro 1000
70033
Rectangular cell 10 mm QUARTZ-UV glas for Spectro 1000
AQUANAL®-PROFESSIONAL REAGENTS 100 tablets/unit for Spectro 1000
70054
Alka-m-Photometer range: 5-200 mg/l
70056
Alka-p-Photometer range: 5-300 mg/l
70057
Ammonia no. 1 use with: Ammonium no. 2
range: 0.02-1 mg/l
70059
Ammonia no. 2 use with: Ammonium no. 1
range: 0.02-1 mg/l
70085
Copper no. 1 use with: Copper no. 2
range: 0.05-5 mg/l
70086
Copper no. 2 use with: Copper no. 1
range: 0.05-5 mg/l
70073
DEHA use with: DEHA solution
range: 0.02-0.5 mg/l
70075
DEHA solution use with: DEHA tablets
range: 0.02-0.5 mg/l
70061
DPD no. 1 determination of bromine, chlorine, chlorinedioxid and ozone
70062
DPD no. 1 High Calcium determination of bromine, chlorine and ozone
70064
DPD no. 3 determination of chlorine and ozone
70084
Hardcheck P / total hardness range: 2-50 mg/l
70076
Iron no. 1 LR use with: iron no. 2 LR
range: 0.01-1 mg/l
70077
Iron no. 2 LR use with: iron no. 1 LR
range: 0.01-1 mg/l
70087
Manganese LR 1 use with: manganese LR 2
range: 0.05-4 mg/l
70091
Molybdate no. 2 use with: molybdate no. 1
70245
Phosphate no. 1 LR use with: phosphate no. 2
range: 0.05-4 mg/l
70095
Phosphate no. 2 LR use with: phosphate no. 1
range: 0.05-4 mg/l
70102
Hydrogen peroxide LR range: 0.01-1.5 mg/l
70070
acidifying GP determination of chlorine
70083
Calibration standard fluoride use with: Spadns reagent
range: 0.02-1.5 mg/l
70071
Chloride range: 5-50 mg/l; liquid reagents
70068
Chlorine HR determination of chlorine
70103
Copper/Zinc LR use with: EDTA and Dechlorine
range: 0.02-1 mg/l
70104
Dechlorine use with: EDTA and copper/zinc
range: 0.02-1 mg/l
70063
DPD 1 Buffer solution determination of bromine, chlorine and chlorine dioxide
range: 0.05-4 mg/l Cl2
70066
DPD 1 Reagent solution determination of bromine, chlorine and chlorine dioxide
range: 0.05-4 mg/l Cl2
70067
DPD 3 Reagent solution determination of chlorine
range: 0.05-4 mg/l Cl2
70105
EDTA use with: copper/zinc and dechlorine
range: 0.02-1 mg/l
70093
Glycine use with: DPD no. 1, DPD n. 3 and DPD High Calcium
range: 0.02-1 mg/l
70106
Nickel range: 0.2-7 mg/l; liquid reagents
70094
Phenolred range: 6.5-8.4 mg/l
70096
Silica no. 1 use with: Silica no. 2
70097
Silica no. 2 use with: Silica no. 1
70081
SPADNS reagent use with: calibration fluoride
range: 0.02-1.5 mg/l
70098
Sulfide no. 1 use with: Sulfide no.2
range: 0.04-0.5 mg/l
70101
Sulfide no. 2 use with: Sulfide no.1
range: 0.04-0.5 mg/l
Back to top
Optics
The AQUANAL®-PROFESSIONAL SPECTRO 1000 is a single-beam spectral photometer (see illustration). The light source is a tungsten halogen lamp with flash function. The lamp is switched on only momentarily for the duration of the measurement process1), and there is therefore no need for a warm-up period. The AQUANAL®-PROFESSIONAL SPECTRO 1000 is ready to perform a self-test as soon as it is switched on. The light passes through an entry slot onto the monochromator, where it is split into spectral ranges. The monochromator is a holographically produced, transparent grating. The movable mirror ensures that light of the desired wavelength is automatically focused in such a way that it passes through the exit slot into the sample chamber and hence through the water sample. The light that is not absorbed by the sample travels to the detector of a silicon photodiode, evaluated by a microprocessor and shown as a result in the display. (Exception: permanent light is used for a wavelength scan).
Multifunctional Sample Chamber
Round vials measuring 16 mm and 24 mm in diameter and rectangular cells with path lengths from 10 to 50 mm may be used without an adapter.
Self-test
After it is switched on, the AQUANAL®-PROFESSIONAL SPECTRO 1000 automatically performs a self-test beginning with a function test of the stepping motor and the halogen lamp, followed by an optics test. For this purpose, the unit has a built-in didymium glass filter. This filter checks the correct wavelength setting. Finally, the memory functions are tested.
Maintenance
Thanks to the design of the AQUANAL®-PROFESSIONAL SPECTRO 1000 , the only maintenance that is required is replacement of the light source. The latter is situated on the rear of the photometer in an easily accessible position. Replacement is fast and simple and does not require any tools. The positioning of the assembly ensures optimum focusing of the halogen lamp.
Power supply for the AQUANAL®-PROFESSIONAL SPECTRO 1000
The required input voltage is 10 V. The AQUANAL®-PROFESSIONAL SPECTRO 1000 is connected to an external power pack as standard. Battery operation is also possible (see Accessories).
New Methods
The number and variety of test methods are continuously adjusted to suit market requirements. These new methods can be added easily and quickly using the supplied software. You can find updates for new methods and additional languages on this website.
Choice of Language
The user prompt in the display can be switched to German, English and French. An additional language can also be integrated. The required software is on the supplied diskette. Latin characters.
Printer/PC Connection
On the rear of the AQUANAL®-PROFESSIONAL SPECTRO 1000 photometer, there is an RS232 interface with a 9-pin D-Sub connector for connection of a PC or a printer with serial interface.
Printing Data
Every result is printed with date, time, reg. no, code no., measuring range and method number.
Storing Data
You can store results of programmed and user-specific methods (polynomes) in a memory with a capacity of 760 data sets. Alongside the result, the data sets contain information on method, date and time of the test.
User Prompt
The user prompt feature is highly convenient and easy to understand. The user prompt guides the reader step by step all the way through to the test result.
Zero Calibration and Measurements
You choose the desired method either from the list of chemical designations (e.g. Cl, COD, Cu) in alphabetical order or by entering a numerical code. An information picture then appears showing the required vial, the reagent type and the measuring range. The date and time are continuously shown in the display. The AQUANAL®-PROFESSIONAL SPECTRO 1000 automatically selects the correct wavelength.
Zero calibration is performed with the water sample by pressing the ZERO key. A characteristic coloration develops when you add the indicator to the water sample. Press the ENTER key to start measurement (which is effected either immediately or after expiry of the coloration time).
Countdown Function
With some methods, the user has to wait for a predefined colour reaction time after adding the indicator to the water sample. This time interval is shown in the display. The remaining time is continuously displayed. An acoustic signal is emitted during the last 10 seconds of the rundown time. Measurement is then started automatically, and the result is shown in the display.
Differentiation of Results
The AQUANAL®-PROFESSIONAL SPECTRO 1000 allows differentiated tests for certain methods. With the Chlorine method, for example, differentiated measurement is possible for free, combined and total chlorine.
Functions
The AQUANAL®-PROFESSIONAL SPECTRO 1000 is ideal for routine laboratory use and is equipped with additional functions for user-specific applications (for example, the creation of a user-defined method for a routine check).
Spectrum
A wavelength scan is performed in the user-defined interval between 330 and 900 nm. The display shows the graph of the spectrum; if the user presses a key, the display also shows a data list with the corresponding maximum and minimum absorption levels.
Kinetics
The Kinetics feature enables the user to obtain information on the development of the reaction over time. The delay time up to the start of the first measurement, the number of test points, and the interval between two measurements are set by the user in line with user requirements. All data points of the wavelength scan and the kinetics can be transferred to a PC for further evaluation.
Absorption/Transmission
Via this function, the user can - for example - effect measurement of standards with different concentrations using the user-selected wavelength in order to obtain the data pairs required for a polynome. Result output is in Abs and % Transmission.
Polynomes
The corresponding polynome is created from data pairs (concentration/absorption) with the help of an external mathematical program. A known polynome may also be used. Up to 10 polynomes of the third order (y = A+Bx+Cx2+Dx3) can be stored together with user-specific parameters such as method name, wavelength, measuring range and unit.
Concentration
This function can be used to measure 2 to 8 standards. On the basis of the obtained concentrations/absorption pairs, the photometer computes a linear equalisation curve that can be stored as a method for further sample measurements. Up to 4 methods can be stored.
Back to top
Photometers home
AQUANAL® home