Spectro 2 - Photometer for rapid water analysis |
 |
AQUANAL® |
 |
 |
| |
|
|
Concept
Spectro 2 photometer for rapid water analysis support your work with the AQUANAL®-plus Testkits.
The battery-powered photometer can be used in the field.
|
 |
Technical Data
- Microprocessor-controlled LED handphotometer
- Light sources: 480 nm, 565 nm, 585 nm and 635 nm
- Extinction range: 0.000 - 2.000
- Electro-optical precision: 1% ± 2 digits for extinction range 0.100 - 1.000
- test chamber for 16 mm tubes, adapter for 13 mm tubes
- Test result displayed in mg/l or in extinction mode
- 40 cations/anions available
- Automatic selection of the wave length
- Automatic blanc setting
- Comprehensive recognition of measurement faults
- Curent saving function, automatic power off
- Memory capacity for 120 calibration curves each with 11 calibration points
- Data memory for 50 readings/test results
- All data are protected from getting deleted
- Supply voltage: 4 x 1.5 V mignon cells or external stabilised 6 V DC (5.5 V to 6.5 V), (power voltage, for INDOOR USE only)
- External dimensions: 180 x 100 x 45 mm
- Weight: approx. 420 g
Ordering Information
|
Product #
|
Product
Name
|
Add to Cart |
| for your convenience we created separate product numbers for menues in the individual languages replacing the old product number 37568:
|
|
37781
|
Spectro 2 Photometer for AQUANAL®-plus reagents, Menue German |
|
|
37782
|
Spectro 2 Photometer for AQUANAL®-plus reagents, Menue English |
|
|
37783
|
Spectro 2 Photometer for AQUANAL®-plus reagents, Menue French |
|
|
37784
|
Spectro 2 Photometer for AQUANAL®-plus reagents, Menue Italian |
|
|
37785
|
Spectro 2 Photometer for AQUANAL®-plus reagents, Menue Spanish |
|
|
for your convenience we created separate product numbers for menues in the individual languages replacing the old product number 37569:
|
|
37786
|
Spectro 2 Photometer for AQUANAL®-plus reagents complete: in a transport case including software interface cable, batteries, tubes, etc., Menue German |
|
|
37787
|
Spectro 2 Photometer for AQUANAL®-plus reagents complete: in a transport case including software interface cable, batteries, tubes, etc., Menue English |
|
|
37788
|
Spectro 2 Photometer for AQUANAL®-plus reagents complete: in a transport case including software interface cable, batteries, tubes, etc., Menue French |
|
|
37789
|
Spectro 2 Photometer for AQUANAL®-plus reagents complete: in a transport case including software interface cable, batteries, tubes, etc., Menue Italian |
|
|
37790
|
Spectro 2 Photometer for AQUANAL®-plus reagents complete: in a transport case including software interface cable, batteries, tubes, etc., Menue Spanish |
|
|
37561
|
Refill pack tubes (5 pcs 16 mm diameter) |
|
Back to top
Photometers home
AQUANAL® home