Concept
We are offering Spectro CSB/COD within the
AQUANAL®-range to determine the Chemical Oxygen
demand (COD). There are four ready to use tube tests
available for this determination: AQUANAL®-plus COD
tube test: 2– 40 mg/L, 10 – 150 mg/L and 1500 –
15,000 mg/L COD.
|
 |
Technical Data of Spectro CSB/COD
- LED photometer (LED = light emitting diode)
- Optics: Low range: LED λ = 430 nm, medium and high range: LED λ = 605 nm
- 9 V block battery (life approx. 600 tests), mains operation not possible
- Ambient conditions: Temperature 5 – 40°C, relative humidity 30 – 90%, non-condensing
- Dimensions: D x W x H = 180 x 110 x 100 mm, supplied in a case
To guarantee ideal conditions for the chemical reaction
AQUANAL®-plus Thermoblock can be used:
Technical Data of AQUANAL®-plus Thermoblock
- Temperature: 100°C and 148°C fixed, can be selected via a membrane keyboard
- Tolerance: ± 1°C
- Heating: 300 W, electronically regulated, with protection against overheating
- Heating block: Eight wells for tubes of 16 mm diameter, 1 well in the centre for tubes of 22 mm diameter
- Time input: 30, 60, 120 minutes or continuous operation at the selected temperature, automatically switched off at the end of the selected reaction time,
acoustic signal
- Casing: Round metal block with epoxide-coated metal casing
- Dimensions: D x W x H = 214 x 1343 x 110 mm, supplied in a cardboard box
- Weight: 2.15 kg
- Mains connection: 230 V, 50 Hz (350 W) or 110 V, 60 Hz (on request)
- Plug: Euroformat
- Safety: CE marking, transparent covering hood
Ordering Information
|
Catalog No.
|
Product
|
Add to Cart |
|
37735
|
AQUANAL®-plus COD tube tests, 2 - 40 mg/L |
|
|
37736
|
AQUANAL®-plus COD tube tests, 15 - 150 mg/L |
|
|
37737
|
AQUANAL®-plus COD tube tests, 100 - 1500 mg/L |
|
|
37738
|
AQUANAL®-plus COD tube tests, 1000 - 15,000 mg/L |
|
|
37570
|
Spectro CSB/COD for AQUANAL®-plus COD tube tests |
|
|
37571
|
AQUANAL®-plus Thermoblock |
|
|
70044
|
Sample chamber lid for Spectro Turbidity |
|
Back to top
Photometers home
AQUANAL® home
|