Analytical / Chromatography
Air Monitoring
Analytical Reagents
AQUANAL
Compact Labs
Dealership
LabInfos
Literature Request
Photometers
Prof. Reagent Tests
Test Kits
Test Sticks
Thermoblock
Tube Tests
Vario H Powder Packs
Waste Water Analysis
Analytic basic reagents
Derivatization Reagents
Electrophoresis
Sensoric Applications
Solvents
Trace Analysis
Water Analysis
Analytical Standards
Gas Chromatography
HPLC
Microbiology
Microscopy
Sample Preparation
Spectroscopy
Syringes
Titration
TLC
Vials
Videos
Events
Literature
Analytix
The Reporter
Reporter Europe
 Quantofix® Test Sticks
AQUANAL®
Quantofix® Test Sticks
AQUANAL Image
Quantofix Test Sticks Image
Concept
Quantofix® test sticks serve for semi-quantitative determination of ions and molecules. The benefits of using Quantofix® test sticks to carry out a determination are evident: it is quick -dip the test stick and read the result- and easy -no further equipment needed. A number of determinations require the use of additional reagents which are enclosed with the test sticks in a single kit. Instructions for use are enclosed as well. The test sticks themselves are about 0.2 mm thick and made of plastic; each pack allows 100 tests to be carried out. Our product range includes the following tests:
Test
Measuring Range
Graduation of
Color Scale in mg/L
Color Reaction
Product No.
Aluminium
0-500 mg/l Al3+
0-5-20-200-500
pink --> red
37207
Ammonium
0-400 mg/l NH4+
0-10-25-50-100-200-400
yellow --> orange
37212
Arsenic
0 - 0.5 mg/l As3+/5+
0 - 0.01 - 0.025 - 0.05 - 0.1 - 0.5
white --> yellowish-brown
37927
Ascorbic Acid
0-2000 mg/l Vitamin C
0-5-100-200-300-500-1000-2000
yellow --> bluish green
37203
Chlorine
0-100 mg/l Cl2
0-1-3-10-30-100
white --> blue
37208
Chromate
0-100 mg/l CrO42-
0-3-10-30-100
pink --> violet
37209
Cobalt
0-1000 mg/l Co2+
0-10-25-50-100-250-500-1000
white --> greenish blue
37199
Copper
0-300 mg/L Cu2+
0-10-30-100-300
white --> red violet
37198
Fluoride
0-100 mg/l F-
0–2–5–10–20–50–100
red --> colorless
37211
Nickel
0-1000 mg/l Ni2+
0-10-25-50-100-250-500-1000
white --> red
37197
Nitrate (Nitrite)
0-500 mg/l NO3-
Nitrate: 0-10-25-50-100-250-500
white --> pink
37204
0-80 mg/l NO2-
Nitrite: 0-1-5-10-20-40-80
Nitrite
0-50 mg/l NO2-
0-1-5-10-25-50
white --> pink
37205
Peroxide
0-100 mg/l H2O2
0-1-3-10-30-100
white --> blue
37206
Phosphate
0-100 mg/l PO43-
0-3-10-25-50-100
white --> greenish blue
37210
Potassium
0-1500 mg/l K+
0-200-400-700-1000-1500
yellow --> orange
37202
Silver (Ag-Fix)
0-10 g/l Ag+
0-0.5-1-2-3-7-10
ochreous --> brown
37196
pH value 4-8
pH value: 4-5-6-7-8
yellow --> blue
Sulfate
0-1600 mg/l SO42-
0-200-400-800-1200-1600
red --> yelow
37213
Sulfite
0-1000 mg/l SO32-
0-10-25-50-100-250-500-1000
white --> pink
37201
Tin
0-500 mg/l Sn2+
0-10-25-50-100-250-500
white --> blue
37195
Water Hardness - Aquadur®
3-21°dH
0 – 4 – 7 – 14 – 21 °d
turquoise --> pink
37194
Back to top
AQUANAL® home