Analytical / Chromatography
Air Monitoring
Analytical Reagents
AQUANAL
Compact Labs
Dealership
LabInfos
Literature Request
Photometers
Prof. Reagent Tests
Test Kits
Test Sticks
Thermoblock
Tube Tests
Vario H Powder Packs
Waste Water Analysis
Analytic basic reagents
Derivatization Reagents
Electrophoresis
Sensoric Applications
Solvents
Trace Analysis
Water Analysis
Analytical Standards
Gas Chromatography
HPLC
Microbiology
Microscopy
Sample Preparation
Spectroscopy
Syringes
Titration
TLC
Vials
Videos
Events
Literature
Analytix
The Reporter
Reporter Europe
 Professional Vario H Powder Packs
AQUANAL®
Introduction
Use
Chlorine, Free and Total
Product List
Introduction
Vario H Powder Packs are designed to be a like for like chemistry replacing Hach® powder pillows.
The use of dry powdered reagents minimises leakage and deterioration problems. Powders are packed in individual, pre-measured foil packs. Each pack contains enough reagent for one test ensuring identical test results compared to Hach® Powder pillows.
Use of Vario H Powder Packs
Back to top
Chlorine, free and total
  • Free Chlorine
    Free Chlorine (Hypochlorous Acid and Hypochlorite Ion) oxidizes DPD. The reactions is pH dependent. DPD and a buffer system (pH 6.3-6.5) cause a magenta colour in the presence of Free Chlorine. Both, our Vario H Powder and Hach® Powder handle high levels of hardness without precipitation.
  • Total Chlorine
    Potassium Iodide is used to determine combined chlorine forms. Chloramines oxidize the iodide to iodine; the liberated iodine reacts with DPD. Both, Sigma-Aldrich Vario H Powder and Hach® Powder contain DPD, Potassium Iodide and a buffer system.
Back to top
Product List
Product #
Range
Product Name
HACH® Prod.No.
Add to Cart
Aluminium
70436
0-0.22 mg/l Al AQUANAL®-professional Vario H Aluminium Set 26037-00
70447
  AQUANAL®-professional Vario H Aluminium ECR, refill pack for 70436 26038-49
70449
  AQUANAL®-professional Vario H Aluminium Hexamethylenetetramine buffer, refill pack for 70436 26039-99
70451
  AQUANAL®-professional Vario H Aluminium ECR Masking Reagent, refill pack for 70436 23801-23
Chlorine, free
70112
0.01-2 mg/l Cl2 AQUANAL®-professional Vario H Chlorine Free (DPD) for 10 ml sample 21055-69
70113
0-5 mg/l AQUANAL®-professional Vario H Chlorine Free (DPD), for 25 ml sample 14070-99
Chlorine, total
70200
0.01-2 mg/l Cl2 AQUANAL®-professional Vario H Chlorine Total (DPD) for 10 ml sample 21056-69
70202
0-5 mg/l Cl2 AQUANAL®-professional Vario H Chlorine Total (DPD), for 25 ml sample 14064-99
Copper
70203
0-5 mg/l Cu AQUANAL®-professional Vario H Copper I 21058-69
Iron, total
70204
0-1.8 mg/l Fetot AQUANAL®-professional Vario H Iron (TPTZ) for 10 ml sample 26087-99
70205
0-3 mg/l Fetot AQUANAL®-professional Vario H Iron (Phenantroline), for 10 ml sample 21057-69
Manganese LR
70441
0-0.7 mg/l Mn AQUANAL®-professional Vario H Manganese Set, low range 26517-00
70443
  AQUANAL®-professional Vario H Manganese Alkaline Cyanide, refill pack for 70441 21057-69
70445
  AQUANAL®-professional Vario H Manganese Ascorbic Acid, refill pack for 70441 14577-99
70446
  AQUANAL®-professional Vario H Manganese PAN, refill pack for 70441 21224-26
Manganese HR
70206
0-20 mg/l Mn AQUANAL®-professional Vario H Manganese Set, high range 24300-00
70208
  AQUANAL®-professional Vario H Manganese Citrate Buffer, refill pack for 70206 21076-69
70207
  AQUANAL®-professional Vario H Manganese Sodiumperiodate, refill pack for 70206 21077-69
Molybdate HR
70209
0-35 mg/l Mo AQUANAL®-professional Vario H Molybdate Set, high range for 10 ml sample 26041-00
70212
  AQUANAL®-professional Vario H Moly 1, refill pack for 70209 26042-99
70213
  AQUANAL®-professional Vario H Moly 2, refill pack for 70209 26043-99
70217
  AQUANAL®-professional Vario H Moly 3, refill pack for 70209 26044-99
70426
  AQUANAL®-professional Vario H Molybdate Set, high range for
25 ml sample
22434-00
70427
  AQUANAL®-professional Vario H Molybdate 1, refill pack for 70426 14146-69
70428
  AQUANAL®-professional Vario H Molybdate 2, refill pack for 70426 14148-69
70429
  AQUANAL®-professional Vario H Molybdate 3, refill pack for 70426 14178-69
Nitrate
70431
0-30 mg/l NO3-N AQUANAL®-professional Vario H Nitrate Set, high range 26053-45
Nitrite LR
70219
0-0.3 mg/l NO2-N AQUANAL®-professional Vario H Nitrite for 25 ml sample 14065-99
70218
0-0.3 mg/l NO2-N AQUANAL®-professional Vario H Nitrite for 10 ml sample 21071-69
Nitrogen, Ammonia LR
70434
0-2.5 mg/l NH3-N AQUANAL®-professional Vario H Ammonia Set, low range 26045-45
70222
  AQUANAL®-professional Vario H Ammonia Cyanurate, refill pack for 70434 and 70423 23954-66
70223
  AQUANAL®-professional Vario H Ammonia Salicylate, refill pack for 70434 and 70423 23952-66
70435
  AQUANAL®-professional Vario H Ammonia Diluent Reagent, refill pack for 70434 n.a.
70226
0-0.5 mg/l NH3-N AQUANAL®-professional Vario H Ammonia Set, low range 26680-00
70227
  AQUANAL®-professional Vario H Ammonia Cyanurate, refill pack for 70226 26531-99
70228
  AQUANAL®-professional Vario H Ammonia Salicylate, refill pack for 70226 26532-99
Nitrogen, Ammonia HR
70423
0-50 mg/l NH3-N AQUANAL®-professional Vario H Ammonia Set, high range 26069-45
70222
  AQUANAL®-professional Vario H Ammonia Cyanurate, refill pack for 70434 and 70423 23954-66
70223
  AQUANAL®-professional Vario H Ammonia Salicylate, refill pack for 70434 and 70423 23952-66
70424
  AQUANAL®-professional Vario H Ammonia Diluent Reagent, refill pack for 70424 n.a.
Nitrogen, Total LR
70411
0-25 mg/l N AQUANAL®-professional Vario H Nitrogen Set, low range 26722-45
Nitrogen, Total HR
70412
10-150 mg/l N AQUANAL®-professional Vario H Nitrogen Set, high range 27141-00
o-Phosphate
70229
0-2.5 mg/l PO4 AQUANAL®-professional Vario H Phosphate for 10 ml sample 21060-69
Silica LR
70417
0-1.6 mg/l SiO2 AQUANAL®-professional Vario H Silica Set, low range 24593-00
70418
  AQUANAL®-professional Vario H Silica Amino Acid F, refill pack for 70417 22540-69
70236
  AQUANAL®-professional Vario H Silica Citric Acid, refill pack for 70232 and 70417 21062-69
70419
  AQUANAL®-professional Vario H Silica Molybdate 3, refill pack for 70417 1995-26
Silica HR
70232
0-100 mg/l SiO2 AQUANAL®-professional Vario H Silica Set, high range 24596-00
70235
  AQUANAL®-professional Vario H Silica Acid RGT, refill pack for 70232 21074-69
70236
  AQUANAL®-professional Vario H Silica Citric Acid, refill pack for 70232 and 70417 21062-69
70237
  AQUANAL®-professional Vario H Silica Molybdate, refill pack for 70232 21073-69
Silica UHR
70239
0-200 mg/l SiO2 AQUANAL®-professional Vario H Silica Set, ultra high range 22443-00
70241
  AQUANAL®-professional Vario H Silica Acid RGT, refill pack for 70239 1042-99
70242
  AQUANAL®-professional Vario H Silica Citric Acid, refill pack for 70239 14548-99
70243
  AQUANAL®-professional Vario H Silica Molybdate, refill pack for 70239 1041-99
Sulfate
70244
0-70 mg/l SO4 AQUANAL®-professional Vario H Sulfate for 25 ml sample 12065-99
70421
  AQUANAL®-professional Vario H Sulfate for 10 ml sample 21067-69
Hach® is a registered trademark of the Hach Company
Back to top
AQUANAL® home