|
Most results of chemical analytical measurements are directly related to reference materials which are
used for the calibration of the measurement process. Two decisive preconditions must be fulfilled in order to obtain a result which is
traceable to the SI. First, the used reference material itself must carry an SI-traceable value and an attached uncertainty. Second, the
whole measurement procedure - from sampling to calculation of the result - must be fully validated and the uncertainty must be
evaluated according to common rules (GUM, Eurachem guide). While the analyst has to validate his procedure and to evaluate his
measurement uncertainty by himself, he may take it for granted, that the value which is declared on the label of the reference material
is traceable to the SI. Thus, the reference material plays a most important role as it serves as a transfer standard in the traceability
chain.
Reference materials are available from a large number of producers. Among them are private companies, federal research
laboratories, metrology institutes and others. ISO has set up several rules to assure a suitable quality of reference materials which
should be clearly indicated in a certificate.
Table 1
|
ISO Guide Number
|
Content
|
|
17025
|
General requirements for the competence of testing and calibration laboratories |
|
30 (1992)
|
Terms and definitions used in connection with reference materials |
|
31 (1996)
|
Contents of certificates of reference materials |
|
32 (1997)
|
Calibration of chemical analysis and use of certified reference materials |
|
33 (1989)
|
Use of certified reference materials (under revision) |
|
34 (1996)
|
Quality system guidelines for the production of reference materials |
|
35 (1989)
|
Certification of reference materials - General and statistical principles (under revision) |
There also have been several discussions on how the traceable value has to be assigned to the reference material and who is in
charge of this duty. However, there is still no valid guideline at present, which is generally accepted. In fact, one has to face a
contradictory situation today. On the one hand, there is a growing demand for reference materials which carry traceable values. On
the other hand, assigning a traceable value means putting much effort in a primary measurement which at least for a private producer
will often be in contrary to economical requirements. Thus it is by far not self-evident, that all the produced reference materials carry
traceable values.
As the need for reference materials will keep growing, a division of tasks may be the most suitable way, where different institutes
and companies focus on their different skills and core activities. While a private company may be able to produce a reference
material most efficient in a large scale, the assignment of a traceable value will not be a special skill as this was not explicitely
required in the past. So far, the declared value was simply regarded as the truth by most analysts. In contrary to that, federal institutes
and among them especially the metrological institutes are predestinated to assign a traceable value, because normally it is one of
their core tasks to provide the ultimative link to the SI by performing a primary measurement. However, they often may not be
equipped well enough for an efficient production. Regarding the large bunch of reference materials that are already available, one
could state, that there is already enough production capacity. There is, however, a lack of capacity for a serious value assignment.
As a consequence, it would be more effective to combine the different skills in a complementary way.
The way described above has been chosen by EMPA and FLUKA GmbH. From the beginning, BAM (Federal Institute for
Materials Research and Testing) was involved as partner. A suitable quality assurance system was required from each partner. An
ISO 9001/2000 certification is in place at EMPA and FLUKA. BAM and EMPA are accredited according to ISO/IEC 17025. After
having agreed to the criteria of EMPA for the value assignment, FLUKA produces the reference material according to ISO guide 34
and eventually further requirements defined by EMPA. EMPA controls the batch for its homogeneity and approximate content before
the material is released for bottling and packaging. The certification analysis is performed by EMPA and BAM using randomly
chosen bottles. Two different methods of measurement are applied. The results are combined to the certified value. Long term
stability tests are carried out by EMPA in parallel. A certificate is provided only when all criteria including stability are met.
Each certified standard is accompanied with a certificate which states:
- The certified property values and their uncertainty
- The primary methods of analysis used for certification
- The intended use, storing and handling instructions, the expiry date
And a certification report whicht states:
- analysis report of EMPA and BAM
- the ion standard solutions and the ion chromatography standard solutions the amount of trace impurities
- the titrimetric standards the homogeneity report
Label of each bottle contents:
- Fluka catalogue number
- Fluka product name
- Certification number
- Batch number
- Expiry date
- Certified property value and the confidence limit
| Table 2: |
These anionic standard solutions for Ion Chromatography (1000 mg/L) are
certified in a cooperation project with BAM and Sigma-Aldrich |
Catalog No. |
Product Name |
Produced from |
Example Certificate |
|
87969
|
Bromide |
NaBr |
|
|
87603
|
Chloride |
NaCl |
|
|
80373
|
Fluoride |
NaF |
|
|
86576
|
Nitrate |
NaNO3 |
|
|
81193
|
Phosphate |
Na2HPO4 |
|
|
80218
|
Sulfate |
Na2SO4 |
|
| These certified standards are prepared with high-purity salts and water and are supplied in 100 mL HDPE bottles. |
For our range of Certified Ion Chromatography Standards please click here.
Back to Top
Table 3. Titrimetric Substances
|
Catalog No.
|
Product Name
|
Use
|
Assay
|
Cas No.
|
Package Size
|
|
17971
|
Arsenic trioxide |
Reductometric standard |
~ 99.5%
|
1327-53-3
|
10 g
|
|
12353
|
Benzoic acid |
Acidimetric standard |
~ 99.5%
|
65-85-0
|
25 g
|
|
15194
|
Lead nitrate |
Complexometric standard |
~ 99.5%
|
10099-74-8
|
25 g
|
|
21067
|
Calcium carbonate |
Complexometric standard |
~ 99.5%
|
471-34-1
|
25 g
|
|
60357
|
Potassiumhydrogenphthalate |
Acidimetric standard |
~ 99.5%
|
877-24-7
|
50 g
|
|
60386
|
Potassiumiodate |
Reductometric standard |
~ 99.5%
|
7758-05-6
|
50 g
|
|
71363
|
Sodium carbonate |
Basimetric standard |
~ 99.5%
|
497-19-8
|
50 g
|
|
71387
|
Sodium chloride |
Argentometric standard |
~ 99.5%
|
7647-14-5
|
25 g
|
|
71804
|
Sodium oxalate |
Reductometric standard |
~ 99.5%
|
62-76-0
|
25 g
|
|
93440
|
Tris hydroxymethyl aminomethane |
Basimetric standard |
~ 99.5%
|
77-86-1
|
50 g
|
Back to top
Back to Certified Reference Material home
Trace Analysis home
Back to Analytical home
|