Analytical / Chromatography
Air Monitoring
Analytical Reagents
Analytical Standards
Gas Chromatography
HPLC
Microbiology
Microscopy
Buffer Substances
Dehydration Agents
Embedding Media
Fixation Agents
Microphoto Award
Solvents
Stains and Dyes
Surface Replication
Various Chemicals
Sample Preparation
Spectroscopy
Syringes
Titration
TLC
Vials
Videos
Events
Literature
Analytix
The Reporter
Reporter Europe
 Embedding and Mounting Media
Microscopy
Fluka is pleased to offer an extended range of embedding media kits and ready-made mounting media for both light and electron microscopy. Due to the increasing importance of fluorescence microscopy non-fluorescent aqueous mounting media that delay fluorescence fading have been introduced. PVA-NPD is the medium of choice for AMCA or Cascade blue-labeled tissue sections and cell cultures. Either PVA-NPD or PVA-DABCO are recommended for FITC, TRITC, DTAF, Cy-3 labeled sections or cell cultures.
Product list of Embedding and Mounting Media
Back to top
Back to Microscopy home
Back to Analytical home
Back to Bioscience home