|
Residue Analysis
Vetranal Standards
|
|
|
Nitrofuran Antibiotics Detailed Product List |
|
Nitrofuran Antibiotics
During an investigation early in 2002, it was discovered that residues of nitrofuran antibiotics occurred in fish products like shrimp and catfish originating from South-East Asian countries. Later on, intensified control revealed that the same residues could also be found in poultry, duck and rabbit imported from this region.
Since the discovery of the occurrence of residues of nitrofurans and their metabolites (AOZ (3-Amino-2-oxazolidinone), AMOZ (5-Methylmorpholino-3-amino-2-oxazolidinone), AHD (1-Aminohydantoin hydrochloride) and SEM (Semicarbazide hydrochloride), numerous scientists have contacted Sigma-Aldrich and asked for analytical standards. In 2003, we launched following 4 standards:
|
| Abbreviation |
Name |
Catalog No. |
|
AOZ
|
(3-Amino-2-oxazolidinone) |
|
|
AMOZ
|
(5-Methylmorpholino-3-amino-2-oxazolidinone): |
|
|
AHD
|
(1-Aminohydantoin hydrochloride) |
|
|
SEM
|
(Semicarbazide hydrochloride) |
|
Afterwards, many customers asked if we can provide also the derivatized and isotope- labelled standards. In order to provide the best and most convenient analytical solution we have developed 7 more Nitrofuran Standardswhich will allow you to reduce the analysis time significantly:
Nitrofuran standards derivatized with 2-nitrobenzaldehyde which are most common standards used in nitrofuran analysis. |
| Abbreviation |
Name |
Catalog No. |
|
2-NP-AOZ
|
(3-(2-Nitrobenzylidenamino)-2-oxazolidinone) |
|
|
2-NP-SCA
|
(2-Nitrobenzaldehyde semicarbazone) |
|
|
2-NP-AMOZ
|
(5-(Morpholinomethyl)-3-(2-nitrobenzylidenamino)-2-oxazolidinone) |
|
|
2-NP-AHD
|
(3-(2-Nitrobenzylidenamino)-2,4-imidazolidinedione) |
|
| For scientists performing nitrofuran analysis by LC-MS we are pleased to provide 3 unique isotope marked standards: |
| Abbreviation |
Pack-size |
Catalog No. |
|
AOZ-D4
|
10mg
|
|
|
AMOZ-D5
|
10mg
|
|
|
SCA-HCL (13C, 15N)
|
10mg
|
|
|
References: "Determination of the metabolites of nitrofuran antibiotics in animals tissues by HPLC-tandem mass spectrometry", A. Leitner, P. Zvllner, W. Lindner, J. Chromatogr. A, 939 (2001) 49-58.
"Analysis of protein-bound metabolites of furazolidone and furaltadone in pig liver by high performance liquid chromatography and liquid chromatography-mass spectrometry.", E. Horne, A. Cadogan, M. O'Keeffe and L.A.P. Hoogenboom, The Analyst, 121 (1996) 1463-1468.
"The use of pig hepatocytes to study the nature of protein-bound metabolites of furazolidone; a new analytical method for their detection.", L.A.P. Hoogenboom, M. van Kammen, M.C.J. Berghmans, J.H. Koeman and H.A. Kuiper, Food and Chemical Toxicology, 29 (1991) 321-328.
Back to Top
Detailed Product List
|
| Prod. No. |
Name |
Application |
Pack Size |
| 46060 |
Amoxicillin, trihydrate |
antibiotic |
250 mg |
| 46006 |
Amphotericin B, trihydrate |
antibiotic |
100 mg |
| 46061 |
Ampicillin, trihydrate |
antibiotic |
250 mg |
| 46062 |
Amprolium hydrochloride |
|
250 mg |
| 46024 |
Androstanolone |
hormone |
250 mg |
| 46025 |
Androstanolone 17-benzoate |
hormone |
250 mg |
| 46033 |
Androstenedione |
hormone |
250 mg |
| 46041 |
Androsterone |
hormone |
250 mg |
| 46042 |
Androsterone 3-acetate |
hormone |
100 mg |
| 46063 |
Arecoline hydrobromide |
|
250 mg |
| 46059 |
4-Arsanilic acid |
|
250 mg |
| 46084 |
Bacitracin A |
antibiotic |
250 mg |
| 46610 |
Benzathine penicillin G, tetrahydrate |
antibiotic |
250 mg |
| 46074 |
Betamethasone 17-valerate |
hormone |
250 mg |
| 46083 |
Bithionol |
|
250 mg |
| 46090 |
Boldenone |
hormone |
250 mg |
| 46091 |
Boldenone 17-acetate |
hormone |
100 mg |
| 46100 |
Carbazole |
phenothiazine |
250 mg |
| 46110 |
Chloramphenicol |
antibiotic |
250 mg |
| 46109 |
Chloramphenicol palmitate |
antibiotic |
250 mg |
| 46116 |
Chlormadinone 17-acetate |
hormone |
100 mg |
| 46125 |
Chlorpromazine hydrochloride |
phenothiazine |
250 mg |
| 46133 |
Chlortetracycline hydrochloride |
antibiotic |
250 mg |
| 46140 |
Cloxacillin sodium salt, hydrate |
antibiotic |
250 mg |
| 46148 |
Corticosterone |
hormone |
100 mg |
| 46149 |
Corticosterone 21-acetate |
hormone |
100 mg |
| 46364 |
Crystal violet chloride |
|
250 mg |
| 46158 |
Dapsone |
|
250 mg |
| 46161 |
Demeclocycline hydrochloride, hemihydrate |
antibiotic |
100 mg |
| 46165 |
Dexamethasone |
hormone |
250 mg |
| 46166 |
Dexamethasone 21-acetate |
hormone |
250 mg |
| 46174 |
Diaveridine |
diaminopyridine |
100 mg |
| 46182 |
Dicloxacillin sodium salt, hydrate |
antibiotic |
100 mg |
| 46190 |
Dienestrol |
hormone |
250 mg |
| 46191 |
Dienestrol diacetate |
hormone |
250 mg |
| 46200 |
Diethylcarbamazine citrate |
|
250 mg |
| 46207 |
Diethylstilbestrol |
hormone |
250 mg |
| 46224 |
Dimetridazole |
imidazole |
250 mg |
| 46232 |
Dipyrone, hydrate |
|
250 mg |
| 46239 |
Equilenin |
hormone |
100 mg |
| 33693 |
Equilenin Solution |
hormone |
2 ml |
| 46247 |
Equilin |
hormone |
100 mg |
| 46256 |
Erythromycin A, dihydrate |
antibiotic |
250 mg |
| 46551 |
17b-Estradiol 17-acetate |
hormone |
100 mg |
| 46552 |
17b-Estradiol 3-benzoate |
hormone |
250 mg |
| 46553 |
17b-Estradiol 3,17-diacetate |
hormone |
100 mg |
| 46554 |
17b-Estradiol 3,17-dipropionate |
hormone |
250 mg |
| 46542 |
17a-Estradiol, hemihydrate |
hormone |
100 mg |
| 46550 |
17b-Estradiol, hemihydrate |
hormone |
250 mg |
| 46555 |
17b-Estradiol 3-methyl ether |
hormone |
250 mg |
| 46556 |
17b-Estradiol 17-propionate |
hormone |
250 mg |
| 46557 |
17b-Estradiol 17-valerate |
hormone |
250 mg |
| 46565 |
Estriol |
hormone |
100 mg |
| 46573 |
Estrone |
hormone |
250 mg |
| 46263 |
17a-Ethinylestradiol |
hormone |
250 mg |
| 46272 |
Ethisterone |
hormone |
250 mg |
| 46280 |
Etiocholanolone |
hormone |
100 mg |
| 35032 |
Fenbendazole |
|
100 mg |
| 46289 |
Furaltadone |
|
250 mg |
| 46297 |
Furazolidone |
|
250 mg |
| 46305 |
Gentamycin 2,5-sulfate, hexahydrate |
antibiotic |
250 mg |
| 46309 |
Griseofulvin |
|
250 mg |
| 46313 |
Hexachloroethane |
|
250 mg |
| 46320 |
Hexestrol |
hormone |
250 mg |
| 46328 |
Hydrocortisone |
hormone |
250 mg |
| 46329 |
Hydrocortisone 21-acetate |
hormone |
250 mg |
| 46337 |
17a-Hydroxyprogesterone |
hormone |
250 mg |
| 46346 |
21-Hydroxyprogesterone |
hormone |
250 mg |
| 46338 |
17a-Hydroxyprogesterone 17-acetate |
hormone |
250 mg |
| 46347 |
21-Hydroxyprogesterone 21-acetate |
hormone |
250 mg |
| 46356 |
Kanamycin A disulfate, dihydrate |
antibiotic |
250 mg |
| 46372 |
Lasalocid A sodium salt |
|
250 mg |
| 46380 |
Leucomycin, hydrate |
antibiotic |
250 mg |
| 46944 |
Levamisole hydrochloride |
imidazole |
250 mg |
| 46387 |
Lincomycin hydrochloride, hydrate |
antibiotic |
250 mg |
| 46396 |
Malachite green oxalate |
|
250 mg |
| 46404 |
Mebendazol |
|
250 mg |
| 46411 |
Medroxyprogesterone |
hormone |
100 mg |
| 46412 |
Medroxyprogesterone 17-acetate |
hormone |
250 mg |
| 46420 |
Megestrol 17-acetate |
hormone |
100 mg |
| 46264 |
Mestranol |
hormone |
250 mg |
| 46429 |
Methimazole |
|
250 mg |
| 46436 |
6a-Methylprednisolone |
hormone |
100 mg |
| 46444 |
17a-Methyltestosterone |
hormone |
250 mg |
| 46453 |
6-Methyl-2-thiouracil |
thiouracil |
250 mg |
| 46461 |
Metronidazole |
imidazole |
250 mg |
| 46468 |
Monensin sodium salt, hydrate |
antibiotic |
100 mg |
| 46473 |
Morantel tartrate |
|
250 mg |
| 46476 |
Nandrolone |
hormone |
250 mg |
| 46477 |
Nandrolone 17-propionate |
hormone |
100 mg |
| 46482 |
Naproxen |
|
250 mg |
| 46486 |
Neomycin B trisulfate sesquihydrate |
|
250 mg |
| 46494 |
Nifuroxazide |
nitrofurane |
250 mg |
| 46502 |
Nitrofurantoin |
nitrofurane |
250 mg |
| 46208 |
Nitrofurazon solution, 100 ng/ml, in acetonitrile |
|
2 ml, 10 ml |
| 46518 |
Nitromide |
|
250 mg |
| 46525 |
19-Norethisterone |
hormone |
250 mg |
| 46527 |
19-Norethisterone 17-acetate |
|
100 mg |
| 46531 |
Novobiocin sodium salt |
|
250 mg |
| 46535 |
Nystatin A1, trihydrate |
antibiotic |
250 mg |
| 46582 |
Oleandomycin phosphate, dihydrate |
antibiotic |
250 mg |
| 46589 |
Oxacillin sodium salt, hydrate |
antibiotic |
100 mg |
| 46598 |
Oxytetracycline hydrochloride |
antibiotic |
250 mg |
| 46609 |
Penicillin G potassium salt |
antibiotic |
250 mg |
| 46616 |
Penicillin V potassium salt |
antibiotic |
250 mg |
| 46624 |
Phenothiazine |
|
250 mg |
| 46901 |
N4-Phthalylsulfathiazole |
sulfonamide |
250 mg |
| 46632 |
Piperazine citrate, pentahydrate |
|
250 mg |
| 46639 |
Polymyxin B 2,5-sulfate, pentahydrate |
antibiotic |
100 mg |
| 46648 |
Praziquantel |
|
250 mg |
| 46656 |
Prednisolone |
hormone |
250 mg |
| 46657 |
Prednisolone 21-acetate |
hormone |
250 mg |
| 46608 |
Procaine hydrochloride |
|
250 mg |
| 46665 |
Progesterone |
hormone |
250 mg |
| 46674 |
Promazine hydrochloride |
|
250 mg |
| 46682 |
Promethazine hydrochloride |
|
250 mg |
| 46689 |
Propionylpromazine hydrochloride |
|
100 mg |
| 46698 |
6-Propyl-2-thiouracil |
|
250 mg |
| 46702 |
Pyrantel pamoate |
|
250 mg |
| 46706 |
Pyrimethamine |
|
250 mg |
| 46713 |
Rifamycin AMP |
antibiotic |
100 mg |
| 46721 |
Rifamycin SV sodium salt, hexahydrate |
antibiotic |
100 mg |
| 46724 |
Roxarsone |
|
250 mg |
| 46725 |
Salbutamol |
|
100 mg |
| 46732 |
Salbutamol hemisulfate |
|
250 mg |
| 46729 |
Salinomycin SV sodium salt, 2,5-hydrate |
antibiotic |
100 mg |
| 46738 |
Spectinomycin dihydrochloride, pentahydrate |
antibiotic |
250 mg |
| 46745 |
Spiramycin (I) |
antibiotic |
100 mg |
| 46754 |
Streptomycin sesquisulfate |
antibiotic |
250 mg |
| 46900 |
N4-Succinylsulfathiazole, monohydrate |
sulfonamide |
250 mg |
| 46762 |
Sulfabenzamide |
|
250 mg |
| 46770 |
Sulfacetamide |
|
250 mg |
| 46778 |
Sulfachlorpyridazine |
sulfonamide |
250 mg |
| 35033 |
Sulfadiazine |
|
100 mg |
| 46794 |
Sulfadimethoxine |
sulfonamide |
250 mg |
| 46802 |
Sulfadimidine |
sulfonamide |
250 mg |
| 46810 |
Sulfadoxine |
|
250 mg |
| 46818 |
Sulfaguanidine, monohydrate |
sulfonamide |
250 mg |
| 46826 |
Sulfamerazine |
sulfonamide |
250 mg |
| 46834 |
Sulfameter |
sulfonamide |
250 mg |
| 46842 |
Sulfamethizole |
sulfonamide |
250 mg |
| 46850 |
Sulfamethoxazole |
sulfonamide |
250 mg |
| 46858 |
Sulfamethoxypyridazine |
sulfonamide |
250 mg |
| 46866 |
Sulfamoxole |
sulfonamide |
250 mg |
| 46874 |
Sulfanilamide |
sulfonamide |
250 mg |
| 46882 |
Sulfanitran |
sulfonamide |
250 mg |
| 46890 |
Sulfapyridine |
sulfonamide |
250 mg |
| 46902 |
Sulfathiazole |
sulfonamide |
250 mg |
| 46908 |
Sulfisomidine |
sulfonamide |
250 mg |
| 46916 |
Sulfisoxazole |
sulfonamide |
250 mg |
| 46923 |
Testosterone |
hormone |
250 mg |
| 46924 |
Testosterone 17-acetate |
hormone |
250 mg |
| 46926 |
Testosterone 17-benzoate |
| |