|
Certified Standards for Demanding Ion Chromatography Applications |
PRIMUS (Primary Multiions Standards for Ion Chromatography) - A Corporate
Project of:
The need of using CRM's
Today, Ion Chromatography (IC) is an important technique within the analytical laboratory. It is used for the aqueous
determination of ions down to the ppt range, being one of the best methods that can provide quantitative analysis of ions at this
level. Moreover, the development of modern IC apparatus and columns makes this technique a fast, uncomplicated and reliable
tool.
The quality of IC standards had to be improved to keep up with the advances in the available hardware. The selection of
appropriate standards for the calibration of the IC system is important. Fluka already offers a wide range of IC
standards, starting from single elements for anion and cation analytic to multielement standards. These standards are
frequently used by the Environmental, Food and Beverage industries. However, other industries like Pharmaceutical or
Semiconductor, may need to work with even higher qualifications and certified standards to comply with GMP. This trend is
furthermore reinforced by official regulations of the European Community within the guideline 96/23/EG on the use of Certified
Reference Materials (CRM):
...In paragraph 3.1.1.2., the determination of trueness (one component of accuracy) is described: trueness can only be
established by means of CRM's. A CRM should be used whenever available] (1).
The American FDA (Food and Drug Administration) demands in their Inspection Guide for analytic methods the usage of
reference standards.
ISO (International Organization for Standardization) has published more then 6 papers about reference materials.
CRM's are guaranteed as validation standards. This means that they have to be traceable to a SI unit and standard deviation has to
be determined. In accordance to ISO Guide 17025 (2), the preparation of a CRM ideally is carried out by metrological institutes
such as NIST (National Institute of Standards and Technology), EMPA (Swiss Federal Laboratories for Materials Testing and
Research) or BAM in Germany (Federal Institute for Materials Research and Testing).
Single- and Multielement Standards developed in collaboration with EMPA and
BAM
For Ion Chromatography, the number of CRM's available is still small. In order to fill this gap, Fluka launched a few months ago
their first multiion CRM's for Ion Chromatography (Table 1).
The development was carried out in collaboration with EMPA, located in St. Gallen, Switzerland. Based on the competence of this
highly regarded metrological institute, the traceability to SI is guaranteed and standard deviation is determined. Therefore, these
CRM's are produced completely at EMPA, giving you the guarantee of certification you desire.
Following a positive customer response, we were also asked to produce certified multielement standards and we are pleased
to offer two new certified multielement standard solutions, PRIMUS (Primary Multiion Standards) (Table
2).
High precision weighing and high purity salts for highest quality
EMPA determines the purity of the starting materials based on high purity salts (Table 2). The content of
these primary materials is determined by two methods for each salt. The analysis is done in collaboration with BAM. The results are
verified and cross-checked at state-of-the-art levels to ensure the quality you require. The really novel part is the production
procedure. First, the solid salts are determined and then they are dissolved in high purity water. Although it sounds simple, one of
the core competences of EMPA is the extremely precise and accurate weighing of these salts. This allows us to offer our customers
CRM's standards having a very low content of ion species (10mg/kg for each ion). To ensure the quality of our products, each bottle
is delivered in a sealed aluminium bag to minimise the loss of water and to keep the concentration stable over more than two years.
All standards are delivered with a Certificate of Analysis, including a detailed report with the calculation of the measurement
uncertainty.
Did you know that ...
... each new Metrohm IC apparatus contains a pack of our PRIMUS multication and
multianion standards?
| Metrohm highly recommends the use of PRIMUS standards for the internal calibration and
validation of their instruments. |
References
- Commission decision of 12 August 2002 implementing Council Directive 96/23/EC concerning the performance of
analytical methods and the interpretation of results. Official Journal of the European Communities, L 221/8
- ISO Guides 30 (1992): Terms and definitions used in connection with reference materials; 31(2000): Reference materials -
contents of certificates and labels; 32 (1997): Calibration in analytical chemistry and use of certified reference materials; 33 (2000):
Uses of certified reference materials; 34 (2000): General requirements for the competence of reference material producers; 35
(1989): Certification of reference materials - General statistical principles
www.sigma-aldrich.com/certified_standards
Table 1: PRIMUS: Multielement Standard Solutions for Ion Chromatography (10 mg/kg)
Catalog No. |
Product Name |
Ions |
Produced from |
Example Certificate
|
|
89316
|
Certified Multication Standard Solution |
Lithium, Sodium, Potassium, Magnesium, Calcium |
Li2CO3, NaCl, KCl, MgO, CaCO3 |
|
|
89886
|
Certified Multianion Standard Solution |
Fluoride, Chloride, Bromide, Nitrate, Phosphate, Sulfate |
NaF, NaCl, NaBr, NaNO3, Na2SO4, Na2HPO4 |
|
|
PRIMUS standards supplied in 50 mL HDPE bottles. A detailed certificate is provided with each bottle including:
SI traceable content, uncertainty, trace impurities and expiry date. |
| Table 2: |
These anionic standard solutions for Ion Chromatography (1000 mg/L) are
certified in a cooperation project with BAM and Sigma-Aldrich |
Catalog No. |
Product Name |
Produced from |
Example Certificate |
|
87969
|
Bromide |
NaBr |
|
|
87603
|
Chloride |
NaCl |
|
|
80373
|
Fluoride |
NaF |
|
|
86576
|
Nitrate |
NaNO3 |
|
|
81193
|
Phosphate |
Na2HPO4 |
|
|
80218
|
Sulfate |
Na2SO4 |
|
|
| These certified standards are prepared with high-purity salts and water and are supplied in 100 mL HDPE bottles. |
| TraceCERT® Standards for IC |
click here for Background Information
Cat. No. |
Ion |
Composition |
Example Certificate
|
Additional Information |
|
Ammonium |
NH4Cl in water |
|
replaces cat. no. 95023 |
|
Bromide |
NaBr in water |
|
replaces cat. no. 17355 |
|
Chloride |
NaCI in water |
|
replaces cat. no. 24889 |
|
Chromate |
KCrO4 in water |
|
replaces cat. no. 27072 |
|
Cyanide |
|
Fluoride |
NaF in water |
|
replaces cat. no. 47070 |
|
Iodide |
KI in water |
|
new product |
|
Nitrate |
NaNO3 in water |
|
replaces cat. no. 72547 |
|
Nitrite |
NaNO2 in water |
|
replaces cat. no. 77012 |
|
Phosphate |
Na2HPO4 in water |
|
replaces cat. no. 79409 |
|
Sulfate |
Na22SO4 in water |
|
replaces cat. no. 86126 |
|
| Certified Eluent Concentrate for Ion Chromatography |
Our new certified eluent concentrates for ion chromatography are traceable by potentiometric titration
to NIST Standard Reference Material. It is certified in accordance with ISO Guide 31. All details about
exact content, uncertainty, traceability and expiry date are described in the certificate.
Table 1: TraceCERT® calibration standards for IC
Cat. No. |
Product Name |
Concentration (mMole) |
Example Certificate |
|
2,6-pyridinedicarboxylic acid |
20 |
|
|
DL-tartaric acid |
100 |
|
|
Methanesulfonic acid |
100 |
|
|
Nitric acid |
100 |
|
|
Perchloric acid |
10 |
|
|
Potassium hydroxide |
100 |
|
|
Sodium bicarbonate |
100 |
|
|
Sodium carbonate |
100 |
|
|
Sodium hydroxide |
100 |
|
|
Sulfuric acid |
100 |
|
|
| Packaging: 1L HDPE bottles with 45mm screw thread |
| Water for Ion Chromatography |
Fluka has developed a water quality especially for the needs of ion chromatography.
This water is
also suitable for trace analysis of anions, cations and also some organics
which are usually analyzed
by IC.
The new product 00612 is available in 2.5L and 5L package size and specified as follows:
Organic Traces:
Acetate, formiate, glycolate, oxalate: ≤ 10 µg/kg each
Cation Traces:
Al, Ba, Bi, Cd, Co, Cr, Cu, Fe, Li, Mg, Mn, Mo, Ni, Pb, Sr, Zn: ≤ 5 ìg/kg each
Ca, K, Na: ≤ 10 µg/kg
each
NH4+: ≤ 50µg/kg
Anion Traces:
Bromide, chloride, fluoride, iodide, nitrate, nitrite, phosphate, sulfate:
1 µg/kg each
Conductivity:
≤ 2 µS/cm
Catalog No. |
Product Name |
Pack Size |
|
00612
|
Water for Ion Chromatography |
2.5 L, 5L |
| Packaging: 2.5L and 5L HDPE bottles with 45mm thread |
|
|
| Standards for Ion Chromatography |
Ion chromatographic standard solutions require slightly different specification than spectroscopic solutions:
- The solution is filtered through a 0.45 mm membrane filter to prevent the column from clogging
- pH > 3
All the solutions are carefully prepared from high-purity salts, preferably nitrates; their concentration is 1000 mg/L. For packaging high-density polyethylene bottles are used. On each label the expiry date of the solution is stated.
Cation Standards for Ion Chromatography (1000mg/L)
|
Catalog No.
|
Product Name |
CAS No.
|
Additional information
|
|
Ammonium |
12125-02-9
|
replaced by 59755 |
|
11719
|
Barium |
10022-31-8
|
|
|
14203
|
Beryllium |
-
|
|
|
20892
|
Cadmium |
10325-94-7
|
|
|
21041
|
Calcium |
10124-37-5
|
|
|
20951
|
Cesium |
7647-17-8
|
|
|
27570
|
Cobalt |
10141-05-6
|
|
|
61151
|
Copper |
3251-23-8
|
|
|
15316
|
Lead |
10099-74-8
|
|
|
62369
|
Lithium |
7790-69-4
|
|
|
63041
|
Magnesium |
10377-60-3
|
|
|
63533
|
Manganese |
10377-66-9
|
|
|
72224
|
Nickel |
13138-45-9
|
|
|
60037
|
Potassium |
7757-79-1
|
|
|
71167
|
Sodium |
7631-99-4
|
|
|
85889
|
Strontium |
10042-76-9
|
|
|
96456
|
Zinc |
7779-88-6
|
|
|
IC standards supplied in 100 mL HDPE bottles.
Matrix: water or 0.001M nitric acid solution. |
Anion Standards for Ion Chromatography (1000 mg/L)
|
Catalog No.
|
Product Name |
CAS No.
|
Additional information |
|
17355
|
Bromide |
7647-15-6
|
replaced by 43147 |
|
24889
|
Chloride |
7647-14-5
|
replaced by 39883 |
|
27072
|
Chromate |
7789-00-6
|
replaced by 40121 |
|
28491
|
Cyanide |
151-50-8
|
|
|
47070
|
Fluoride |
7681-49-4
|
replaced by 77365 |
|
72547
|
Nitrate |
7631-99-4
|
replaced by 74246 |
|
Nitrite |
-
|
replaced by 67276 |
|
79409
|
Phosphate |
7778-77-0
|
replaced by 38364 |
|
86126
|
Sulfate |
7757-82-6
|
replaced by 90071 |
|
| IC standards supplied in 100 mL HDPE bottles. Matrix: water |
Multiion Standards for Ion Chromatography
Catalog No. |
Product Name |
Ion Concentration |
CAS No. |
|
87029
|
Multielement Anion |
Br (20mg/kg), Cl(10mg/kg), F(3mg/kg), NO3(20mg/kg), PO4(30mg/kg), SO4(20mg/kg) |
-
|
|
63597
|
Multielement I |
Br (1000mg/L), Cl(1000mg/L), F(1000mg/L) |
-
|
|
63598
|
Multielement II |
NO3(1000mg/L), PO4(1000mg/L), SO4(1000mg/L) |
-
|
|
| IC standards supplied in 100 mL HDPE bottles. Matrix: water |
Eluent Concentrates for Ion Chromatography
Catalog No. |
Product Name |
Pack Size |
CAS No. |
|
72064
|
Sodium hydroxide solution, eluant, 50-52% |
500 mL |
1310-73-2
|
|
86854
|
Tetrabutylammonium hydroxide, ~40% |
100 mL, 500mL, 2.5L |
2052-49-5
|
|
Organic Acid Standards for Ion Chromatography (1000mg/L)
Due to the omnipresence of organic acids in foods and beverages such as fruit drinks and wines and their link to certain nutritional, quality and safety aspects of the food, interest in measuring their concentration is growing rapidly. Ion chromatography is a common method for the analysis of organic acids.
To meet the requirements of our analytical customers, we developed a group of organic acids standards for ion chromatography (Table 1). The concentration is 1.000 g/L in water. All standards are carefully prepared from high purity starting materials, dissolved in water 17749 for analytical usage, filtered through a 0.22µm membrane filter to prevent column fouling and clogging. All standards packaged in high density polyethylene bottles under sterile conditions. An expiration date is clearly provided on each bottle.
Catalog No. |
Brand |
IC Standard |
|
Fluka |
Acetate |
|
Fluka |
Citrate |
|
Fluka |
Formate |
|
Fluka |
Malonate |
|
Fluka |
Oxalate |
|
Fluka |
Phthalate |
|
Fluka |
Propionate |
|
Fluka |
Tartrate |
|
| IC standards supplied in 100 mL HDPE bottles. |
Back to Top
Back to Trace Analysis home
Back to Analytical home
|