For certified calibration solutions, we maintained a long-standing partnership with Swiss Federal Institute for Materials Science and Technology (EMPA). However, EMPA terminated all their activities in chemical metrology in 2004 and no longer acts as a national metrological institute. Recognizing that EMPA’s exit created the need for a first class CRM supplier in the market, Sigma-Aldrich made a strategic decision to acquire the metrological competency from EMPA and transfer it to our Buchs, Switzerland site.
We began in late 2005 to replicate the EMPA metrological environment in order to provide a seamless transition. First, we purchased from EMPA the unique equipment which was custom-designed to produce intercomparison samples of the highest metrological level (CCQM Key Comparisons). Second, we built a special, dedicated laboratory at the Buchs site where the high-precision weighing capabilities, homogenization and clean room bottling equipment were installed.
|
| TraceCERT® reference materials are developed and produced in an accredited laboratory fulfilling both ISO 17025 (General requirements for the competence of testing and calibration laboratories) and ISO Guide 34 (General requirements for the competence of reference material producers). |
TraceCERT® products are characterized by
- Unique (metrological) level of accuracy and lot-specific values
- Traceability to at least two independent references (i.e. NIST, BAM or SI unit kg)
- Certification according to ISO Guide 31
- Highest-purity starting materials available
- Comprehensive documentation including proper uncertainty calculation,
expiry date and storage data
- Certificates of ICP standards list up to 70 trace impurities
- ICP standards packaged in light- and gas-tight aluminum foil bags
- Competitively priced
Certified Element Standards for AAS and ICP
| Element |
Composition |
Cat. no.
ICP Standard |
Example Certificate |
Add to Cart |
Cat. no.
AAS Standard |
Example Certificate |
Add to Cart |
| Aluminum |
Al(NO3)3 x 9H2O + HNO3 |
|
| Antimony |
Sb metal + HNO3 + HF |
|
| Arsenic |
As2O3 + HNO3 (+ NaOH for AAS) |
|
| Barium |
BaCO3 + HNO3 |
|
| Boron |
H3BO3 + H2O |
|
| Cadmium |
Cd metal + HNO3 |
|
| Calcium |
CaCO3 + HNO3 |
|
| Cesium |
CsNO3 + HNO3 |
|
| Chromium |
(NH4)2Cr2O7 + HNO3 |
|
| Cobalt |
Co metal + HNO3 |
|
| Copper |
Cu metal + HNO3 |
|
| Gold |
Au metal + HCl (HNO3 traces) |
|
| Iron |
Fe metal + HNO3 |
|
| Lead |
Pb(NO3)2 + HNO3 |
|
| Magnesium |
Mg metal + HNO3 |
|
| Manganese |
Mn metal + HNO3 |
|
| Mercury |
Hg metal + HNO3 |
|
| Nickel |
Ni metal + HNO3 |
|
| Palladium |
Pd(NO3)2 + HCl |
|
| Phosphorus |
H3PO4 + H2O |
|
| Platinum |
Pt metal + HCl (HNO3 traces) |
|
| Potassium |
KNO3 + HNO3 |
|
| Scandium |
Sc2O3 + HNO3 |
|
| Selenium |
Se metal + HNO3 |
|
| Silicon |
Si metal + NaOH |
|
| Silver |
Ag metal + HNO3 |
|
| Sodium |
NaNO3 + HNO3 |
|
| Strontium |
Sr(NO3)2 + HNO3 |
|
| Sulfur |
H2SO4 + H2O |
|
| Tin |
Sn metal + HCl |
|
| Vanadium |
V2O5 + HNO3 |
|
| Yttrium |
Y2O3 + HNO3 |
|
| Zinc |
Zn metal + HNO3 |
|
ICP standards supplied in 100 mL HDPE bottles and sealed in an aluminum bag, including certificate.
AAS standards supplied in 250 mL HDPE bottles, except for Hg which is bottled in 100 mL borosilicate white glass bottle.
The following AAS standards are supplied in 100 mL HDPE bottles: Pd, Au, Pt, Sc
Certified Ion Standards for IC (TraceCERT®)
Cat. No. |
Ion |
Composition |
Example Certificate |
Additional Information |
|
Ammonium |
NH4CI in water |
|
replaces cat. no. 95023 |
|
Bromide |
NaBr in water |
|
replaces cat. no. 17355 |
|
Chloride |
NaCI in water |
|
replaces cat. no. 24889 |
|
Chromate |
K2CrO4 in water |
|
replaces cat. no. 27072 |
|
Cyanide |
|
Fluoride |
NaF in water |
|
replaces cat. no. 47070 |
|
Iodide |
KI in water |
|
new product |
|
Nitrate |
NaNO3 in water |
|
replaces cat. no. 72547 |
|
Nitrite |
NaNO2 in water |
|
replaces cat. no. 77012 |
|
Phosphate |
Na2HPO4 in water |
|
replaces cat. no. 79409 |
|
Sulfate |
Na22SO4 in water |
|
replaces cat. no. 86126 |
|
Production and certification of TraceCERT® standards
To actually produce high quality CRMs, the entire development process, beginning from the selection of an appropriate starting material to the choice of a purpose-specific packaging, must be highly sophisticated.
Figure 1: Schematic representation of the production design and certification of Sigma-Aldrich TraceCERT® products.
This approach was established for the preparation of intercomparison samples at the highest metrological stage.
Not only is the bottled solution compared to a reference material (NIST, BAM, etc.), the starting material is checked against a second, independent reference. A key step is that the weighing operation leads to direct traceability to the SI-unit kilogram. Gravimetric preparation using pure materials is a practical and, in many cases, the most accurate calculation of concentration units, through conversion of mass and mole fraction to mass fraction.
As a final step, we provide detailed, lot-specific information in the Certificate of Analysis supplied with every TraceCERT® product. Our well-defined approach to CRM production leads to multiple-traceability and, as a further benefit, ensures that significant biases and systematic deviations are excluded to the fullest extent possible. |
Serie of ANALYTIX articles about certified reference materials
There is much confusion and many misconceptions over traceability, uncertainty calculation and the proper use of reference materials. In the next seven issues of Analytix, we will attempt to enlighten interested readers on the meaning behind important CRM-related terms and concepts, including
- What does traceability really mean?
- The proper method to calculate uncertainty
- Characterization and purity statement of high-purity starting materials
- Production, liquid handling and packaging on a metrological level
- Certification and accreditation
- Certification of custom standards
Now available:
Part 1: Introduction - Swiss precision meets analytical competence (PDF, 104KB)
Part 2: What treaceability really means (PDF, 172KB)
Part 3: Challenges in the characterization of high-purity starting materials (PDF, 102 KB)
Part 4: Production, handling and storage of high-precision calibration solutions (PDF, 129 KB)
Part 5: Reliability is a matter of proper uncertainty calculation (PDF, 106 KB)
Part 6: Double accreditation - increasing the reliability and and value of analytical results (PDF, 168 KB)
Back to Application Area / Analytical Technique
|