|
Biocide Preservatives |
 |
Analytical / Chromatography |
 |
 |
| |
|
| Kathon® Preservative |
| Kathon CG/ICP preservative eliminates the problem of microbial
growth, reducing downtime and extending the lifetime of your media and chromatographic
system. This preservative is a highly effective, broad-spectrum biocide
with excellent compatibility and stability and low toxicity at in-use levels.
|
| |
| ProClin™ Preservative |
| ProClin preservative is a highly effective biocide for
controlling microorganisms in reagents and products intended for in vitro
diagnostic use. These preservatives are an excellent choice for replacing
thimerosal, sodium azide, and gentamicin preservatives. |