|
Super Clean™ Gas Purifiers |
 |
Gas Chromatography |
 |
 |
| |
|
| Click-On Inline
Super Clean Gas Purifiers |
Changing out a traditional inline purifier allows the open gas line to
be exposed to unpurified room air, requiring a lengthy and unproductive
flushing process. Getting dirt in the threads or over-tightening of gas
line fittings can prevent proper sealing, providing a source for gas contamination.
The Click-On Inline Super Clean Gas Purifiers have none of those disadvantages.
They are used for the removal of hydrocarbons, oxygen, and moisture in
carrier gas and fuel gases for your GC or GC-MS system. Click-On Inline
Super Clean Gas Purifiers are available with or without visual indicators.
In sharp contrast to traditional inline purifier replacement, change out
of the Click-On Inline Super Clean Gas Purifiers is simple and eliminates
the risk of damaging compression fittings. Once the Click-On connectors
are installed into the gas line, those connections never need to be broken.
Subsequent replacement of purifiers can be done in seconds. The new purifier
is snapped in place and the fittings are hand tightened. Each Click-On
connector contains a needle valve that snaps shut the instant the purifier
is removed. This prevents introduction of room air into the gas line and
allows the change out without shutting down the GC. Click-On connectors
are available in either brass or stainless steel with either 1/8 inch
or 1/4 inch compression fittings. |
| Click-On InLine Traps |
| Cat. No. |
Description |
Add to order |
| 28861-U |
Moisture Trap |
|
| 28862-U |
Oxygen Trap |
|
| 28863-U |
Hydrocarbon Trap |
|
| 28864-U |
Triple Oxygen/Moisture/Hydrocarbon Trap for Carrier
Gas |
|
| 28865-U |
Gas Specific (He) Triple Oxygen/Moisture/Hydrocarbon
Trap |
|
| 28866-U |
Combi Moisture/Hydrocarbon Trap for Fuel Gas |
|
| 28867-U |
Indicating Triple Oxygen/Moisture/Hydrocarbon Trap |
|
| |
| Click-On Connectors |
| Cat. No. |
Description |
Add to order |
| 28868-U |
Click-On Connector 1/4” Brass (Pk. 2) |
|
| 28869-U |
Click-On Connector 1/8” Brass (Pk. 2) |
|
| 28872-U |
Click-On Connector 1/4” SS (Pk. 2) |
|
| 28873-U |
Click-On Connector 1/8” SS (Pk. 2) |
|
| 28874-U |
Click-On Double Version 1/8” (Pk. 2) |
|
| |
| Accessories |
| Cat. No. |
Description |
Add to order |
| 28875-U |
Replacement O-ring Set (Pk. 10) |
|
| 28876-U |
Wall Mounting Clamp |
|
|
| back to top |
|
Super Clean Gas Purifiers
Super Clean Gas Purifiers use a base plate design. Replacement purifiers
can be installed in seconds without exposing gas lines to room air. The
specially designed base plate rests on the bench top and has needle valves
that instantly close to provide a diffusion-proof seal when the gas purifier
is removed. Replacement gas purifiers need only be held in place on the
base plate while the retaining ring is hand tightened.
Super Clean Gas Purifiers purify carrier, fuel and other gases for your
GC or GC-MS system by removing hydrocarbons, oxygen (color indicating),
and moisture (color indicating). |
| Super Clean Cartridge Systems |
| Cat. No. |
Description |
Add to order |
| SU861021 |
Ultra-High Capacity Moisture Filter with indicator |
|
| SU861022 |
Ultra-High Capacity Oxygen Filter with indicator |
|
| SU861023 |
Ultra-High Capacity Hydrocarbon Filter (charcoal)
without indicator |
|
| SU861025 |
Combi GC Filter, replacement fuel gas filter (charcoal/moisture) |
|
| SU861026 |
Triple GC Filter Replacement (oxygen/moisture/charcoal) |
|
| SU861027 |
Triple GC Filter Replacement for Helium (oxygen/moisture/charcoal)
|
|
| 28878-U |
Triple GC Kit for Carrier Gas, 1 position |
|
| SU861040 |
GC-MS Kit for He (Base plate + 1 triple filter) |
|
| SU861043 |
GC-FID Kit for 3 filters, Triple + 2x Combi Cartridge-
(Charcoal/moisture) |
|
| |
| Super Clean Cartridge Systems -
Hardware only |
| Cat. No. |
Description |
Add to order |
| SU861011 |
Single-Position Base plate |
|
| SU861012 |
2-Position Base plate |
|
| SU861013 |
3-Position Base plate |
|
| back to top |
|
|
Super Clean LC-MS Gas Purifiers
Super Clean Gas LC-MS Purifiers, a new revolution in clean
gas purifiers, purify nitrogen from a nitrogen generator for LC-MS systems.
These point-of-use glass/metal, diffusion-proof purifiers can remove hydrocarbons
to produce nitrogen that is better than 6.0 (99.9999%) gas quality, independent
of the original gas quality.
The Super Clean Fast-Connect design avoids MS source damage and helps
to eliminate LC-MS downtime. The two cartridge purifier systems, used
in parallel, is the only fully glass/metal cartridge purifier capable
of purifying LC-MS N2 carrier gas for all major hydrocarbon contaminants.
The activated charcoal bed of the indicating hydrocarbon filter adsorbs
organic compounds larger than ethane. The indicating material changes
from bright yellow to dark green when the trap has reached saturation
and needs to be replaced. |
| Super Clean Gas Filter Kits |
| Cat. No. |
Description |
Add to order |
| SU861045 |
High Flow Special Moisture Super Clean Filter Kit
(2 filters/base plate) |
|
| SU861046 |
High Flow LC-MS Super Clean Filter Kit for Nitrogen
Purification without
indicator (2 charcoal filters) |
|
| |
| Super Clean Gas Filter Cartridges |
| Cat. No. |
Description |
Add to order |
| SU861028 |
High Flow Special Moisture Filter: bundle of 2 replacement
cartridges |
|
| SU861029 |
Charcoal for LC, Nitrogen Purification Super Clean
Filter, bundle of 2 charcoal cartridges |
|
| 28877-U |
High Flow Charcoal Super Clean Filter for LC-MS |
|
| |
| Base Plate |
| Cat. No. |
Description |
Add to order |
| 28879-U |
2-Position Base plate for High Flow LC-MS |
|
| |
| Accessories |
| Cat. No. |
Description |
Add to order |
| SU861016 |
Wall Mounting Bracket for Super Clean Cartridge
System |
|
| SU861050 |
Replacement O-Rings for Super Clean Cartridges (10
large/10 small) |