|
Inertness of Supelco Low Bleed GC Capillary Columns |
 |
Gas Chromatography |
 |
 |
| |
|
| |
| The Proof: Inertness |
| |
SLB columns achieve their high level of inertness by
virtue of a Proprietary Surface Deactivation that we apply to the
fused silica tubing.
Inertness, a measure of the absence of adsorptive or active sites in
the column, is an important attribute in that it influences peak signal.
Increased peak signal, in addition to decreased noise, is the best way
to consistently achieve low detection limits. Additionally, a column that
does not possess good inertness may end up reducing profits due to additional
labor requirements to keep the system in working order. |
| Inertness = Sharp Peaks = Low Detection Limits
Most analytes have at least one distinguishing functional group in their
molecular structure. The functionality may be subtle, such as a double
bond, or pronounced, such as a chlorine substitution, and may adversely
affect the chromatography. Polar functional groups, such as hydroxyls
(-OH) or amines (-NH2), are problematic due to their tendency
to adsorb to active sites on any surface (syringe needle, injection port
liner, column, etc.) with which they come into contact. Adsorption results
in broad, short peaks. Because more of the peak area is in the noise region,
it tends to get lost by the software algorithm. Adsorption is essentially
equivalent to the loss of area counts.
In Figure 1, the Extracted Ion Current Profile (EICP), also known
as an Extracted Ion Chromatogram (EIC), for 2,4-dinitrophenol from a US
EPA Method 8270D analysis on an SLB-5ms column is shown. This analyte
contains a hydroxyl (-OH) and two nitro groups (-NO2) attached
to an aromatic ring, and is historically a very troublesome analyte for
this method. Figures 2 and 3 show EICPs of 4-nitrophenol and 4-nitroaniline,
respectively, from the same analysis. 4-Nitrophenol contains a hydroxyl
group and a nitro group attached to an aromatic ring, whereas 4-nitroaniline
contains an amine group and a nitro group attached to an aromatic ring.
These analytes are also known to be troublesome, although to a lesser
degree than 2,4-dinitrophenol. Note the sharpness of the traces of the
quantitation m/z, 184, 139, and 138, respectively, on the SLB-5ms column
shown in these three Figures. It is important to keep in mind that sharper
peaks result in greater peak area that is above the noise level. Since
most analytes possess one or more groups of varying functionality, it
is important to use a deactivated column, like SLB-5ms, to ensure good
peak shape and sensitivity for all components of the sample.
Figure 1. EICP of 5 ng On-column of 2,4-Dinitrophenol.
Figure 2. EICP of 5 ng On-column of 4-Nitrophenol.
Figure 3. EICP of 5 ng On-column of 4-Nitroaniline.
|
Inertness = Negligible Peak Tailing = Minimal Preventative
Maintenance
As previously discussed, an active surface can lead to adsorption. Besides
causing broad, short peaks, adsorption can cause peak tailing. Many environmental
and pharmaceutical methods have a peak symmetry test that must be satisfied
prior to the analysis of samples. Excessive peak tailing makes it difficult
to pass the test. Failure to pass the test requires the analyst to perform
maintenance (chemically treating the activity, clipping the column, replacing
the column, etc.). These activities increase instrument downtime, which
keeps the analyst from analyzing samples.
US EPA Method 625 requires that two analytes, one acidic and one basic,
must pass a daily tailing factor test prior to the analysis of any sample
extract. Figures 4 and 5 show EICPs on an SLB-5ms for pentachlorophenol
and benzidine, respectively, at the method-required concentrations. Both
tailing factors were extremely close to a value of 1, indicating excellent
peak shape for both analytes in addition to falling well within the method-required
criteria.
Figure 4. EICP of 50 ng On-column of Pentachlorophenol.
Figure 5. EICP of 100 ng On-column of Benzidine.
|
|
| Inertness = Great RRFs = Minimal Preventative Maintenance
Many US EPA Methods that employ the use of a mass selective detector
(MSD) require the use of internal standards, compounds that are structurally
similar to the target analytes. The internal standards are added to the
sample extract shortly before analysis and are used to compensate for
injection discrepancies and drift in MSD response. The response for every
target analyte relative to the response for a specific internal standard
is calculated. This relative response factor (RRF) is plotted against
the calibration curve to determine concentration.
These methods list minimum RRF criteria for poor responding target analytes,
sometimes referred to as System Performance Check Compounds (SPCCs), as
a measure of system performance. If an RRF is below the minimum criteria
in either an initial or a continuing calibration, the analyst must take
action to correct the problem prior to analysis of samples. This usually
involves cutting off a section of column from the injector end to remove
surface activity.
RRF values for these poor performing target analytes from US EPA Methods
8270D and OLM04.2 SVOA are shown in Table 1. Target analytes with
polar functionality tend to have low RRFs due to adsorption. The observed
RRFs clearly exceed the minimum RRF criteria specified for these analytes.
|
| |
| Table 1. RRFs at 50 ng On-column (Internals
at 40 ng On-column) of Several Poor Performers. |
| Analyte |
Analyte
m/z |
Internal
Standard * |
8270D
Criteria |
OLM04.2
Criteria |
Observed |
| Bis(2-chloroisopropyl)ether |
45 |
1 |
- |
0.010 |
1.702 |
| N-nitroso-di-n-propylamine |
70 |
1 |
0.050 |
0.500 |
1.020 |
| 4-chloroaniline |
127 |
2 |
- |
0.010 |
0.441 |
| Hexachlorobutadiene |
225 |
2 |
- |
0.010 |
0.153 |
| Hexachlorocyclopentadiene |
237 |
3 |
0.050 |
0.010 |
0.301 |
| 2-nitroaniline |
65 |
3 |
- |
0.010 |
0.365 |
| Dimethyl phthalate |
163 |
3 |
- |
0.010 |
1.272 |
| 3-nitroaniline |
138 |
3 |
- |
0.010 |
0.348 |
| 2,4-dinitrophenol |
184 |
3 |
0.050 |
0.010 |
0.160 |
| 4-nitrophenol |
139 |
3 |
0.050 |
- |
0.879 |
| 4-nitrophenol |
109 |
3 |
- |
0.010 |
0.152 |
| Diethyl phthalate |
149 |
3 |
- |
0.010 |
1.290 |
| 4-nitroaniline |
138 |
3 |
- |
0.010 |
0.339 |
| 2-methyl-4,6-dinitrophenol |
198 |
4 |
- |
0.010 |
0.128 |
| N-nitrosodiphenylamine |
169 |
4 |
- |
0.010 |
0.653 |
| 2,4,6-tribromophenol (surr.) [1] |
330 |
3 |
- |
- |
0.169 |
| 2,4,6-tribromophenol (surr.) [2] |
330 |
4 |
- |
0.010 |
0.106 |
| 4-bromophenyl phenyl ether |
248 |
4 |
- |
0.100 |
0.229 |
| Pentachlorophenol |
266 |
4 |
- |
0.050 |
0.132 |
| Carbazole |
167 |
4 |
- |
0.010 |
0.961 |
| Di-n-butyl phthalate |
149 |
4 |
- |
0.010 |
1.088 |
| Butylbenzyl phthalate |
149 |
5 |
- |
0.010 |
0.615 |
| 3,3'-dichlorobenzidine |
252 |
5 |
- |
0.010 |
0.401 |
| Bis(2-ethylhexyl)phthalate |
149 |
5 |
- |
0.010 |
0.769 |
| Di-n-octyl phthalate |
149 |
6 |
- |
0.010 |
1.258 |
[1] Using 8270D specified internal standard.
[2] Using OLM04.2 specified internal standard. |
* Internal Standards
1 = 1,4-dichlorobenzene-d4, m/z 152
2 = Naphthalene-d8, m/z 136
3 = Acenaphthene-d10, m/z 164
4 = Phenanthrene-d10, m/z 188
5 = Chrysene-d12, m/z 240
6 = Perylene-d12, m/z 264 |