|
HPLC Accessories
Chiralyser MP
|
|
|
Feat ures
- 100x0.5mm flow cell (analytical)
- LED lasts 100,000 hours
- Low maintenance
- Energy efficient
- Variable voltage range
- No adsorption phenomena
- LED wavelength chosen for maximum sensitivity
Benefits
- Robust
- Sets up in minutes
- Easy-to-use
- Excellent sensitivity
- Identifies enantiomer pairs
- Identifies elution orders
- Validates peaks as (-) and (+)
| Specifications |
| Operation principle: |
Faraday compensation |
| Measuring range: |
Minimum 32x10-4 degree Maximum 2048x10-4 degree in 8 ranges |
| Resolution: |
16 bit |
| Noise: |
2x10-5 degree |
| Stability: |
5x10-5 degree/hour |
| Detection limit: |
0.5 mg/L (closed loop, [a=90°], S/N=2) |
| Flow cell: |
Glass coated stainless steel 100 mm, 20µL (analytical) 100 mm, 300µL (preparative) |
| Flow rate: |
0.3 to 3 mL/min, analytical cell 2.5 to 100 mL/min, preparative cell |
| Size and weight: |
450x190x90mm, 12 Kg |
| Product Bulletin |
| |
CHIRALYSER MP |
| |
|
This presentation requires the Acrobat plug-in for proper viewing. Download the reader for FREE.
|
|
For ordering information, go to our