Analytical / Chromatography
Air Monitoring
Analytical Reagents
Analytical Standards
Gas Chromatography
HPLC
Accessories
Columns
Ascentis Express
Ascentis
Chiral
Discovery
apHera - High pH
Supelcosil
TSK-GEL
Solvents
Microbiology
Microscopy
Sample Preparation
Spectroscopy
Syringes
Titration
TLC
Vials
Videos
Events
Literature
Analytix
The Reporter
Reporter Europe
 Ascentis Express Fused-Core™ HPLC Columns
HPLC
image for fused core hplc column
Ascentis Express columns provide a breakthrough in HPLC column performance.
Based on Fused-Core™ particle technology, Ascentis Express provides the benefits of high speed and high efficiencies of sub-2 µm particles at much lower backpressures. Due to the high efficiencies at low backpressures, Ascentis Express can benefit both conventional HPLC users as well as UPLC™ or other ultra pressure system users.
Ascentis Express Extreme Performance benefits include:
  • Double the efficiencies of conventional 3 µm particles
  • Equal efficiencies of sub-2 µm columns at half of the backpressure
  • Rugged design capable of high pressure operation
Available in C18, C8, and (NEW!) HILIC
Ascentis Express C18 and C8 provide classic reversed-phase selectivity with performance benefits for Fast HPLC and High Resolution HPLC. Ascentis Express HILIC provides all of the benefits of Fused-Core particle technology to the emerging technique of HILIC mode chromatography.
HILIC chromatography is complementary to reversed-phase separations due to increased retention for polar compounds. The elution order is typically the opposite of that for C18 with the non-polar compounds eluting early and the highly polar compounds having greater retention and selectivity.
Ordering information for Ascentis Express HPLC columns
Technical Resources
Sign up to receive the Fused-Core Report! A quarterly report on the lastest applications, publications, and new products related to fused-core particle technology.
Publication Title Type Size (kb) No. Pages
Ascentis Express Brochure (view / request) Brochure 1,075 8
NEW! Ascentis Express HILIC HPLC Columns Flyer 360 2
High Performance HPLC Fittings/Interconnects Product Info. 304 2
Ascentis Express: A Fused-Core™ Particle HPLC Column for High Speed and High Efficiency Separations with Low Back Pressures App Note 181 91 2
Guidelines for Optimizing Performance with Ascentis Express HPLC Columns Data Sheet 31 1
Improving USP methods using Ascentis Express (Reporter volume 26.1) Article 482 3
Fast HPLC using Ascentis Express (Reporter volume 25.5) Article 181 3
High Resolution HPLC Using Ascentis Express (TheReporter volume 25.4) Article 307 2
Transferring Methods Using Ascentis Express (TheReporter volume 25.3) Article 273 2
Introducing Ascentis Express (TheReporter volume 25.2) Article 482 3
Trademarks
Ascentis - Sigma-Aldrich Co.
Fused-Core - Advanced Materials Technology, Inc.
UPLC - Waters Corp.