Analytical / Chromatography
Air Monitoring
Analytical Reagents
Analytical Standards
Gas Chromatography
HPLC
Accessories
Columns
Ascentis Express
Ascentis
Chiral
Discovery
apHera - High pH
Supelcosil
TSK-GEL
Solvents
Microbiology
Microscopy
Sample Preparation
Spectroscopy
Syringes
Titration
TLC
Vials
Videos
Events
Literature
Analytix
The Reporter
Reporter Europe
 apHera™ Polymer-Based Reversed Phase Achiral HPLC Columns
HPLC Columns
Astec Products | Technical Resources | Chiral Services
> Chiral HPLC Columns > apHera Polymer-Based Reversed Phases
Ordering Information
Features
  • Stable vinyl copolymer base
  • Stable pH range 2-12
  • 300 Å pore size
  • Sharp resolution, high NTP
  • Stable in all organic solvents
  • Expanded applications: peptides, proteins
  • Efficient separation of any basic substance
  • Amenability to washing with alkaline solutions
  • Available as C4, C8 and C18
  • In recent years reversed phase chromatography has become predominant throughout high performance liquid chromatography applications. Column packings have generally been alkyl-bonded silica gels such as ODS, and, to a lesser extent, polystyrene-based porous gels. Silica based phases are mechanically stable and generally high in NTP. However, they cannot be used under alkaline conditions and their residual silanol groups tend to adsorb organic bases.
    Conventional polystyrene gels are free from residual silanol groups and can be used under alkaline conditions but they are also low in NTP and undergo excessive shrinkage and swelling with various solvents, thus limiting the range of eluents that can be used. Polymer based reversed phase columns have, therefore, generally been viewed as inferior in strength and separation efficiency.
    apHera™ reversed phase columns were developed specifically to provide the superior advantages of both silica and polystyrene columns, without the disadvantages of either. This was accomplished using a vinyl alcohol copolymer base that keeps the surface wetted even with high carbon loads. The columns are packed with butyl (C4), octyl (C8) or octadecyl (C18) packings obtained by the introduction of the alkyl function on the hydroxyl groups of vinyl alcohol copolymers. The porous structure has an average pore diameter large enough to produce ideal results for small analytes, peptides and small proteins. These columns equal silica based columns in separation efficiency with organic solvents but provide efficiency with buffered and alkaline solutions not possible on silica. Shrinkage and swelling are minimal in a broad range of solvents, and the high NTP values of these columns are practically unaffected by differing solvent polarities, unlike polystyrene based polymer columns. One of the most significant features is the logical elution order of alkylated bases where retention increases proportionately with increasing chain length.
    The apHera™ columns represent a fundamental advance in liquid chromatography with a greatly expanded field of reversed phase applications.
    We also offer apHera NH2 Amino columns for carbohydrate analysis.