Analytical / Chromatography
Air Monitoring
Analytical Reagents
Analytical Standards
Gas Chromatography
HPLC
Microbiology
Basic Ingredients
Agar
Extracts
Plant Peptones
Protein sources
Saccharide Substrates
Identification Tests
Microbiology Manual
Product Lines
Quality Assurance
Theory
Microscopy
Sample Preparation
Spectroscopy
Syringes
Titration
TLC
Vials
Videos
Events
Literature
Analytix
The Reporter
Reporter Europe
 Plant Peptones
Microbiology
In a time of animal diseases, like BSE, TSE or foot and mouth disease, it is important to have a good alternative possibility. 100% vegatable protein source that means no risk of BSE/TSE, no problems with certificates and regulations. But it is not the only reason for a change, because several test showed that it is possible to get a better growth or yield by using Plant Peptone. This facts helps you to save time an money. Nowadays, the applications using Plant Peptone increases drastically, although there are thousands of older existing standard methods. The plant peptones in table 1 are specially tested as replacement for animal based peptones. For further request please contact our technical service.
Lists:
  1. plant peptones tested as replacement for animal based peptones
  2. plant peptones tested for growth of different microorganisms
Prod. No. Plant Peptone Animal Peptone Description
18332
Peptone (vegetable) Peptone Pepton (vegetable) is a meat-free alternative to peptones from meat (Fluka 70175). It supports good growth of a wide variety of microorganisms in culture media in routine diagnostic and research bacteriology.
51841
Peptone (vegetable) acid hydrolysate Casein acid hydrolysate Peptone (vegetable) acid hydrolysate is a purified hydrolysate which provides the necessary nitrogenous material for culture media or can be used when amino acid mixtures are specified. It can be used for preparing Antibiotic Sensitivity Test Media including Mueller Hinton Agar, in media requiring quantitative addition of tryptophan, in vaccine preparation media as a source of high concentration of free amino acids.
19942
Peptone (vegetable),
no 1
Meat peptone Peptone (vegetable) no 1 is a meat-free alternative to traditional peptones. A highly nutritious general purpose peptone for growth of bacteria and fungi.
61854
Peptone (vegetable),
no 2
Gelatine peptone Peptone (vegetable) No. 2 is prepared under controlled conditions by enzymic digestion of vegetable proteins. It has nutritional characteristics that matches with Gelatin peptones (Fluka 70176). It can be employed in media for fermentation studies: Purple Agar base, Sugar free Agar, MacConkey Agars, Violet Red Bile Agar, etc.
92976
Peptone special (vegetable) Peptone special (Neopeptone) Manufactured under controlled conditions from vegetable proteins. It is especially adapted for the preparation of media for culturing fastidious bacteria and the support growth conditions found in Peptone special (Fluka 68971). Used for cultivation and fermentation media for fastidious bacteria.
29185
Proteose peptone (vegetable) Proteose peptone Proteose Pepton (vegetable) is an enzymic hydrolysate of vegetable protein and can be employed in media for bulk production of antibiotics, enzymes, veterinary preparation, bacterial toxins, etc. It can successfully replace Proteose peptone (Fluka 82450).
16922
Tryptone (vegetable) Tryptone Tryptone (vegetable) is free of animal protein and an alternative to Tryptone enzymatic digest from caseine (Fluka 95039).
12331
Tryptose (vegetable) Tryptose Tryptose (vegetable) is an animal protein-free alternative to traditional tryptose (Fluka 70937). It is used in several media, e.g. tryptose media for cultivation of fastidious microorganisms or it is well suited for preparing blood agars. Also in vaccine preparation it can be used for rapid and luxuriant growth as desired for large scale manufacturing of vaccines and toxins.
05138
Vegetable extract Beef extract Vegetable Extract is a meat-free alternative to traditional peptones.With a wide distribution of peptides it meets the requirements for replacing Peptone from meat, enzymatic digest (Fluka 82962) to yield maximum growth of fastidious microorganisms.
04316
Vegetable extract no 1 Meat extract Vegetable Extract no 1 is a meat-free alternative to traditional peptones. With a wide distribution of peptides it meets the requirements for replacing meat extract (Fluka 70164) to yield maximum growth of fastidious microorganisms.
49869
Vegetable extract no 2 Liver extract powder Vegetable extract No. 2 is a specially prepared dehydrated extract of vegetable proteins. Growth response of this vegetable extract is comparable to Liver extract powder (Fluka 70165). It can be employed for the cultivation of fastidious anerobic bacteria such as Brucella and Clostridia by adding to Thioglycollate media. It can also be incorporated in Blood Agar Base for the cultivation of a wide variety of pathogenic microorganisms.
07436
Vegetable hydrolysate no 2 Liver hydrolysate Vegetable hydrolysate no 2 is a meat-free alternative to traditional peptones.With a wide distribution of peptides it meets the requirements for replacing Liver Powder, dehydrated (Fluka 70165) to yield good growth of fastidious anaerobic bacteria such as Clostridia, Bacteroides and Brucella.
67381
Vegetable infusion powder Heart infusion powder Vegetable infusion powder is dehydrated infusion obtained from vegetable proteins under controlled conditions. Growth supporting properties of this infusion is comparable with Heart Infusion Powder (Fluka 57462). It can be used in media employed for cultivation of fastidious organisms like Brucella, Mycoplasma, Pneumococci, Gonococci, Actinomycetes, fungi etc. and antibiotic sensitivity tests.
95757
Vegetable special infusion powder Brain heart infusion A dehydrated infusion from vegetable proteins under controlled conditions. Growth supporting properties of this infusion is comparable with Infusion powder from bovine heart (Fluka 57462). It is suitable for cultivation of fastidious organisms, for the fermentative production of vaccines and for preparing Blood Agar.
Tested Microorganisms in Cultivation Medium with Plant Peptones
18332
Peptone (veget.)
x
x
x
x
51841
Peptone (veget.) acid hydrolys.
x
x
x
x
x
x
19942
Peptone (veget.) no 1
x
x
61854
Peptone (veget.) no 2
x
x
x
x
x
92976
Peptone special (veget.)
x
x
29185
Peptose Peptone (veget.)
x
x
x
x
x
x
x
x
16922
Tryptone (veget.)
x
x
x
x
x
x
12331
Tryptose (veget.)
x
x
x
x
05138
Veget. extract
x
x
x
x
04316
Veget. extract no 1
x
x
x
x
x
49869
Veget. extract no 2
x
x
x
x
x
07436
Veget. hydrol. no 2
x
x
x
x
67381
Veget. infusion powd.
x
x
x
x
x
x
x
x
95757
Veget. special infusion powd.
x
x
x
x
x
x
x
83059
Peptone from potatoes
x
x
x
x
x
93491
Peptone from broad bean
x
x
93492
Peptone from wheat
x
x
  </