| Introduction:
Rapid detection, identification and quantification of microorganisms in beverages,
food and water |
The new HybriScan Test System, which uses sandwich hybridization, provides fast, sensitive and reliable detection, identification and quantification of spoilage and pathogenic microorganisms in beverages, food and water. It is ideal for the comprehensive and reliable routine control of raw materials and concentrates in all production steps up to the quality check of finished goods. HybriScan is a simple, time-saving assay that can be performed with standard laboratory equipment (see Figure 1).
Figure 1: Work flow of HybriScan test method
| Benefits over conventional detection methods and PCR |
HybriScan has significant time- and labor-saving benefits over traditional methods. It also has benefits over PCR and real time PCR, which, although highly sensitive, are susceptible to experimental interferences, like template inhibition from insufficient purification [1], and lack quantification accuracy due to biases associated with PCR and reverse transcription reactions (a general accepted error connected to these methods). In contrast, the HybriScan method is nearly independent of the influences of sample matrix and is able to distinguish between live and dead cells. It also permits the detection of non-culturable microbes. Table 1 compares the benefits and disadvantages of the various methods.
| Advantages of HybriScan over other detection techniques |
| Detection technology |
Advantage |
Disadvantage |
| HybriScan |
- differentiates live/dead cells
- minimal interference by sample matrix
- high specificity
- low cross-reactivity
- easy handling
- cost-efficient read-out devices
- quantitative and qualitative
- high sample throughput (96-microwell plates)
- detects of non-culturable microbes
|
- no differentiation of serotypes or subspecies
- limited probe design (rRNA target)
|
| PCR |
- high sample throughput
- sensitive
- quantitative
|
- no live/dead cell differentiation
- sensitive to matrix interference (high extraction effort)
- susceptible to polymerase inhibition
|
| ELISA |
- differentiation of serotypes or subspecies
- high sample throughput (96-microwell plates)
- quantitative and qualitative
|
- low sensitivity
- low specificity, higher cross-reactivity
- slow and expensive assay development
|
| Conventional cultivation-based methods |
- relatively inexpensive
- simple
- specific
- widely accepted method
|
- time-consuming (up to 10 days)
- no detection of non-culturable microbes
- low sample throughput
- laborious
|
| Principles of the HybriScan method |
The HybriScan method is based on the detection of rRNA via hybridization events and specific capture and detection probes (Figure 2). Sandwich hybridization assays from crude cell samples or in connection to PCR have been extensively used in clinical diagnostics for detection of nucleic acids from bacteria [3, 9, 10] and viruses [11]. Specificity is achieved by targeting conserved or unique rRNA sequences. A labeled capture probe is used to immobilize the target sequence on a solid support plate (coated microtiter plate). A labeled detection probe provides an enzyme-linked optical signal read out. Detection results from application of antibody labeled enzyme. The bound complex is visualized by chromogenic substrates. Photometric data are measured at 450 nm and compared with standard solutions. The HybriScan software enables easy measurement and data analysis.
Figure 2: Principle of the HybriScan sandwich hybridization assay.
| Sensitivity, specificity, flexibility and applicability of HybriScan technology |
Sandwich hybridization is very sensitive, detecting attomoles of the respective target rRNA molecules. [2] The ideal hybridization target for bacteria and yeast is rRNA. These cells contain a large number of rRNA-containing ribosomes; a single cell therefore contains several thousand copies of rRNA but only one DNA. Sandwich hybridization also provides sensitivity in crude biological samples because it is not susceptible to matrix interference.
By using specific probes, HybriScan allows flexible group- and species-specific detection. It is applicable to many analytical fields, including monitoring the microbial content of beer, wine, non-alcoholic beverages, drinking water, a wide variety of foods and wastewater. HybriScan rapidly and accurately identifies, detects and quantifies many important pathogenic species, including Salmonella, Campylobacter, Listeria and Legionella including the most relevant species L. pneumophila. [3, 4, 5]
| HybriScan Listeria monocytogenes: Rapid and innovative test system |
One of the most important foodborne pathogens is Listeria monocytogenes (Figure 3), which poses a health threat in foods that have long, refrigerated shelf lives. [6] Listeriosis, caused by ingestion of foods contaminated with Listeria monocytogenes, has increased dramatically in recent years, causing a great deal of distress and even death. Milk, cheese, ice cream and meat contaminated with this pathogen have led to recent outbreaks of listeriosis. [7] L. monocytogenes proliferates at refrigeration temperatures and is able to grow over a wide pH range from 4.39 to 9.40. These are important characteristics particularly with regard to food safety [12].
Figure 3: Listeria monocytogenes colonies grown on PALCAM agar. HybriScan Listeria monocytogenes permits rapid identification of suspect colonies within one hour.
Conventional culture-based methods to detect L. monocytogenes generally involve selective enrichment followed by culturing on selective medium, isolation and biochemical identification. [8] This laborious and time-consuming approach often takes several days to show results. Also, compared to molecular biological and immunological methods, culture-based methods often give false negatives.
HybriScan Listeria monocytogenes is an excellent alternative to lengthy culture-based methods. It is as reliable and comprehensive as classical methods, but permits rapid detection and quantification with results available within 48 hours. The species-specific probe permits direct detection of L. monocytogenes, thereby eliminating false positives caused by other Listeria species. Even more compelling, suspected single colonies can be identified within one hour using the HybriScanI identification kit without need for further cultivation. Figure 4 shows the validation results of HybriScan Listeria monocytogenes. Food samples were analyzed with the HybriScan method and compared to the culture-based method according to § 64 LFGB. Five different food categories were tested. Results of validation showed a relative accuracy of 99.2 %, relative specificity of 98.5% and relative sensitivity of 99.6%.
Two versions are available. HybriScanI Listeria monoytogenes is used for the extremely rapid, sensitive and economical identification of suspect colonies of L. monocytogenes. HybriScanD Listeria monocytogenes is used for the detection, identification and quantification of L. monocytogenes in different food matrixes.
HybriScan kits are the result of a joint project between Sigma-Aldrich and Scanbec GmbH.
Figure 4: Validation of HybriScan Listeria monocytogenes. 355 food samples were analyzed and compared to culture-based method according to § 64 LFGB. The blue values are the number of L. monocytogenes positive tested food samples in each category. Validation was according to ISO 16140:2003 (ASU L00.00-22).
| Product list of HybriScan Kits |
| Cat. no. |
Description |
old
Cat. No. |
old Name |
Reactions |
Add to Cart |
| Detection Kits |
| 62533 |
HybriScanD Beer |
FS G 01 |
FastScan Beer |
96 |
|
| 68301 |
HybriScanD Drinks |
FS G 02 |
FastScan Drinks |
96 |
|
| 96343 |
HybriScanD E. coli |
FS F 05 |
FastScan E. coli |
96 |
|
| 59744 |
HybriScanD Lactobac |
FS G 04 |
FastScan Lactobac |
96 |
|
| 16593 |
HybriScanD Legionella |
FS W 01 |
FastScan Legionella |
96 |
|
| 07190 |
HybriScanD Legionella pneumophila |
FS W 02 |
FastScan Legionella pneumophila |
96 |
|
| 55661 |
HybriScanD Listeria |
FS F 03 |
FastScan Listeria |
96 |
|
| 49699 |
HybriScanD Listeria monocytogenes |
FS F 04 |
FastScan Listeria monocytogenes |
96 |
|
| 55662 |
HybriScanD Salmonella |
FS F 01 |
FastScan Salmonella |
96 |
|
| 02349 |
HybriScanD Total Bacterial Count |
FS B 01 |
FastScan Total Bacterial Count |
96 |
|
| 04447 |
HybriScanD Waste Water Microthrix parvicella |
FS W 04 |
FastScan Waste Water Microthrix parvicella |
96 |
|
| 78436 |
HybriScanD Waste Water Total Bacterial Count |
FS W 03 |
FastScan Waste Water Total Bacterial Count |
96 |
|
| 61397 |
HybriScanD Yeast |
FS G 03 |
FastScan Yeast |
96 |
|
| Identification Kits |
| 79742 |
HybriScanI Brettanomyces |
FS I 09 |
FastScan Brettanomyces |
48 |
|
| 19503 |
HybriScanI Candida albicans |
FS I 08 |
FastScan Candida albicans |
48 |
|
| 76545 |
HybriScanI E. coli |
FS I 04 |
FastScan E. coli |
48 |
|
| 75724 |
HybriScanI Lactobacillus brevis |
FS I 10 |
FastScan Lactobacillus brevis |
48 |
|
| 80065 |
HybriScanI Lactobacillus buchneri |
FS I 12 |
FastScan Lactobacillus buchneri |
48 |
|
| 86827 |
HybriScanI Lactobacillus lindneri |
FS I 11 |
FastScan Lactobacillus lindneri |
48 |
|
| 49417 |
HybriScanI Legionella pneumophila |
FS I 06 |
FastScan Legionella pneumophila |
48 |
|
| 77007 |
HybriScanI Leuconostoc |
FS I 13 |
FastScan Leuconostoc |
48 |
|
| 49712 |
HybriScanI Listeria monocytogenes |
FS I 01 |
FastScan Listeria monocytogenes |
48 |
|
| 42875 |
HybriScanI Megasphaera |
FS I 17 |
FastScan Megasphaera |
48 |
|
| 33018 |
HybriScanI Pectinatus cerevisiiphilus |
FS I 16 |
FastScan Pectinatus cerevisiiphilus |
48 |
|
| 73582 |
HybriScanI Pectinatus frisingensis |
FS I 15 |
FastScan Pectinatus frisingensis |
48 |
|
| 67289 |
HybriScanI Pediococcus damnosus |
FS I 14 |
FastScan Pediococcus damnosus |
48 |
|
| Auxiliary Equipment |
| 44492 |
HybriScan Software |
|
Table 2: HybriScan products (HybriScanD = detection kit; HybriScanI = identification kit)
- Bustin, S.A. Absolute quantification of mRNA using real-time reverse transcription polymerase chain reaction assays. J. Mol. Endocrinol. 2000, 25, 169-193.
- Tenhunen, J.; Eloranta, J.; Kallio, A.; Soderlund, H. A solution hybridization method for quantification of mRNAs: determining the amount and stability of oncogene mRNA. Genet. Anal. Tech. Appl. 1990, 7, 228-233.
- Huhtamella, S.; Leinonen, M.; Nieminen, T.; Fahnert, B.; Myllykoski, L.; Breitenstein, A.; Neubauer, P. RNA-based sandwich hybridisation method for detection of lactic acid bacteria in brewery samples. J. Microbiol. Methods 2007, 68(3), 543-53.
- Leskela, T.; Tilsala-Timisjarvi, A.; Kusnetsov, J.; Neubauer, P.; Breitenstein, A. Sensitive genus-specific detection of Legionella by a 16S rRNA based sandwich hybridization assay. J. Microbiol. Methods 2005, 62(2), 167-79.
- Rautio, J.; Barken, KB.; Lahdenpera, J.; Breitenstein, A.; Molin, S.; Neubauer, P. Sandwich hybridisation assay for quantitative detection of yeast RNAs in crude cell lysates. Microb. Cell Fact. 2003, 2(1), 4-12.
- Mellefont, L.A.; McMeekin, T.A.; Ross, T. Effect of relative inoculum concentration on Listeria monocytogenes growth in co-culture. Int. J. Food Microbiol. 2008, 121, 157-168.
- McLauchlin, J. The relationship between Listeria and listeriosis. Food Control 1996, 7, 187-193.
- Donnelly, C.W. Detection and isolation of Listeria monocytogenes from food samples: implications of sublethal injury, J. AOAC Int. 2002, 85, 495-500.
- Casademont I, Bizet C, Chevrier D and Guesdon JL Rapid detection of Campylobacter fetus by polymerase chain reaction combined with non-radioactive hybridization using an oligonucleotide covalently bound to microwells. Mol Cell Probes 2000, 14, 233-240
- Chevrier D, Popoff MY, Dion MP, Hermant D and Guesdon JL Rapid detection of Salmonella subspecies I by PCR combined with non-radioactive hybridisation using covalently immobilised oligonucleotide on a microplate. FEMS Immunol Med Microbiol. 1995, 10(3-4), 245–501.
- Albretsen C, Kalland KH, Haukanes BI, Havarstein LS and Kleppe K Applications of magnetic beads with covalently attached oligonucleotides in hybridization: isolation and detection of specific measles virus mRNA from a crude cell lysate. Anal. Biochem. 1990, 189, 40-50
- Uyttendaele, M., De Troy, P., Debevere, J. Incidence of Salmonella, Campylobacter jejuni, Campylobacter coli, and Listeria monocytogenes in poultry carcasses and different types of poultry products for sale on the Belgian retail market. Journal of Food Protection 1999, 62, 735–740.
| HybriScan Waste Water Calculator |
For the Waste Water Kits
(04447 HybriScanD Waste Water Microthrix parvicella,
78436 HybriScanD Waste Water Total Bacterial Count )
we provide an excel sheet for calculation of total bacterial count, Microthrix count and ratio of Microthrix : Total Bacterial Count.
For download the file please click on the links below.
|