Analytical / Chromatography
Air Monitoring
Analytical Reagents
Analytical Standards
Gas Chromatography
HPLC
Microbiology
Basic Ingredients
Identification Tests
Microbiology Manual
Product Lines
Bacillus anthracis
Culture Media Index
Equipment
Ferrioxamine E
Prepared Plates
Pseudomonas Agar
Signaling Compounds
Vegitone Media
Quality Assurance
Theory
Microscopy
Sample Preparation
Spectroscopy
Syringes
Titration
TLC
Vials
Videos
Events
Literature
Analytix
The Reporter
Reporter Europe
 HiFluoro Pseudomonas Agar Base
Microbiology
By Jvo Siegrist, Product Manager Microbiology, isiegris@europe.sial.com
HiFluoro Pseudomonas Agar Base is used as a selective medium for the isolation of Pseudomonas aeruginosa from diverse sample. Pseudomonas aeruginosa breaks the fluorogenic compound to release the fluorogen which produces a visible fluorescence under long wave UV light.
HiFluoro Pseudomonas Agar Base is formulated based on the formula described by King et al. (1) expect fluorogenic mixture. The medium is reach in essential growth nutrients. Magnesium chloride and Potassium sulfate stimulate pyocyanin production. Cetrimide (Cetyltrimethylammonium bromide) is incorporated in the medium to inhibit bacteria other than Pseudomonas aeruginosa. It acts as a quaternary ammonium compound, cationic detergent which causes nitrogen and phosphorus to be released from bacterial cells other than Pseudomonas aeruginosa. Pseudomonas aeruginosa breaks the fluorogenic compound to release the fluorogen which produces a visible fluorescence under long wave UV light.
Pseudomonas aeruginosa on HiFluoro Pseudomonas Agar Pseudomonas aeruginosa on HiFluoro Pseudomonas Agar under UV light
References:
  1. 1. E.O. King, et al., Media for the demonstration of pyocyanin and fluorescein, J. Lab. Clin. Med. 44, 301 (1954)
  2. G.J. Bonde, Bacterial Indicators of Water Pollution, (1962)
  3. L.C.D Formiga, New possibilies for the laboratory diagnosis of diphtheria, Brazilian J. Med. Biol. Res., 18, 401 (1985)
  4. D.J. Blazevic, M.H. Köpcke, J.M. Matsen, Incidence and identification of Pseudomonas fluorescens and Pseudomonas putida in the clinical laboratory, Appl. Microbiol., 25, 107 (1973)
For further fluorogenic media click here
Back to top
Microbiology Home