|
No more problems with animal derived compounds in media. Approved new media with plant peptones and synthetic detergents for all kind of microorganisms.
In the Analytix Issue 3 (2005) we wrote about plant peptones as a good alternative to animal derived peptones. Meanwhile we can provide you a nice range of approved dehydrated media with plant peptones and synthetic detergent as replacement for bile salts. We can assure that this media are free from BSE and GMO! As well we have seen higher recovery rates and better growth performance with plant peptones. The new developed media are just a choice of some different kind of applications and microorganisms. If there is demand for other important formulations we are open to check the possibility to provide you a new composition. Because of less import and export problems and costs we can provide you also a lower price for this media than for the equivalent with animal derived components.
| Product # |
Product Name |
Species |
Add to Cart | | 41159 |
Actinomyces Agar, Vegitone |
Actinomyces |
|
| 40834 |
Actinomyces Broth, Vegitone |
Actinomyces |
|
| 28943 |
Andrade peptone water, Vegitone |
Citrobacter, Escherichia coli, Klebsiella, Proteus, Providencia, Salmonella, Shigella, Yersinia & other coliforms |
|
| 19057 |
Lactose Broth, Vegitone |
Escherichia coli & Coliforms |
|
| 19344 |
LB Agar, Vegitone |
Escherichia coli, diverse other bacteria |
|
| 28713 |
LB Broth, Vegitone |
Escherichia coli, diverse other bacteria |
|
| 19352 |
Mac Conkey Agar No 1, Vegitone |
Escherichia coli, Proteus, Salmonella, Shigella & other coliforms, Staphylococci, Enterococci |
|
| 38954 |
Malt Extract Agar modified, Vegitone |
Aspergillus, Penicilliums, Saccharomyces, Pichia, Zygosaccharomyces, Candida, diverse other moulds |
|
| 07012 |
M-FC Agar modified for Klebsiella, Vegitone |
Escherichia coli, Klebsiella & other coliforms |
|
| 43291 |
M-FC Agar, Vegitone |
Escherichia coli & other coliforms |
|
| 49522 |
M-Lauryl Sulfate Broth, Vegitone |
Escherichia coli, Salmonella & other coliforms |
|
| 41782 |
MRS Agar, Vegitone |
Lactobacilli |
|
| 38944 |
MRS Broth modified, Vegitone |
Lactobacilli |
|
| 04163 |
Nutrient Agar No 2, Vegitone |
diverse moulds and bacteria |
|
| 16336 |
Nutrient Broth No 3, Vegitone |
diverse bacteria |
|
| 15776 |
PALCAM Listeria Selective Agar, Vegitone |
Listeria |
|
| 59859 |
PALCAM Listeria Selective Enrichment Broth, Vegitone |
Listeria |
|
| 40893 |
Peptone Water, phosphate-buffered, Vegitone |
Citrobacter, Escherichia coli, Klebsiella, Proteus, Providencia, Salmonella, Shigella, Yersinia & other coliforms, Staphylococci, Streptococci, Pseudomonas, diverse other bacteria |
|
| 19718 |
Plate Count Agar, Vegitone |
diverse moulds and bacteria |
|
| 14432 |
Tryptic Soy Agar, Vegitone |
diverse moulds and bacteria |
|
| 41298 |
Tryptic Soy Broth, Vegitone |
diverse moulds and bacteria |
|
| 39964 |
Tryptone water, Vegitone |
Escherichia coli, Enterococci, Klebsiella, diverse other bacteria |
|
| 41960 |
Vegitone Infusion Broth |
Coliforms, Neisseria, Staphylococci, Streptococci, diverse other bacteria |
|
| 42376 |
Violet Red Bile Agar, Vegitone |
Escherichia coli & other coliforms |
|
| 53605 |
Violet Red Bile Glucose Agar without Lactose, Vegitone |
Escherichia coli, Salmonella, Shigella & other coliforms |
|
| Back to top |
References:
- Analytix Issue 3, Sigma-Aldrich (2005)
- www.sigma-aldrich.com/microbiology
Back to top
Microbiology Home
|