|
Microrganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the environment, microbes have adapted to the habitats most suitable for their needs, in the laboratory, however, these requirements must be met by a culture medium. This is basically an aqueous solution to which all the necessary nutrients have been added. Depending on the type and combination of nutrients, different categories of media can be made.
Categories
Complex media are rich in nutrients, they contain water soluble extracts of plant or animal tissue (e.g., enzymatically digested animal proteins such as peptone and tryptone). Usually a sugar, often glucose is added to serve as the main carbon and energy source. The combination of extracts and sugar creates a medium which is rich in minerals and organic nutrients, but since the exact composition is unknown, the medium is called complex.
Defined media are media composed of pure ingredients in carefully measured concentrations dissolved in double distilled water i.e., the exact chemical composition of the medium is known. Typically, they contain a simple sugar as the carbon and energy source, an inorganic nitrogen source, various mineral salts and if necessary growth factors (purified amino acids, vitamins, purines and pyrimidines).
Selective/differential media are media based on either of the two categories above supplemented with growth-promoting or growth-inhibiting additives. The additives may be species- or organism-selective (e.g., a specific substrate, or an inhibitor such as cyclohexamide (artidione) which inhibits all eucaryotic growth and is typically used to prevent fungal growth in mixed cultures).
| Media |
Purpose |
|
| Complex |
Grow most
heterotrophic organisms |
| Defined |
Grow
specific heterotrophs and are often mandatory for chemoautotrophs, photoautotrophs and for
microbiological assays |
| Selective |
Suppress unwanted
microbes, or encourage desired microbes |
Differential
|
Distinguish
colonies of specific microbes from others |
| Enrichment |
Simlar to
selective media but designed to increase the numbers of desired microorganisms to a
detectable level without stimulating the rest of the bacterial population |
| Reducing |
Growth of
obligate anaerobes |
|
The mixture of necessary nutrients can be used as a liquid medium, or a solidifying agent can be added. "Agar agar" is a natural polysaccharide produced by marine algae and is the most commonly used solidifying agent added to media (end concentration usually 1.5 % w/v). If hydrolysis of the agar is suspected, a silica gel is used as a replacement solidifying agent.
Aseptic Techniques
Before inoculation with the desired microorganisms, microbiological media and all materials coming into contact with it must be sterile. During any subsequent handling of the bacterial cultures, unwanted or contaminant organisms must be excluded employing aseptic techniques.
Sterilisation implies the complete destruction of all microorganisms including spores, this is accomplished by the use of heat, chemicals, radiation, filtration.
Heat
Denatures and coagulates vital proteins. There are various forms of heat sterilisation.
Red Heat
Inoculating wires or loops are sterilised by holding them in a Bunsen flame until they are red hot.
Moist Heat
Bacteria are more readily destroyed by moist heat (steam) than dry heat. Usually used for the sterilisation of culture media, aqueous solutions and the destruction of discarded cultures. Air must first be removed in order to achieve the 121 °C necessary for successful sterilisation. This is accomplished by the use of an autoclave (the technical version of a pressure cooker), which follows automatic cycles of heating under pressure for the required time.
Dry Heat
Usually employed for materials which could either be corroded by steam or must remain dry before use. These include metal instruments, glass petri dishes, flasks and pipettes and cotton wool. In practice, dry heat sterilisation requires longer time intervals and higher temperatures than steam sterilisation, e.g.steam sterilisation 121°C for 15mins or dry heat sterilisation 160°C for 120 minutes.
Chemical
Usually employed for delicate equipment such as optical instruments and electrical devices which would be damaged by heat. Due to the toxicity of the chemicals used, this is not the most popular form of sterilisation. Chemicals employed include: gaseous ethylene oxide, which alkylates amino, sulfhydryl, carboxyl and hydroxyl groups of microbial cell compounds; formaldehyde, used as a fumigant; and hydrogen peroxide vapour used in aseptic packaging.
Radiation
Employed for heat-sensitive materials and for environmental samples such as soil and sediment where structural changes caused by heat need to be avoided. Two forms of radiation are used:
UV
Initiates the excitation of atoms which in nucleic acids leads to fatal mutations. UV light cannot penetrate materials so is used mainly for surface treatments e.g. Iaminar flow benches, and air and water.
Ionizing Radiation
Can penetrate samples, causing ionization within cells. Gamma radiation generated through a 60Co a-source is used to sterilise complex matrices such as soil and foodstuff. Microorganisms show increased resistance to radiation under anoxic conditions (2-5x) and also in frozen samples.
Filtration
Filtration sterilisation operates through the exclusion rather than destruction of microorganisms. It is safe for the user and is employed for sensitive liquids and gases. Three types of filters are currently in use:
Depth Filters
These are made of columns packed with fibrous materials such as glass wool or cotton wool. The twisting and turning fibres entrap particles and so act as filters; they show little resistance to flow and are used mainly for gases or as pre-filters for membrane filters which are easily clogged.
Membrane Filters
Act by screening out particles. Their effectiveness depends on the size of the membrane pores and the electrostatic attractions present. The most commonly used filters in microbiology are usually made of cellulose acetate or cellulose nitrate.
Size of filter pores required to screen out:
Yeast 0.45 -1.2 µm
Bacteria 0.2 µm
Viruses and mycoplasmas 0.01-0.1µm
Membrane filtration is usually employed for heat-sensitive substances, e.g. vitamin solutions; the filters are heat-sterilised before use.
Nucleation Track (Nuclepore) Filters
These filters consist of very thin polycarbonate films which have been treated with nuclear radiation and then etched with a chemical to create very uniform vertical holes. They are employed for the same material as membrane filters but have the disadvantage that they are more easily clogged.
Media Supplements
Table of the most commonly used Media Supplements, Methods of Sterilisation, Solubilities
| Substance |
Solubility
in Water at 25°C |
Comments/Sterilisation |
|
Actidione
(Cyclohexamide) |
2.1
g/100 ml |
Destroyed
by boiling in aqueous solution at pH 7 for 1 hr. Filter sterilise |
| L(+)-Arabinose |
1 g/1 ml |
|
| Arginine* |
15
g/100 ml |
Sat.
solution bly alkaline, absorbs CO2 pK1, 2.18 pK2 9.09
pK3 13.2 |
| Asparagine* |
2.16
g/100 ml |
Stable
in aq sol. at 100°C, hydrolysed in acidic solutions |
| Biotin |
22
mg/100 ml |
pH of
0.01% sol. = 4.5
acidic solutions can be heat-sterilised |
| Cysteine* |
soluble |
Neutral
slightly alkaline solution is oxidised to cystine pK1, 1.71 pK2 8.33 pK3
10.78 |
| Dextrin |
soluble in 3
parts boiling water |
|
| Ehrlichs reagent
Fructose |
soluble
soluble |
|
| Fuchsin |
1 g/7 ml |
|
| Galactose |
|
a) soluble in 0.5 parts
water, freely soluble in hot water
b) soluble
in 1.7 parts water at 17°C |
| Glucose |
1 g/1 ml |
pH of 0.5 M aq solution = 5.9 |
| Glycerol |
miscible |
|
| Glycogen |
soluble |
with opalescence |
| Lactose |
21.6 g/100 ml |
a) 1 g/5 ml
b) 1 g/2.2
ml at 15°C |
| Maltose |
soluble |
mp 102-103°C |
| Mannitol |
soluble |
|
| Niacin |
soluble |
Stable to
autoclaving at 120°C for 20 mins |
| Ornithine* |
soluble |
Aqueous solution
alkaline pK, 1.94 pK2 8.65 pK3 10.76 |
| Phenylalanine* |
|
L 29.6 g/L at
25°C
D 1 g/35.5ml at 16°C
DL 14.11 g/L at 25°C
pK1 2.58 pK2 9.24 |
| Resazurine |
insoluble
in water
soluble in dil alkali hydroxides |
Indicator
0.1 g in 20ml 1 N NaOH + water up to 500 ml
pH 3.8 = orange, pH 6.5 dark violet |
| Ribose |
soluble |
|
| Citric acid |
soluble 59.2 % at
20°C |
pH of 0.1 N
solution = 2.2 |
| Sorbitol |
soluble Up to83% |
mp 100/112°C |
| Starch |
insoluble |
|
| Sucrose |
1g/0.5ml |
|
| EDTA (disodium
salt) |
soluble |
Used to complex
iron in media |
| EDTA (acid) |
insoluble |
|
All stock solutions of amino acids can be autoclaved at 120°C for 20 mins.
Interaction of Media Components
When undertaking research where medium composition plays an important role, e.g. toxicity studies, care must be taken to observe the various interactions which can take place between media components, for example;
|
Yeast,
peptone
and
amino acids |
Bind large amounts of divalent Hg, Pb, Ag and
Cu ions |
| Glucose |
exhibits
non-selective metal binding which can cause metal deficiency of essential ions or decrease
the toxicity of toxic ions. |
| EDTA |
chelates
essential trace metals causing deficiency; the effect can be reversed by the addition of
other metals e.g. Cu and Fe. |
| NaCI |
increases
Zn toxicity (formation of a Zn-chloro complex), decreases Cd toxicity. |
| Agar |
in
solid medium, Sn toxicity increases (formation of a soluble toxic tinagar complex). |
| Silica
gel |
decreases
toxicity of Sn, Cd, Pb, Ni and Zn divalent ions. |
| Phosphate |
precipitation*
of insoluble phosphates decreasing toxicity of toxic ions |
|
(e.g
Cd and Pb) or causing deficiency of essential ions (e.g. Fe) |
| Carbonate |
precipitation
of insoluble carbonates
decreased Pb toxicity |
| Citrate |
non-selective
metal binding |
| Tris |
non-selective
metal binding |
|
* Can be avoided by addition of a HEPES buffer to reduce phosphate.
Back to top
Microbiology Home
|