Analytical / Chromatography
Air Monitoring
Analytical Reagents
Analytical Standards
Gas Chromatography
HPLC
Microbiology
Microscopy
Sample Preparation
SPE
Diffusive Sampling
What is it
What is Radiello
How to use
Applications
Analytical Service
Products & Accessories
Literature/Downloads
Brochure and CD
Site Map
SPME
VersaFlash System
Versapure Büchner Funnels
Spectroscopy
Syringes
Titration
TLC
Vials
Videos
Events
Literature
Analytix
The Reporter
Reporter Europe
 How to use radiello®
radiello® diffusive sampling System
radiello Parts and Components
The essential parts of radiello are the adsorbing cartridge, the diffusive body, the supporting plate and the with the bar code indication. Apart from the adsorbing cartridge, all of the other components can be used multiple times.
The adsorbing cartridges for thermo desorption can also be reused.
(1) Open the plastic bag, draw the adsorbent cartridge out from the tube and put it in the diffusive body. The lower part of the diffusive body holds a seat for the central positioning of the cartridge (see right). The cartridge must be inserted fully into the diffusive body.
(2) Keeping the diffusive body in a vertical position, screw it onto the supporting plate
(3) Insert a label in the pocket without peeling it off
(4) Place assembled device on worker or location to be monitored. Keep note of the date and time and expose radiello .
Sampling has started!
(5) Keep note of the date and time of the end of exposure. Place the cartridge into the tube, peel off the label and stick it onto the tube such that the barcode is parallel to the axis of the tube. Forward the ttube with the adsorber cartridge to the lab for analysis.
(6) Analytes are desorbed from the radiello tube using either solvent desorption (CS2, CH3CN, etc.) or thermal desorption. Spent cartridges are discarded. (Thermal desorption cartridges can be reused.)
Complete analytical protocols for each analyte class are supplied in the radiello Manual.
Back to Top
Back to radiello diffusive sampling Home