Analytical / Chromatography
Air Monitoring
Analytical Reagents
Analytical Standards
Gas Chromatography
HPLC
Microbiology
Microscopy
Sample Preparation
SPE
Diffusive Sampling
What is it
What is Radiello
How to use
Applications
Analytical Service
Products & Accessories
Literature/Downloads
Brochure and CD
Site Map
SPME
VersaFlash System
Versapure Büchner Funnels
Spectroscopy
Syringes
Titration
TLC
Vials
Videos
Events
Literature
Analytix
The Reporter
Reporter Europe
 Calibration Solutions and Kits
radiello® Products & Accessories
Calibration solutions
     -H2S (Hydrogen Sulfide) Calibration Standard
     -Aldehyde Calibration Standard
Calibration kits
     -BTEX Calibration Kit (CS2 Desorption)
     -BTEX Calibration Kit (Thermal Desorption)
     -VOCs Calibration Kit (Workplace Environment)
Calibration solutions
H2S (Hydrogen Sulfide) Calibration Standard
Methylene blue concentrate that, once diluted 1:50, provides the same absorbance value of hydrogen sulfide at 665 nm at a concentration of 1.145 μg·ml-1 sulfide ions. This concentration value is the highest absorbance value with the linear range of the spectrophotometer and can be used as the stock solution to prepare standards for the calibration curve. It is suitable for preparing 50 calibration curves using the recipe described in the table. The standard is stable for at least one year.
Solution
mL of
mL of water
Equivalent to μg/mL of S2-
A
2 of code 171
98
1.145
B
25 of A
25
0.572
C
10 of A
40
0.229
D
5 of A
45
0.115
radiello Code
Description
Pkg Qty.
Supelco
Cat. No.
Add to Cart
171
H2S Methylene Blue Calibration Standard, 100 mL
1
Aldehyde Calibration Standard
The Aldehyde Calibration Standard consists of nine 2,4-dinitrophenylhydrazones (2,4-DNPH) diluted in 10 mL acetonitrile. Certified concentrations of each compound are described in the table. Actual concentrations are certified for each lot. The standard stock solution is shipped in a pierceable-septum crimped cap vial, and is stable for at least 4 months when stored in the dark at 4°C.
2,4-DNPH of
μg/mL as aldehyde
Formaldehyde
50
Acetaldehyde
50
Acrolein
10
Propanal
50
Butanal
50
Isopentanal
50
Pentanal
50
Hexanal
50
Benzaldehyde
50
radiello Code
Description
Pkg Qty.
Supelco
Cat. No.
Add to Cart
302
Aldehyde Calibration Standard, 10mL
1
radiello Calibration kits
The kits consist of twelve cartridge adsorbents (Cat. No. RAD130 or RAD145). Three of which are blanks and nine preloaded, divided into three concentration groups. Concentrations are described in the listed tables. The values shown are indicative. Actual concentrations are certified for each lot. The cartridges are stable for at least four months when stored at 4°C.
BTEX Calibration Kit (CS2 Desorption)
The BTEX Calibration Kit (CS2 Desorption) is designed for the analysis of BTEX in urban environments. The kit may be used for both routine calibration and quality control. The kit consists of twelve Code 130 Cartridge Adsorbents (Cat. No. RAD130). The BTEX preloaded simulate 7 day exposures (100,080 minutes).
code 405
simulated concentrations in μg·m-3 (7 days exposure equivalent)
Group 1
Group 2
Group 3
benzene
1
10
50
toluene
2
20
100
ethylbenzene
1
10
50
m-xylene
1
10
50
p-xylene
1
10
50
o-xylene
1
10
50
radiello Code
Description
Pkg Qty.
Supelco
Cat. No.
Add to Cart
405
BTEX Calibration Kit (CS2 Desorption)
1
BTEX Calibration Kit (Thermal Desorption)
The BTEX Calibration Kit (thermal desorption) was designed for the analysis of BTEX in urban environments followed by thermal desorption. The kit may be used for both routine calibration and quality control. The kit consists Code 145 Cartridge Adsorbents (Cat. No. RAD145). The BTEX preloaded simulate 7 day exposures (100,080 minutes). BTEX is spiked on the cartridges by injecting vaporized BTEX standards in methanol under nitrogen flow. A methanol peak may be visible during chromatographic analysis.
code 407
simulated concentrations in μg·m-3 (7 days exposure equivalent)
Group 1
Group 2
Group 3
benzene
1
5
25
toluene
2
10
50
ethylbenzene
1
5
25
m-xylene
1
5
25
p-xylene
1
5
25
o-xylene
1
5
25
radiello Code
Description
Pkg Qty.
Supelco
Cat. No.
Add to Cart
407
BTEX Calibration Kit (Thermal Desorption)
1
VOCs Calibration Kit (Workplace Environment)
The VOC Calibration Kit is ideal for conducting scheduled quality control runs when analyzing workplace environments. The kit consists of Code 130 Cartridge Adsorbents (Cat. No. RAD130).
Three of which are blanks and nine, divided into three concentration groups, are preloaded with VOCs to simulate 8 hour exposures (480 minutes). The composition of VOCs represents a broad range of polarity, and the spiked concentrations represent 0.5, 1.0, and 2.0 times the threshold limit values (TLV) for each compound. VOCs are spiked on the cartridges by injecting vaporized VOC standards in CS2 under nitrogen flow.
code 406
simulated concentrations in mg·m-3 (8 hours exposure equivalent)
Group 1
Group 2
Group 3
benzene
0.1
0.2
0.4
toluene
19
38
76
ethylbenzene
12
24
48
m-xylene
12
24
48
p-xylene
12
24
48
o-xylene
12
24
48
butanol
15
30
60
2-ethoxyethyl acetate
2.5
5
10
radiello Code
Description
Pkg Qty.
Supelco
Cat. No.
Add to Cart
406
VOC Calibration Kit (workplace environment)
1
Back to Top
Back to radiello Products & Accessories
Back to radiello diffusive sampling Home