|
Products Accessories
Cartridge Adsorbents
|
Several different cartridge adsorbents are available. Each of which are specific for different classes of compounds. The dimensions for each cartridge are 60 mm L x 4.8 or 5.8 mm diameter. Each cartridge is designed for one time use with the exception of Thermal Desorption (TD) cartridge adsorbents
A sufficient number of adhesive labels with barcode is included with every pack of cartridges for easy tracking of sampling date and time. |
 |
| Compound Class |
radiello Code
|
Description of adorber bed |
Pkg Qty.
|
Supelco Cat. No.
|
Add to cart
|
VOCs, and BTEX CS2 Desorption |
130
|
SS net (100mesh, 5.8mm diam),activated charcoal (35-50 mesh) |
20
|
|
|
VOCs and BTEX Thermo Desorption |
145
|
SS net (3x8um mesh, 4.8mm diam), 350mg Carbograph 4 (35-50 mesh) |
20
|
|
|
| Aldehydes |
165
|
SS net w/ 2,4-DNPH coated florisil |
20
|
|
|
| Ammonia |
168
|
Microporous PE impregnated with phosphoric acid |
20
|
|
|
| Anaesthetic gases/vapors |
132
|
SS net w/ mix of mol sieve & activated charcoal (35-50 mesh) |
20
|
|
|
| HCl |
169
|
SS net w/silica gel (0.1-0.4mm particle size) |
20
|
|
|
| HF, NO2 & SO2 |
166
|
Microporous PE coated w/ wet TEA |
20
|
|
|
| Hydrogen Sulfide |
170
|
Microporous PE impregnated with zinc acetate |
20
|
|
|
| Ozone |
172
|
Micropore PE tube with silica coated with 4,4'-dipyrldylethylene |
20
|
|
|
Phenolic Compounds Thermo Desorption |
147
|
SS net 100 mesh, 4.8mm diam), 250mgTenax-TA (20-35 mesh) |
20
|
|
|
Back to radiello Products & Accessories Back to radiello diffusive sampling Home