Sigma-Aldrich
Fluka & Supelco - Analytical Logo
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
radiello® Products & Accessories
Ready-to-use Samplers
The radiello ready-to-use diffusive samplers come pre-assembled with the cartridge adsorbent pre-contained within the diffusive body, and sealed with a polycarbonate screw-thread cap. To avoid premature sampling, the entire unit (diffusive body with pre-sealed and installed cartridge adsorbent) is enclosed in an airtight polypropylene (PP) container. Just before use, the unit is removed from the PP container, and snapped into the ready-to-use vertical adapter pre-fixed to the triangular support plate. Once sampling is complete, the diffusive sampling unit is removed from the support plate and resealed into the PP container for shipment to the lab.
diffusive_samplers
Each ready-to-use sampler includes: a sampler unit (sealed diffusive body with cartridge adsorbent), glass or plastic tube, ready-to-use vertical adapter, and barcode label and PP container. Please order the triangular support plates (RAD121) separately.
Note that the ready-to-use diffusive samplers are ideal for workplace sampling campaigns, but are not for sampling low concentrations in outdoor or domestic environments. Shelf life of this samplers is 3 months.
Application
radiello Code
Description
Pkg Qty.
Supelco
Cat. No.
Add to Cart
VOCs and BTEX
CS2 Desorption
123-1
Code 120 White Diffusive Body (Cat. No. RAD120) and Code 130 Cartridge Adsorbent (Cat. No. RAD130)
5
VOCs and BTEX
Thermo Desorption
123-2
Code 120-2 Yellow Diffusive Body (Cat. No. RAD1202) and Code 145 (Cat. No. RAD145) Cartridge Adsorbent
5
HF, NO2 & SO2
123-3
Code 120-1 Blue Diffusive Body (Cat. No. RAD1201) and Code 166 Cartridge Adsorbent (Cat. No. RAD166)
5
Aldehydes
123-4
Code 120-1 Blue Diffusive Body (Cat. No. RAD1201) and Code 165 Cartridge Adsorbent (Cat. No. RAD165)
5
Ozone
123-5
Code 120-1 Blue Diffusive Body (Cat. No. RAD1201) and Code 172 Cartridge Adsorbent (Cat. No. RAD172)
5
Hydrogen Sulfide
123-6
Code 120 White Diffusive Body (Cat. No. RAD120) and Code 170 Cartridge Adsorbent (Cat. No. RAD170)
5
Ammonia
123-7
Code 120-1 Blue Diffusive Body (Cat. No. RAD1201) and Code 168 Cartridge Adsorbent (Cat. No. RAD168)
5
HCl
123-8
Code 120 White Diffusive Body (Cat. No. RAD120) and Code 169 Cartridge Adsorbent (Cat. No. RAD169)
5
Back to radiello Products & Accessories
Back to radiello diffusive sampling Home
<