Sigma-Aldrich
Fluka & Supelco - Analytical Logo
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Solid Phase Extraction
Dispersive SPE Tubes
dispersive spe tubes
Dispersive SPE (dSPE), often referred to as the “QuEChERS” (Quick, Easy, Cheap, Effective, Rugged, and Safe) method, is an emerging sample prep technique that is becoming increasingly popular in the area of multi-residue pesticide analysis in food and agricultural products.
In dSPE, food/agricultural samples are first extracted with an aqueous miscible solvent (e.g., acetonitrile) in the presence of high amounts of salts (e.g., sodium chloride and magnesium sulfate) and/or buffering agents (e.g. citrate) to induce liquid phase separation and stabilize acid and base labile pesticides. Upon shaking and centrifugation, an aliquot of the organic phase is subjected to further clean up using SPE. Unlike traditional methods using SPE tubes, in dispersive SPE, clean up is facilitated by mixing bulk amounts of SPE (e.g., Supelclean PSA, ENVI-Carb, and/or Discovery DSC-18) with the extract. After sample clean up, the mixture is centrifuged and the resulting supernatant can either be analyzed directly or can be subjected to minor further treatment before analysis.
Supelco now carries a line of vials and centrifuge tubes containing pre-determined amounts of salts and SPE sorbents to support the most common method configurations used today.
For a limited time, you may request free dSPE sample MultiPaks for method development.
Note: If there is a certain dispersive SPE configuration you require that is not listed, please contact technical service at 800-359-3041/814-359-3041 or email: techservice@sial.com. We may be able to offer the configuration you require as a custom product.
For ordering information, go to our On-Line Catalog
Technical Resources
Subject No.
Pages
File Size I.D.
Dispersive SPE (dSPE) Product Line Data/Instruction Sheet 2 37 Kb T706013
The Extraction & Analysis of Multi-Residue Pesticides in Oranges Using Dispersive SPE & GC-MS (poster) 20 196 Kb T406100
Extraction and Analyses of Agricultural Pesticides from Oranges Using the "QuEChERS" Method (TheReporter 24.3) 3 381 Kb T206003
NEW Custom “Dispersive” SPE Products for Pesticide and Acrylamide Analysis (TheReporter 23.4) 2 383 Kb T205004