Reversed-phase SPE
Ion-exchange SPE
Normal phase SPE
Supelclean™ SPE Method Development Kits
96-Well SPE Method Development Plates
FREE SPE MultiPaks for Method Development
Application Assistance
Our kits consist of an assortment of SPE phase chemistries and cartridge configurations
ideal for SPE method development. The range of phase chemistries available for
each kit allows the user to profile for compund retention, elution and sample
matrix selectivity.
| Supelclean SPE Method Development Kit |
Kit
A |
Kit
B |
Kit
C |
Kit
NP-3 |
Kit
IX-3 |
| Supelclean Packing |
Sorbent Qty.
/ Tube Size |
| LC-Si |
500 mg/3 mL |
100 mg/1 mL |
500 mg/6 mL
1 g/6 mL |
500 mg/3 mL |
|
| LC-8 |
500 mg/3 mL |
100 mg/1 mL |
500 mg/6 mL |
|
|
| LC-18 |
500 mg/3 mL |
100 mg/1 mL |
500 mg/6 mL |
|
|
| LC-CN |
500 mg/3 mL |
100 mg/1 mL |
500 mg/6 mL |
|
500 mg/3 mL |
| LC-Diol |
500 mg/3 mL |
100 mg/1 mL |
|
500 mg/3 mL |
|
| LC-NH2 |
500 mg/3 mL |
100 mg/1 mL |
|
500 mg/3 mL |
500 mg/3 mL |
| LC-Ph |
500 mg/3 mL |
100 mg/1 mL |
|
|
|
| LC-SAC |
500 mg/3 mL |
100 mg/1 mL |
|
|
500 mg/3 mL |
| LC-SCX |
500 mg/3 mL |
100 mg/1 mL |
|
|
500 mg/3 mL |
| LC-WCX |
500 mg/3 mL |
100 mg/1 mL |
|
|
500 mg/3 mL |
| LC-Alumina-A |
|
|
1 g/6 mL |
1 g/3 mL |
|
| LC-Alumina-B |
|
|
1 g/6 mL |
1 g/3 mL |
|
| LC-Alumina-N |
|
|
1 g/6 mL |
1 g/3 mL |
|
| LC-Florisil |
|
|
1 g/6 mL |
|
|
| Quantity each tube |
6 |
12 |
3 |
6 |
12 |
| Catalog Number |
57019 |
57009-U |
57075-U |
57074-U |
57073 |
|
Supelco
96-well SPE Method Development Plates contain an assortment of SPE phase
chemistries ideally suited for method development. The mix of phase chemistries
contained within this 96-well SPE plate allows researchers to screen for
analyte retention, recovery, and selectivity when achieving ones
sample prep objectives.
For ordering information, go to our
|
SPE MultiPaks consist of an assortment of SPE phase chemistries and tube dimensions
ideally suited for method development. The mix of phase chemistries available
in these MultiPaks allows you to screen for optimal retention and selectivity
required to achieve your sample prep objectives.
Available SPE MultiPaks
- Dispersive SPE (dSPE) MultiPaks
- Discovery Reversed-Phase SPE MultiPaks
- Discovery Normal-Phase SPE MultiPaks
- Discovery Ion-Exchange SPE MultiPaks
- Discovery DSC-MCAX (Mixed-Mode Cation Exchange) SPE MultiPak
- Discovery DPA-6S (Polyamide) SPE MultiPak
- Supelclean ENVI-Carb (Graphitized Carbon) SPE MultiPak
- Discovery Ag-Ion SPE MultiPak
- Supelclean Dual Layer SPE MultiPak (for multi-residue pesticide analysis
- Supelclean PSA SPE MultiPak
- Supelclean Sulfoxide SPE MultiPak
- Sample Prep Pipette Tip MultiPaks
View additional information on Discovery, Supelclean,
Sample Prep Pipette Tip, and Dispersive SPE MultiPaks. You can also contact Technical
Service at 814-359-3041 (800-359-3041 within the U.S.) or techservice@sial.com.
Refer to our library of Technical Resources
for more information.
For application assistance, please use the Application
Assistance Form to provide your requirements. A technical service representative
will contact you to assist you in developing/troubleshooting your sample prep
method.
|