Analytical / Chromatography
Air Monitoring
Analytical Reagents
Analytical Standards
Gas Chromatography
HPLC
Microbiology
Microscopy
Sample Preparation
SPE
Overview
Phase Selection
Specialty Products
Accessories
Method Development
Technical Resources
Custom Capabilities
Diffusive Sampling
SPME
VersaFlash System
Versapure Büchner Funnels
Spectroscopy
Syringes
Titration
TLC
Vials
Videos
Events
Literature
Analytix
The Reporter
Reporter Europe
 SPE Cartridges (Tubes) - Phase Selection Guide
Solid Phase Extraction
Standard SPE Phases
Specialty SPE Phases
Standard SPE Phases back to top
Your Sample
Matrix is:
Aqueous
(biological fluids, water,
aqueous extracts of tissues, etc.)
Organic
(organic extracts of tissues,
hexane, dichloromethane, etc.)
  ________|_________________    ________________|_______  
|_________________________|    |_______________________|  
Recommended
Retention
Mechanism:
Reversed-Phase Ion-Exchange Normal Phase
Analyte
Characteristics:
Moderately polar to
non-polar compounds
Weak
cations / anions
Strong
cations / anions
Polar to moderately
polar compounds
Application Pharma. Environ. Pharma. Environ. Pharma. Environ. Pharma. Environ.
Recommended
SPE Phase:
DSC-18
DSC-18Lt
DSC-8
DSC-Ph
DSC-CN
DPA-6S
ENVI™-18
ENVI-8
ENVI-Chrom P
LC-18
LC-8
LC-Ph
LC-CN
DSC-SAX
DSC-SCX
LC-SAX
LC-SCX
DSC-WCX
DSC-NH2
LC-WCX
LC-NH2
PSA
DSC-Si
DSC-CN
DSC-Diol
DSC-NH2
ENVI-Florisil®
LC-Alumina
LC-Florisil
LC-Si
LC-NH2
LC-Diol
LC-CN
PSA
The SPE Tube Configuraton Guide will help you choose the appropriate bed weight and cartridge dimensions.
NEW! Supelco now offers SPE sample MultiPaks to make the phase selection process easier.
Specialty SPE Phases back to top
Phase Description Application
SupelMIP™ SPE Tubes Molecular imprinted polymers for the highly selective extraction of trace analytes in difficult sample matrices
Supelclean™ Sulfoxide SPE Trace level extraction of PCBs and other aromatic compounds from difficult oil samples under normal-phase conditions
Supelclean Coconut Charcoal SPE Extraction of nitrosamines in drinking water (US EPA Method 521)
Supelclean ENVI-Carb Plus SPE Spherical carbon particles engineered for the extraction of highly polar compounds from aqueous samples such as drinking and ground water.
Discovery DSC-MCAX Mixed-mode cation exchange for superior selectivity/sample cleanup when extracting basic compounds (most pharmaceuticals) from biological fluids (e.g., plasma, urine, etc.)
Discovery Ag-Ion Silver Ion SPE for the fractionation of cis-trans isomers and other FAMEs
Polymer SAX Rezorian™ Cartridge Strong cation and anion exchanger on a styrene base particle. Offers much higher ion-exchange capacity Polymer SCX Reversible Tube than silica based ion-exchangers
Discovery DPA-6S Polyamide resin that adsorbs polar compounds containing multi –OH and –COOH groups. Useful for extracting polyphenolics and other natural compounds (e.g., flavenoids, chlorophyll, humic acid, etc.) from plant extracts
Supelclean ENVI-18
and –8 DSK SPE Disks
Provides fast flow rates for processing large volumes of water samples
(≥ 0.5 L). Used in EPA 500 series methods – Drinking Water
Supelclean ENVI-Carb Extreme affinity for polar compounds in aqueous samples and water miscible organic extracts.
Commonly used in pesticide analysis of food samples
Multi-layer Supelclean
SPE Products (ENVI-Carb™, SAX, PSA, NH2)
Developed to provide superior cleanup when conducting multi-residue pesticide analysis in food/agricultural matrices
Supelclean LC-4 (wide pore) Used for desalting proteins/peptides and other macromolecules
Dual Layer Florisil/Na2SO4 For total petroleum hydrocarbon index according to European Method EN9377-2
We also offer a variety of Specialty Products.