Glass SPE Tubes
&
PTFE & SS Frits
Common in environmental
analysis to reduce leachables
from PP hardware and PE frits |
VersaPure® Prepacked
Büchner Funnels
Used for large scale purification
(medicinal chemistry purification) |
Reversible
SPE Tubes
Reverse SPE tubes prior to elution
to minimize elution volume for
strongly retained compounds |