|
VersaPure™ Prepacked, Disposable Büchner Funnels |
 |
Solid Phase Extraction |
 |
 |
| |
|
Medicinal Chemistry is a branch within the
pharmaceutical industry that merges expertise in organic, analytical, computational
chemistry, and drug metabolism to conceptualize, design, and create/synthesize
novel compounds for potential therapeutic use.
VersaPure™ Prepacked Büchner Funnels are designed to augment
other larger scale purification techniques such as flash chromatography
and preparative HPLC by offering a convenient, rapid, reliable and simple
approach to larger scale sample prep/ purification. |
| |
| Reduce Your Risk, Prevent Problems, Save Time and Money |
 |
Prepared to eliminate exposure to inhalable
particulates Compressed bed eliminates channeling
Porous frit helps to spread sample across the adsorbent
bed Prepackaged design saves time in packing, cleaning,
and disposal Reallocate dedicated
lab (hood) space for packing Space savings for sorbent
inventory Quality - reliability - reproducibility
Personal safety, during packing and clean-up |
VersaPure Büchner Funnel Specifications
Semi-translucent polypropylene body Sealed
retaining ring compresses and seals bed in place Polyethylene
frits, 20 µm porosity 0.7 µm glass fiber
membrane below bottom frit
Dimensions available:
| |
Bed wt. |
I.D. (mm) |
Height (mm) |
| |
~12.5 g |
55 |
30 |
| |
~25 g |
70 |
40 |
| |
~50 g |
90 |
48 |
| |
~100 g |
110 |
66 |
Empty Disposable Büchner Funnels
packed with: 0.7 µm GF membrane 20
µm PE frit retaining ring |
To order from the list of available Büchner Funnels below, go to the
|
Discovery® DSC-Si
Merck-Si
Charcoal
Magnesium Sulfate
Celite®
Florisil® |
Alumina-A
Alumina-B
Alumina-N
Discovery DPA-6S
Empty disposable |