|
We carry the popular and high-quality Hamilton, SGE and VICI® Precision
Sampling analytical gas and liquid syringes and accessories. The selection of
brands gives you choices based on the product offering, application relevance,
instrument compatibility, and personal preference. We continually work with
our customers, suppliers, and instrument manufacturers to make sure our syringe
line stays up-to-date. As with all of our products, they are backed by Sigma-Aldrich’s
commitment to innovation, quality, and service.
The right choice of syringe is vital to ensure proper fit and operation within
the instrument, accurate and precise injections, and long functional lifetime.
Structurally, syringes must ensure a tight and secure fit in the injection
port and withstand repeated injection cycles without bending or breaking. Compositionally,
they must be able to withstand chemical and mechanical forces within a defined
range. Chemically, they must be inert and not introduce extraneous, interfering
compounds into the sample. Syringes also must deliver accurate and precise volumes
with no dead volume or carryover between injections.
The termination is the interface, or connection, between the syringe barrel
and the needle. The choice depends on the application. Each manufacturer has
its own specification nomenclature.
Fixed (cemented) – Economical, reproducible injections, for autosamplers,
one syringe-one method, do not heat above 50 °C
Removable needle – One syringe-many methods, simplicity of fi
xed needle, but needle can be replaced if damaged or clogged, do not heat above
115 °C
Knurled hub – Creates a leak-tight seal that can be retightened
to compensate for wear
Luer tip – Easy, fast needle replacement, syringe fi lter or
pump priming, Luer tip is ground glass suitable for mounting chromatographic
or PTFE needles, syringes can be autoclaved (without plunger or needle)
Luer-Lok – Security of a locked needle, syringe fi lter or pump
priming, PTFE, male Luer taper with nickel-plated brass locking hub for use
with KEL-F® or metal hub needles and universal connectors
Syringe-Valves – Syringe with On-Off valves, head space applications,
collect and store volatile samples, examples are SampleLock (Hamilton) or Pressure-Lok
(VICI).
Hamilton Termination Nomenclature:
N – Fixed needle, RN – Removable needle, LT – Luer tip, KH
– Knurled hub, TLL – PTFE Luer-Lok, WG – Plunger guides which
prevent bending the syringe, CH – Chaney adapter, SL – SampleLock
SGE Termination Nomenclature:
F – Fixed needle provides reproducible injects at an economical price,
R – Removable needle, LL – Luer-Lok, LT – Luer tip, B –
Plunger-in-Needle
Standard needles are most often point style 2 (sharp point) or 3 (blunt point).
For HPLC injections and sample pipetting, the blunt-tip style 3 is preferred.
The blunt tip has chamfered and polished edges that eliminate damage to the
valve’s rotor seal and stator face. This style can also be used for pipetting
liquids.
For manual GC injections, a beveled tip, like style 2 is preferred for optimum
septum penetration and to prevent septum coring. Also, Hamilton point style
AS and SGE cone-tip needles are specially designed for this application. When
using an autosampler, consult the instrument manual for the preferred point
style.
In syringes with point style 5, conical with side port, liquid samples can
be filled and dispensed through the hole in the side. Septum damage is minimized
by the solid domed tip.
| Point Style 2 |
 |
Sharp, beveled, curved,
non-coring needle point recommended for septum penetration.
Available gauges: 33-10. |
| Point Style 3 |
 |
Blunt needle point for use
with HPLC injection valves and for sample pipetting.
Available gauges: 33-10 |
| Point Style 4 |
 |
Standard 12° beveled
needle point is recommended for life science applications.
Available gauges: 33-10
Special point styles such as 30°, 45°, or any other angle are available
upon request. |
| Point Style 5 |
 |
Conical needle with side
port for penetration of septa, thin-gauged vinyls and plastics
without coring. Available gauges: 26-10 |
| Point Style AS |
 |
Spherical conical style
needle point used on autosampler syringes. The non-coring needle point is
recommended for septum penetration.
Available gauges: 26-22 |
Gauge is the thickness of the needle. The gauge required depends on the injector.
For example, in HPLC applications, Waters instruments use a 25 gauge needle,
while Rheodyne, Valco, Altex and SSI valves use 22 gauge.
23 or 23s Single Gauge (0.64
mm)
- Merlin Microseal™ septa
- Packed column injector ports
- Split/splitless injector ports
|
 |
26 or 26s Single Gauge (0.47
mm)
- Packed column injector ports
- Split/splitless injector ports
|
 |
Dual-Gauge 23-26 or 23s-26s
(0.64 - 0.47 mm) Tapered
- Durability of a 23-gauge
- Versatility of a 26-gauge for split/splitless and on-column injection