|
Supelco carries the most popular VICI Precision Sampling syringes from Valco.
These syringes have wide applicability for both gases and liquids. The pressurization
option is valuable when dealing with transport and storage of volatile samples.
We can also supply any Precision Sampling syringe not shown on these pages.
Gas / Liquid Syringes
Series A
Series A-2
Series C
Series D
Magnum Series
Liquid Syringes
Series C-160
Series C-120
Series CG-130
Series D-140
Pressurized storage with built-in rotary valve
Removable needle, bevel, open end
Syringe Volumes: 100 µL, 250 µL, 500 µL, 1 mL, 2 mL, 5 mL,
10 mL
- Integral sample storage valve - actuated by turning the nosepiece
- Positive rear flange plunger stop - prevents plunger from blowing out of
barrel at elevated pressure
- Carrier back-flushing is eliminated - resulting in sharper peaks and better
separations
- Modular construction - all components may be quickly disassembled for easy
cleaning or replacement
- Leak tight to 250 psi for both liquids and gases
- Has all the standard Pressure-Lok features such as a PTFE plunger tip,
PTFE-sealed needle, and ultra-smooth bore
Pressurized storage with built-in push-button valve
Removable needle, bevel, open end, also available with Luer needle
Syringe Volumes: 25 µL, 50 µL, 100 µL, 250 µL, 500
µL, 1 mL, 2 mL, 5 mL, 10 mL
- Push-button valve - allows sample storage to 250 psi in syringes as small
as 25 µL and instantaneous injection
- Smaller volumes - great for small volatile samples
- Useful with small liquid samples containing low-boiling components which
would be lost through evaporation using ordinary syringes
- Positive rear stop (in 250 µL and larger sizes) prevents plunger
blowout at elevated pressures, protecting against sample loss or operator
injury
- Has all the standard Pressure-Lok features such as a PTFE plunger tip,
PTFE-sealed needle, and ultra-smooth bore
The perfect syringe for everyday use
Removable needle, bevel, open end, also available with Luer needle or with
Chaney adapter
Syringe Volumes: 100 µL, 250 µL, 500 µL, 1 mL, 2 mL, 5 mL,
10 mL
- Positive rear plunger stop
- Low-dead volume removable needle
- Has all the standard Pressure-Lok features such as a PTFE plunger tip,
PTFE-sealed needle, and ultra-smooth bore, but without a valve for pressurized
sample storage
- Sample storage capability can be added with a Mininert syringe valve
- Optional Chaney adapter offers much lower dead volume than Luer-style connectors
Economically priced gas syringe
Removable needle, bevel, open end
Syringe Volumes: 1 mL, 2 mL
- Leak-tight to 250 psi
- Pressure-Lok plunger tip
- No dead volume barrel design
- Special low dead volume needle connector for accurate delivery of gas samples
Pressurized storage of large volumes for trace sample isolation
Removable needle, bevel, open end
Syringe Volumes: 20 mL, 50 mL, 100 mL, 250 mL, 500 mL
- Ideal for collection, trace analysis and sample concentration of environmental
contaminants
- A low-dead volume needle is used in this syringe
- A rotary valve allows on-site sampling for later analysis
- Positive displacement capability of the durable PTFE plunger seal permits
use in a variety of subsequent procedures involving extraction or concentration
of trace components
- The standard termination is a female threaded fitting (1/4-28), which
accepts a variety of Cheminert connectors or adapters. Specially formed seals
permit leak-tight operation to 30 psi (approximately 2 atmospheres).
VICI basic PTFE plunger-tipped microsyringe
Fixed and removable needles, bevel, open end
Syringe Volumes: 5 µL, 10 µL, 25 µL, 50 µL, 100 µL
- Day-in, day-out dependability at an economical price
- Leak-tight to 250 psi - liquids and gases
- PTFE plunger tip is self-lubricating and durable
- PTFE needle seat at rear of needle prevents possible dissolution of needle
cement or contamination of sample
Barrel extension for attaching a vacuum bulb or line
Fixed needle, bevel, open end
Syringe Volumes: 5 µL, 10 µL, 25 µL, 50 µL, 100 µL
- Leak-tight to 250 psi - liquids and gases
- PTFE plunger tip
- Barrel extension beyond the rear flange – permits attachment of a
vacuum bulb or line for easier filling or cleaning
- Standard Pressure-Lok features
Reinforced plunger to prevent bending
Fixed and removable needles, bevel, open end
Syringe Volumes: 5 µL, 10 µL
- Leak-tight to 250 psi - liquids and gases
- Heavy, reinforced plunger within an enlarged plunger guide - eliminates
plunger bending
- Lower plunger section confined within the guide section
- Sturdy upper section absorbs the force of a rapid down-stroke
For small (<10 µL) injections
Removable needle: size and style vary with syringe size
Syringe Volumes: 1 µL, 2 µL, 5 µL, 10 µL
- Designed for small volume injection into gas chromatographs
- Total sample displacement - the entire sample is contained in the needle
- Positive rear plunger stop - eliminates accidental plunger removal
- Superior alignment - permits repeated reassembly without difficulty or
damage
- PTFE seals, needle, plunger, and barrel are all replaceable
|