Analytical / Chromatography
Air Monitoring
Analytical Reagents
Analytical Standards
Gas Chromatography
HPLC
Microbiology
Microscopy
Sample Preparation
Spectroscopy
Syringes
Titration
Complexometric Titration
HYDRANAL
ISE Equipment
Volumetric solutions
pH Electrodes
TLC
Vials
Videos
Events
Literature
Analytix
The Reporter
Reporter Europe
 Complexometric Titration
Titration
IDRANAL® - Reagents for complexometric titrations
Sigma-Aldrich offers a few of the aminopolycarboxylic acids marked by the registered trade mark
IDRANAL®. For use in complexometric titration.
  IDRANAL® I IDRANAL® II IDRANAL® III IDRANAL® IV IDRANAL® V IDRANAL® VI IDRANAL® VII
Abr. NTA EDTA EDTA-Na2 DCTA DTPA EGTA HEDTA-Na3
Art. 34539 31064 34549 34588 34589 34596 34461
View complete product list for complexometric titration
Indicator buffer tablet
Visual indicators are used in complexometric titrations. Eriochrome Black T is the indicator
most widely used but whose aqueous solution is not stable. Therefore IDRANAL Indicator buffer
tablets are used. These tablets contain Eriochrome Black T as the metal indicator and a buffer substance.
They are suitable to titrating Ca, Mg, Zn, Pb and for the determination of water hardness.
Application A 100 ml water sample is boiled after adding 2 drops of concentrated hydrochloride acid -
e.g. Art. 30721- cooled to 40 °C, and mixed with an IDRANAL Indicator buffer tablet and 1 ml of 25% ammonia solution- e.g. Art. 30501. Titration is carried out with IDRANAL solution.
A high carbonate content in the water can slow down the colour change. It can be overcome by boiling the weakly acidified water.To avoid this slow down colour change totally the water containing little or no magnesium should be titrated with IDRANAL II-Zinc, or with IDRANAL A, B or C which already contains IDRANAL II-Zinc.
Titration Home