Analytical / Chromatography
Air Monitoring
Analytical Reagents
Analytical Standards
Gas Chromatography
HPLC
Microbiology
Microscopy
Sample Preparation
Spectroscopy
Syringes
Titration
Complexometric Titration
HYDRANAL
Additional Reagents
Aldehydes and Ketones
Applications
Coulometric Reagents
HYDRANAL® Manual
HYDRANAL®-E types
Instruments
Karl Fischer Seminars
Literature
Moisture Test Kit
Multimedia Guide CD
NEW Literature request
One-Component Rgt.
PhEur requirements
Technical Helpline
Theory
Two-Component Rgt.
Water Standards
ISE Equipment
Volumetric solutions
pH Electrodes
TLC
Vials
Videos
Events
Literature
Analytix
The Reporter
Reporter Europe
 Coulometric Karl Fischer titration
HYDRANAL
Composition
Coulometry normally necessitates two reagent solutions, an anolyte and a catholyte. The anolyte is added to the anodic compartment and the catholyte to the cathodic compartment.
HYDRANAL®-Coulomat A reagents are used as anolyte. Every anolyte contains iodide and a sulphur dioxide/imidazole buffer in a suitable solvent. HYDRANAL®-Coulomat C reagents are used as catholytes.
Toxicity
HYDRANAL®-Coulomat G-Types have been developed as anolyte as well as catholyte.
These reagents are:
  • Pyridine free.
  • Chloroform free.
  • Carbon tetrachloride free.
  • 2-methoxyethanol free.
HYDRANAL®-Coulomat G reagents contain methanol as the most hazardous component. The non-toxic HYDRANAL®-Coulomat E is mainly based on ethanol and must not wear the skull and crossbones on the label.
Stability
All reagents have a shelf life of at least five years, as long as the bottles remain unopened.
Advantages of the coulometric titration
  • Easy to use.
  • High accuracy.
  • Suitable for the determination of 10 mg - 10 mg of water.
  • Reagents available with and without halogenated hydrocarbons.
View Product List and Ordering Information
Literature
English
Karl Fischer titration The Coulometric Cell
HYDRANAL®-Coulomat Oil A new reagent for the coulometric water determination in oils
Analytix 3/2008 Coulometric water determination according to Karl Fischer
Deutsch
Karl-Fischer-Titration Die Coulometriezelle
HYDRANAL®-Coulomat Oil Ein neues Reagenz für die coulometrische Wasserbestimmung
in Ölen
Back to top
Back to HYDRANAL®-home