Analytical / Chromatography
Air Monitoring
Analytical Reagents
Analytical Standards
Gas Chromatography
HPLC
Microbiology
Microscopy
Sample Preparation
Spectroscopy
Syringes
Titration
Complexometric Titration
HYDRANAL
Additional Reagents
Aldehydes and Ketones
Applications
Coulometric Reagents
HYDRANAL® Manual
HYDRANAL®-E types
Instruments
Karl Fischer Seminars
Literature
Moisture Test Kit
Multimedia Guide CD
NEW Literature request
One-Component Rgt.
PhEur requirements
Technical Helpline
Theory
Two-Component Rgt.
Water Standards
ISE Equipment
Volumetric solutions
pH Electrodes
TLC
Vials
Videos
Events
Literature
Analytix
The Reporter
Reporter Europe
 HYDRANAL®-E types - Non-toxic reagents for Karl Fischer Titration
HYDRANAL Karl Fischer Titration Reagents
At the center of the continuous developments is the replacement of toxic methanol which is widely used as solvent in the titration vessel or as solvent for Karl Fischer reagents by the non-toxic ethanol. HYDRANAL®-E types represent the first non-toxic Karl Fischer reagent line for the volumetric and the coulometric titration.
Advantages of non-toxic HYDRANAL®-E types
  • Compared with methanol based Karl Fischer reagents handling is less dangerous for employees.
  • Reagents meet requirements of ISO EN DIN 14001.
  • Improved solubility of long-chained hydrocarbons.
  • Ketones like acetone can be titrated in HYDRANAL®-CompoSolver E without side-reaction.
  • Compared with the traditional two-component reagents the non-toxic reagents have the same titration speed, identical shelf life and titre parameters.
  • The non-toxic HYDRANAL®-Coulomat E can be used as one-component-reagent for coulometric cells with diaphragm.
  • Comparative applications are available on request
View Product List and Ordering Information
Literature
English
HYDRANAL®-E types Reagents for methanol-free Karl Fischer titration
AnalytiX 1/2008 HYDRANAL®-E - type reagents
German
HYDRANAL®-E Typen Reagenzien für die methanolfreie Karl-Fischer-Titration
Back to top
Back to HYDRANAL®-home