Analytical / Chromatography
Air Monitoring
Analytical Reagents
Analytical Standards
Gas Chromatography
HPLC
Microbiology
Microscopy
Sample Preparation
Spectroscopy
Syringes
Titration
Complexometric Titration
HYDRANAL
Additional Reagents
Aldehydes and Ketones
Applications
Coulometric Reagents
HYDRANAL® Manual
HYDRANAL®-E types
Instruments
Karl Fischer Seminars
Literature
Moisture Test Kit
Multimedia Guide CD
NEW Literature request
One-Component Rgt.
PhEur requirements
Technical Helpline
Theory
Two-Component Rgt.
Water Standards
ISE Equipment
Volumetric solutions
pH Electrodes
TLC
Vials
Videos
Events
Literature
Analytix
The Reporter
Reporter Europe
 HYDRANAL®-Moisture Test Kit
HYDRANAL Karl Fischer Titration Reagents
This ready to use Test Kit is intended to determine the amount of water in various samples, such as in organic solvents. It includes all the equipment and reagents needed in one packing. The individual reagents can be ordered separately.
The HYDRANAL®- Moisture Test Kit allows to determine the amount of water in colorless liquid samples. This ready to use Test Kit can be inserted on site, for example, directly at a solvent or fuel tank, and it is likewise suitable for use in laboratories that do not have a Karl Fischer titrator.
The HYDRANAL®- Moisture Test Kit includes all equipment and reagents needed in one packing. These are:
  • 34732 HYDRANAL®-Titrant E
  • 34730 HYDRANAL®-Solvent E
  • 34813 HYDRANAL®-Standard 5.00
  • A syringe to measure out the sample.
  • A bottle acting as titration vessel.
How to use the HYDRANAL®- Moisture Test Kit?
  • Determine the titre of HYDRANAL®-Titrant Component with HYDRANAL®-Standard 5.00.
  • With a syringe add HYDRANAL®-Titrant Component to HYDRANAL®-Solvent E until the color changes from yellowish to slightly brownish. Now the system is equilibrated to analyse the sample.
  • With the syringe supplied add a defined amount of sample.
  • Add HYDRANAL®-Titrant Component with the scaled syringe until the colour changes to brownish again. The content of water can be calculated from the volume of HYDRANAL®-Titrant Component, the titre and the sample size.
  • Several samples can be titrated one after another in the same vessel.
View Product List and Ordering Information
Literature
Analytix II/05
Back to top
Back to HYDRANAL®-home