Analytical / Chromatography
Air Monitoring
Analytical Reagents
Analytical Standards
Gas Chromatography
HPLC
Microbiology
Microscopy
Sample Preparation
Spectroscopy
Syringes
Titration
Complexometric Titration
HYDRANAL
Additional Reagents
Aldehydes and Ketones
Applications
Coulometric Reagents
HYDRANAL® Manual
HYDRANAL®-E types
Instruments
Karl Fischer Seminars
Literature
Moisture Test Kit
Multimedia Guide CD
NEW Literature request
One-Component Rgt.
PhEur requirements
Technical Helpline
Theory
Two-Component Rgt.
Water Standards
ISE Equipment
Volumetric solutions
pH Electrodes
TLC
Vials
Videos
Events
Literature
Analytix
The Reporter
Reporter Europe
 One-component reagents for volumetric Karl Fischer titration
HYDRANAL Karl Fischer Titration Reagents
Composition
HYDRANAL®-Composite is the world's most frequently used pyridine-free Karl Fischer reagent for one-component titration. It contains all the reactants, i.e. iodine, sulfur dioxide and imidazole, dissolved in diethyleneglycol monoethyl ether (DEGEE).
The R & D laboratory at Riedel-de Haën has recently developed an improved formulation for HYDRANAL®-Composite by using a mixture of imidazole and 2-methylimidazole instead of imidazole only. Significant improvements in the properties of HYDRANAL®-Composite have been obtained as a result of this change.
Stability
HYDRANAL®-Composite is stabilized with DEGEE. The loss of titre is approx. 5% a year. Other Karl Fischer reagents contain 2-methoxyethanol (2-ME).
Toxicity
DEGEE is non-harzardous unlike 2-ME which is fetotoxic and teratogenic. All HYDRANAL®-Composites are pyridine free and free of 2-methoxyethanol. Toxicological studies are done.
Advantages of the one-component titration
  • Unlimited water capacity.
  • Three different titres available.
  • Also convenient for the analysis of ketones.
  • Widely used: many different applications for the determination of i.e. 1 - 100 mg water.
  • Suitable for customary volumetric Karl Fischer titrators.
View Product List and Ordering Information
Literature
English
HYDRANAL®-Composite Improved one-component reagent for volumetric Karl Fischer Titration
HYDRANAL®-KetoSolver For volumetric moisture determination
HYDRANAL®-Medium K Excellent performance without toxic components
HYDRANAL®-Methanol Rapid For fast Karl Fischer titrations
HYDRANAL®-Solver (Crude) Oil Reliable moisture determination in oil and oily substances
Deutsch
HYDRANAL®-Composite Verbessertes Einkomponentenreagent für die volumetrische Karl-Fischer-Titration
HYDRANAL®-KetoSolver Für die volumetrische Wasserbestimmung
HYDRANAL®-Medium K Exzellente Leistung ohne toxische Komponenten
HYDRANAL®-Methanol Rapid Für reaktionsschnelle Karl Fischer-Titration
HYDRANAL®-Solver (Crude) Oil Bewährte Wasserbestimmung in Ölen und Rohölen
Back to top
Back to HYDRANAL®-home