Analytical / Chromatography
Air Monitoring
Analytical Reagents
Analytical Standards
Gas Chromatography
HPLC
Microbiology
Microscopy
Sample Preparation
Spectroscopy
Syringes
Titration
Complexometric Titration
HYDRANAL
Additional Reagents
Aldehydes and Ketones
Applications
Coulometric Reagents
HYDRANAL® Manual
HYDRANAL®-E types
Instruments
Karl Fischer Seminars
Literature
Moisture Test Kit
Multimedia Guide CD
NEW Literature request
One-Component Rgt.
PhEur requirements
Technical Helpline
Theory
Two-Component Rgt.
Water Standards
ISE Equipment
Volumetric solutions
pH Electrodes
TLC
Vials
Videos
Events
Literature
Analytix
The Reporter
Reporter Europe
 Theory of Karl Fischer Titration
HYDRANAL Karl Fischer Titration Reagents
Introduction
Sigma-Aldrich is a pioneer in the supply of safe, reliable and easy-to-use pyridine-free Karl Fischer reagents.
Water can affect product quality, texture, shelf life, chemical stability and reactivity. The Karl Fischer titration is a universally acknowledged method for measuring water content in all types of substances including chemicals, oils, pharmaceuticals and food. In 1979, Riedel-de Haën researcher Dr. Eugen Scholz improved the Karl Fischer titration by replacing noxious pyridine with imidazole.
This innovation became the foundation of HYDRANAL®, the world’s leading pyridine-free reagents for Karl Fischer titration. Since Dr. Scholz’s pioneering research and with a spirit of ongoing product improvement, Riedel-de Haën has launched a wide range of Karl Fischer reagents for both volumetric and coulometric titrations for nearly all types of samples.
Advantages of HYDRANAL® reagents:
  • High-speed titrations
  • Stable end points
  • Accurate results
  • No unpleasant odor
  • Lower toxicity for increased safety and decreased environmental impact
  • Long shelf life
  • Wide applicability
Table 1: HYDRANAL® Product Line at a Glance
HYDRANAL®-Composite The most frequently used pyridine-free Karl Fischer reagents for one-component volumetric titration
HYDRANAL®-Solvent / Titrant Volumetric titration with two-component reagents for higher accuracy and stability
HYDRANAL®-Coulomat Coulometric titration for samples with low water content
HYDRANAL®-K types Specially designed reagents for determining water in aldehydes and ketones
HYDRANAL®-E types Reduced toxicity over ethanol-containing reagents for both volumetric and coulometric titrations
HYDRANAL®-Water Standards Standards with certified water content for titer determination
In the following chapters, you will find the basic principles and possibilities of Karl Fischer titration.
Additional technical information can be obtained by requesting our HYDRANAL®-Manual,
Multimedia Guide CD, Literature or Applications. For personal assistance, please feel free to contact
our Karl Fischer helplines.
Chapter 1 Basic principles of Karl Fischer titration
Chapter 2 Basic forms of Karl Fischer titration
Chapter 3 Variations of Karl Fischer titration
Literature
[1] Eugen Scholz „Karl-Fischer-Titration“
  Springer-Verlag, Berlin, Heidelberg, New York, Tokyo (1984), 140 pages
  ISBN 3-540-12846-8 German Edition
  ISBN 3-540-13734-3 English Edition
Back to Top
Back to HYDRANAL® home