Sigma-Aldrich
Fluka & Supelco - Analytical Logo
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
HYDRANAL® Karl Fischer Titration Reagents
Coulometric Titration
Advantages of coulometric titration:
  • Easy to use
  • Detects low concentrations of water
  • High accuracy
Product range HYDRANAL® coulometric reagents
Composition
Coulometric Karl Fischer titrations usually require two reagent solutions: an anolyte (solution in the anodic compartment) and a catholyte (solution in the cathodic compartment). HYDRANAL®-Coulomat A type reagents are used as the anolyte. They contain iodide and a sulfur dioxide/imidazole buffer in a suitable solvent. HYDRANAL®-Coulomat CG reagents are used as catholytes. The non-toxic HYDRANAL®-Coulomat E is used as both anolyte and catholyte; it is based on ethanol and does not require hazardous labeling.
Toxicity
Except for HYDRANAL®-Coulomat AK containing 2-methoxyethanol, all HYDRANAL®-Coulomat reagents are free of toxic and noxious compounds, such as pyridine, carbon tetrachloride and 2-methoxyethanol. In most reagents, methanol is the most hazardous component, and it can be eliminated by using HYDRANAL®-Coulomat E where ethanol replaces methanol.
Stability
When unopened, most HYDRANAL®-Coulomat reagents have a shelf life of at least five years.
Using HYDRANAL®-Coulomat Reagents
The following procedure is generally used for the coulometric determination of water (Figure 2.3):
  1. Fill the anodic compartment with HYDRANAL®-Coulomat A/AG/AD/AG-H/AG-Oven/AK/E
  2. Fill the cathodic compartment with HYDRANAL®-Coulomat CG/CG-K/E
  3. Titrate the cell to dryness
  4. Add sample
  5. Titrate the water content
Coulometric titration with HYDRANAL® -Coulomat reagents
Figure 2.3: Coulometric titration with HYDRANAL® -Coulomat reagents.
Coulometry without Diaphragm
Coulometric titration cells are also available without diaphragm. They comprise only one compartment and require only one reagent. HYDRANAL®-Coulomat E, AD, AG, AG-H, AG Oven and AK can be used with diaphragm-less cells. To use these reagents, follow the general procedure for coulometric determination of water without the use of a catholyte solution.
Back to Introduction page
Back to HYDRANAL® Home