Sigma-Aldrich
Fluka & Supelco - Analytical Logo
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Analytical / Chromatography > Titration > HYDRANAL® > Learning Center > Theory of Karl Fischer Titration > Volumetric Titration with two-component reagents
Titration
Volumetric Titration with two-component reagents
Advantages of two-component titration:
  • Higher titration speed
  • Higher accuracy for small amounts of water
  • Higher buffer capacity
  • Exact and stable concentration
Product range HYDRANAL® volumetric two-component reagents
Composition
With two-component reagents, the reactants are separated in two solutions: the solvent and the titrating agent. HYDRANAL®-Solvent is a solution of sulfur dioxide and imidazole in methanol. HYDRANAL®-Titrant contains a solution of iodine with a precisely defined concentration.
Stability
The titration agents have a shelf life of three years, the solvents five years.
Toxicity
Non-toxic HYDRANAL®-E type reagents based on ethanol instead of methanol allow safer Karl Fischer titrations.
Using HYDRANAL®-Solvent / Titrant
We recommend the following procedure for the two-component titration technique (Figure 2.2):
  1. Fill the burette with HYDRANAL®-Titrant (E)
  2. Add HYDRANAL®-Solvent (E) to the titration vessel
  3. Titrate to dryness with HYDRANAL®-Titrant (E)
  4. Add sample
  5. Titrate the water content with HYDRANAL®-Titrant (E)
Volumetric two-component titration with HYDRANAL® -Solvent (E) / Titrant (E) Figure 2.2: Volumetric two-component titration with HYDRANAL® -Solvent (E) / Titrant (E).
Back to Introduction page
Back to HYDRANAL® Home