Analytical / Chromatography
Air Monitoring
Analytical Reagents
Analytical Standards
Gas Chromatography
HPLC
Microbiology
Microscopy
Sample Preparation
Spectroscopy
Syringes
Titration
Complexometric Titration
HYDRANAL
Additional Reagents
Aldehydes and Ketones
Applications
Coulometric Reagents
HYDRANAL® Manual
HYDRANAL®-E types
Instruments
Karl Fischer Seminars
Literature
Moisture Test Kit
Multimedia Guide CD
NEW Literature request
One-Component Rgt.
PhEur requirements
Technical Helpline
Theory
Two-Component Rgt.
Water Standards
ISE Equipment
Volumetric solutions
pH Electrodes
TLC
Vials
Videos
Events
Literature
Analytix
The Reporter
Reporter Europe
 Two-component reagents for volumetric Karl Fischer titration
HYDRANAL Karl Fischer Titration Reagents
Composition
With two-component reagents, the reactants are in two separate solutions. HYDRANAL®-Solvent is a solution of sulfur dioxide and imidazole in methanol. HYDRANAL®-Titrant is a solution of iodine, dissolved in a suitable solvent, with an exact titre and a high stability of the titre.
Stability
The titrants have a shelf life of three years, the solvents of five years.
Toxicity
Non-toxic E-Type reagents based on ethanol make safer Karl Fischer titration possible.
Advantages of the two-component titration
  • Higher titration speed compared with volumetric one-component titration.
  • Greater accuracy for small amounts of water.
  • Higher buffer capacity.
  • Exact and stable titre.
View Product List and Ordering Information
Literature
English
HYDRANAL®-E Types The new dimension in non-toxic Karl Fischer titration
Deutsch
HYDRANAL®-E Types Reagenzien für die nicht toxische Karl Fischer-Titration
Back to top
Back to HYDRANAL®-home