Analytical / Chromatography
Air Monitoring
Analytical Reagents
Analytical Standards
Gas Chromatography
HPLC
Microbiology
Microscopy
Sample Preparation
Spectroscopy
Syringes
Titration
TLC
Vials
Videos
Events
Literature
Analytix
The Reporter
Reporter Europe
Product Lines
Vials
Click to see a product selection Guide
Vials > Sampling > Crimp Seal > vials taking 8 mm seal | vials taking 11 mm seal
Vials taking 8 mm seal
Supelco offers five different sizes of vials with a 8 mm crimp finish. They are available in both clear and amber glass. All of these vials can use the same 8 mm crimp cap. The sizes of vial that we offer are:
• 6 x 32 mm
• 7 x 30 mm
• 7 x 32 mm
• 7 x 40 mm
• 8 x 30 mm
Vials only
Crimp seals with septa included
Shell vials with plugs
Shell vials (no plugs)
Plugs for shell vials
Inserts and springs
Vials only back to top
Product Name Product # 
6 mm x 32 mm  
0.2 mL clear glass (tapered bottom) pack of 200 27283
0.2 mL clear glass (tapered bottom) pack of 1000 27284
0.3 mL clear glass (round bottom) pack of 200 24725
0.3 mL clear glass (round bottom) pack of 1000 24726
7 mm x 30 mm  
0.6 mL amber glass (conical) pack of 200 27312
0.6 mL amber glass (conical) pack of 1000 27313
Chromacol Vial PTFE (non-disposable sleeve) O.D. × height 12 mm × 32 mm for use with 7 x 30 mm vials pack of 40 27316
7 mm x 32 mm  
0.8 mL amber glass (round bottom) pkg of 100 27314
0.8 mL amber glass (round bottom) pack of 1000 27315
7 mm x 40 mm  
0.8 mL clear glass pack of 200 24738-U
0.8 mL clear glass pack of 2000 24739
0.8 mL amber glass pack of 200 24740-U
0.8 mL amber glass pack of 2000 24741
0.7 mLclear glass (conical) pack of 200 24742
0.7 mL clear glass (conical) pack of 1000 24743
0.7 mL amber glass (conical) pack of 200 24744
0.7 mL amber glass (conical) pack of 1000 24745
8 mm x 30 mm  
0.5 mL clear glass pack of 200 33275
0.5 mL clear glass pack of 1000 33276
0.5 mL amber glass pack of 2000 24724
Crimp seals (8 mm) with septa included back to top
Product Name Product # 
red PTFE/silicone/red PTFE pack of 200 33265-U
red PTFE/silicone/red PTFE pack of 1000 33266-U
PTFE/silicone pack of 200 33273-U
PTFE/silicone pack of 1000 33274-U
PTFE/silicone pack of 100 27359
red PTFE/silicone pack of 1000 27372
PTFE/red rubber pack of 200 33135-U
PTFE/red rubber pack of 1000 33136
Shell vials with plugs back to top
Product Name Product # 
8 mm x 35 mm  
0.75 mL clear glass pack of 200 24727
0.75 mL clear glass pack of 1000 24728
0.75 mL amber glass pack of 200 24729
0.75 mL amber glass pack of 1000 24730
8 mm x 40 mm  
1 mL clear glass vial pack of 200 33321-U
1 mL clear glass vial pack of 1000 33326
1 mL amber glass pack of 250 27290
1 mL amber glass vial pack of 1000 27291
0.7 mL polypropylene vial pack of 100 27537
Shell vials (no plugs) back to top
Product Name Product # 
8 mm x 40 mm  
1 mL clear glass pack of 200 33320-U
1 mL clear glass pack of 1000 33325-U
Plugs for shell vials (8 mm) back to top
Product Name Product # 
polypropylene pack of 100 27289
natural polypropylene pack of 200 33322-U
natural polypropylene pack of 1000 33327
red polypropylene pack of 200 33323
red polypropylene pack of 1000 33328-U
blue polypropylene pack of 200 33324
blue polypropylene pack of 1000 33329
Inserts and springs back to top
Product Name Product # 
Insert for Shell Vial size 0.20 mL clear glass conical (requires bottom spring, 24735) pack of 200 24733
Insert for Shell Vial size 0.20 mL clear glass conical (requires bottom spring, 24735) pack of 1000 24734
Springs for Inserts pack of 200 24735
Insert for Shell Vial size 0.10 mL clear glass conical (with bottom spring) pack of 100 24737
Insert for 2 mL Standard Opening Vial size 0.15 mL polypropylene conical (with bottom spring) O.D. × height 5 mm × 30 mm pack of 100 24708