Vial
racks  |
back
to top |
| Economical and convenient storage for samples
in your screw cap and crimp seal vials. Can be stacked when empty or full.
Autoclavable, 120 °C. |
|
| Product
Name |
Product # |
|
for use with 6-8mm vial (96 vials per rack) pack of 5 |
23204-U |
|
for use with 11-12mm vial (50 vials per rack) pack of 5 |
23207 |
|
for use with 15-16mm vial (48 vials per rack) pack of 5 |
23205-U |
|
for use with 17mm vial (90 vials per rack) pack of 5 |
23202 |
|
for use with 21-23mm vial (36 vials per rack) pack of 5 |
23201 |
|
for use with 29mm vial (50 vials per rack, for 40mL EPA vial) pack of
5 |
23206 |
Storage
bottles, plastic coated  |
back to top |
Ideal for storing fluids, dry chemicals, and other
lab materials Slip resistant plastisol coating reduces potential
breakage and protects lab personnel Wide opening , special lip,
and sloping shoulders make pouring and cleaning easier Low alkali
borosilicate gl ass content does not harm fluids Autoclavable
Each bottle comes with a white polypropylene screw cap with a wedge-shaped
inner sealing ring and polypropylene pour ring.
|
|
| Product
Name |
Product # |
| 100 mL clear glass O.D.
× height 59 mm × 10 mm pack of 12 |
23171 |
| 500 mL clear glass O.D.
× height 91 mm × 187 mm pack of 12 |
23172 |
| 1000 mL clear
glass O.D. × height 104 mm × 224 mm pack
of 12 |
23173 |
| Cap for Mobile Phase Reservoir
System GL 45 solid (no holes) pack of 12 |
23170-U |
Storage
bin  |
back to top |
| A convenient way to organize your vials.
Available filled with supplies or unfilled. |
|
| Product
Name |
Product # |
| Nine-Drawer Work Station filled with 2000
2 mL standard opening screw top clear glass vials, red PTFE/silicone
septa, and black phenolic caps (recommended for Varian and Spectra Physics
autosamplers) |
27264-U |
| Nine-Drawer Work Station
filled with 3000 2 mL large opening crimp top clear glass vials, PTFE/red
rubber 11 mm septa, and seals |
27231-U |
| Nine-Drawer Work Station
unfilled (cabinet only) |
27263 |
Glass
magnets  |
back to top |
| Our Glass Magnet Sheet prevents your glass apparatus from
being knocked over accidentally. This 2' x 2' sheet (61 cm x 61 cm), with
its tacky surface, effectively secures the vessels in place. It can be cut
with a razor blade or utility knife to fit strategically on counter tops
or in drawers containing fragile glassware. Add it to carts when transporting
glass to another lab or into the field. The Glass Magnet Vial Holder keeps
individual vials, ampuls, and other vessels in place. |
|
| Product
Name |
Product # |
| Glass Magnet mat width
× length × thickness 24 in. × 24 in. × 3/16 in. pack
of 1 |
57269 |
| Glass Magnet diam. ×
thickness 4 in. × 3/16 in. pack of 2 |
57270 |
| GLASS MARKING PEN |
64800-U |
| 2.5
mL vials, crimp caps and septa |
back to top |
| Product
Name |
Product # |
| 2.5 mL vial (requires
13 mm seal) O.D. × height 11.7 mm × 34.5 mm pack
of 288 |
33121 |
| Crimp Seal (without septum)
diam. 13 mm removable center pack of 100 |
27222-U |
| Crimp Seals with PTFE/Rubber
Septa diam. 13 mm removable center pack of 100 |
27223 |
| Crimp Seals with PTFE/Rubber
Septa diam. 13 mm removable center pack of 1000 |
27451 |
| Stoppers septum diam. 13 mm butyl
rubber pack of 100 |
27224 |
| Stoppers septum diam. 13 mm red
rubber pack of 100 |
27226 |
| Crimp Seals with PTFE/Silicone
Septa diam. 13 mm pack of 100 |
27452 |
| Special
use vials |
back to top |
| Product
Name |
Product # |
| Shimadzu 5mL Screw Top
Vial, 12 X 96mm, 10/425 Thread size 5 mL clear N-51A borosilicate
glass body thread 10-425 |
27319-U |
| Shimadzu 5mL Screw Top
Vial, 12 X 96mm, 10/425 Thread size 5 mL clear N-51A borosilicate
glass body thread 10-425 |
27320-U |
| Shimadzu 5mL Vial, 12 X
96mm, PP Hole Cap with PTFE Septa Shimadzu 5mL vial and closure pre-assembled closure
type, screw cap polypropylene caps (10-425) (PTFE Septa) |
27321 |
| Fisons 2.5mL Vial, 12 X
40mm size 2.5 mL O.D. × height 12 mm × 40 mm vial
type, crimp top clear KG-33 borosilicate glass body |
27323 |
| Shimadzu 5mL Vial, 12 X
96mm size 5 mL O.D. × height 12 mm × 40 mm vial
type, crimp top |
27324 |
| Shimadzu 5mL Vial, 12 X
96mm size 5 mL O.D. × height 12 mm × 40 mm clear
KG-33 borosilicate glass body vial type, crimp top |
27325 |