Analytical / Chromatography
Air Monitoring
Analytical Reagents
Analytical Standards
Gas Chromatography
HPLC
Microbiology
Microscopy
Sample Preparation
Spectroscopy
Syringes
Titration
TLC
Vials
Videos
Events
Literature
Analytix
The Reporter
Reporter Europe
Product Lines
Vials
Click to see a product selection Guide
Vials > Bottles & Jars > Serum bottles | Amber bottles
Amber bottles
These large amber screw top septum bottles will protect your sample from sunlight. Only glass and Teflon contact the contents.
30 mL (32 x 80 mm)
60 mL (38 x 95 mm)
120 mL (49 x 114 mm)
240 mL (61 x 143 mm)
30 mL (32 x 80 mm) back to top
Product Name Product # 
This size bottle uses a 20-400 cap and 18 mm septum  
Bottle, black phenolic hole cap, PTFE/silicone septum pack of 12 23228
Bottle only pack of 12 23232
black phenolic hole cap (20-400) pack of 100 27176
PTFE/Silicone septum diam. x thickness 18 mm x 0.060 in. pack of 144 23242-U
black phenolic solid cap (20-400), with polyseal liner pack of 144 23238
60 mL (38 x 95 mm) back to top
This size bottle uses a 20-400 cap and 18 mm septum  
Bottle, black phenolic hole cap, PTFE/silicone septum pack of 12 23229
Bottle only pack of 12 23233
black phenolic hole cap (20-400) pack of 100 27176
black phenolic solid cap (20-400), with polyseal liner pack of 144 23238
PTFE/Silicone septum diam. × thickness 18 mm × 0.060 in. pack of 144 23242-U
120 mL (49 x 114 mm) back to top
This size bottle uses a 22-400 cap and 20 mm septum  
Bottle, black phenolic hole cap, PTFE/silicone septum pack of 12 23230-U
Bottle only pack of 12 23234
black phenolic hole cap (22-400) pack of 144 23236
black phenolic solid cap (22-400), with polyseal linerpack of 144 23240-U
tan PTFE(0.002 in.)/silicone septum O.D. × thickness 20 mm × .125 in. pack of 100 27236
PTFE(0.010 in.)/Silicone septum diam. × thickness 20 mm × .100 in. pack of 100 27237-U
240 mL (61 x 143 mm) back to top
This size bottle uses a 24-400 cap and 22 mm septum  
Bottle, black phenolic hole cap, PTFE/silicone septum pack of 12 23231
Bottle only pack of 12 23235
black phenolic hole cap (24-400) pack of 144 23237
black phenolic solid cap (24-400), with polyseal liner pack of 144 23241
PTFE/Silicone septum diam. × thickness 22 mm × 0.100 in. pack of 144 23245-U