|
Product
Lines
Vials |
Click to see a product selection Guide |
| Many products are offered to accompany the vial products
that we sell. We offer a range of vial racks, made of polypropylene material,
to use to store your samples. In addition, safety-coated storage bottles,
glass magnet products, Mini-Vaps, and Block Heaters are listed for usage
in your application. |
| Block
heater |
|
Constant temperatures for
derivatization and other reactions. The block heater features:
Two temperature ranges, ambient to 60 °C, 50 °C to 130 °C
Three-position power switch and two adjustable thermostats
Pilot light shows heat cycling operation
Two models: 120 volts (100 watts, 0.83 amps, 50/60 Hz) and 240 volts (100
watts, 0.42 amps, 50/60 Hz).
Order blocks separately. Block 1 has eight holes (21 mm wide and 30 mm deep),
and accepts our 3 mL or 5 mL micro reaction vessels. Block 2
has twelve holes (17 mm wide and 24 mm deep), and accepts our 0.1 mL
to 2 mL micro reaction vessels. |
Block heater
(8-hole block not included) |
8-hole block for heater |
|
|
| |
|
| Product
Name |
Product # |
| Block Heater AC input 110
V block (not included) |
33315 |
| Block Heater AC input 240
V CE compliant block (not included) |
33318-U |
| Block #1 |
33316 |
| Block #2 |
33317-U |