|
Product
Lines
Vials |
Click to see a product selection Guide |
| Many products are offered to accompany the vial products
that we sell. We offer a range of vial racks, made of polypropylene material,
to use to store your samples. In addition, safety-coated storage bottles,
glass magnet products, Mini-Vaps, and Block Heaters are listed for usage
in your application. |
Mini-Vap Evaporator
/ Concentrator
The 6-Port Mini-Vap concentrator/evaporator processes six miniature vials
or containers at one time. Comes with six stainless steel needles, fine
control needle valve, and three feet (0.9m) of plastic tubing.
The adjustable Mini-Vap is a versatile concentrator/evaporator that can
be used with a single container. The unique design allows for any size container,
from the smallest vial to a 250 mL beaker. The Mini-Vap includes a needle
valve for fine metering of air or nitrogen drying gas. Ideal for a wide
variety of needs in evaporation or concentration. Adjusts to 11 ½
in. |
|
 |
|
| Product
Name |
Product # |
| Mini-Vap Evaporator/Concentrator, 6 port
length × width × base height 7 ½ in. (19 cm)
× 1 ½ in. (4 cm) × ¾ in. (2 cm) for
use with 1-250 mL containers 1 ea |
22971 |
| Mini-Vap Evaporator/Concentrator, adjustable,
size 1 ½ in. × 8 ½ in. 1 ea |
22970 |