|
Product
Lines
Vials |
Click to see a product selection Guide |
| Mininert valves |
Mininert push-button valves are leak-tight closures that
minimize contact with the sample. Only PTFE and glass are in contact when
used with a glass vial. They are easy to use.
1. Push the green button to open the valve
2. Insert the needle through the septum and take a sample
3. Withdraw the needle
4. Push the red button to close the valve
Benefits:
Leak-tight closure - your sample will not have any contact to the
atmosphere
Simple installation - no tools are required to use these valves Replaceable septum - this part is very easy
to replace by pushing the new one into the septum location in the valve.
Mininert valves are available for screw-cap and crimp-top vials. The screw
cap version is available in 13, 15, 18, 20 and 24 mm sizes. The crimp-top
version, for 20 mm I.D. glassware, slides into the neck of the vial and
has a threaded flange which is turned to secure a tight fit.
Additional Resources:
"Operating Instructions for Mininert Valves" (DS11884) |
| Mininert
valve |
back to top |
| Product
Name |
Product # |
| Screw top |
|
| for vials taking 13 mm
cap pack of 12 |
33300 |
| for vials taking 15 mm
cap pack of 12 |
33301 |
| for vials taking 18 mm
cap pack of 12 |
33302 |
| for vials taking 20 mm
cap pack of 12 |
33303 |
| for vials taking 24 mm
cap pack of 12 |
33304 |
| Crimp top |
|
| for vials taking 20 mm
seal pack of 12 |
33305 |
| |
|
| Septa
& insterter |
back to top |
| Product
Name |
Product # |
| Mininert septa, red silicone,
length x diameter 0.308 in. x 0.125 in., pack of 50 |
33310-U |
| Mininert septum inserter
1 ea |
33311 |