| Serum bottles |
|
| These bottles are offered in sizes that
range from 2.5 mL to 100 mL. The 5-100 mL vials require a 20 mm crimp seal
to be used. They are available in both clear and amber glass.We also offer
20 mm crimp seals. |
|
 |
|
| |
|
| Amber
glass |
back to top |
| Product
Name |
Product # |
| 5 mL O.D.
× height 25 mm × 40 mm pack of 36 |
33141-U |
| 5 mL O.D.
× height 25 mm × 40 mm pack of 288 |
33142-U |
| 10 mL O.D. ×
height 25 mm × 50 mm pack of 36 |
33143 |
| 10 mL O.D.
× height 25 mm × 50 mm pack of 288 |
33144 |
| Clear
glass |
back to top |
| Product
Name |
Product # |
| 2.5 mL (requires
13 mm seal) O.D. × height 11.7 mm × 34.5 mm pack
of 288 |
33121 |
|
5 mL O.D. × height 25 mm × 40 mm
pack of 36 |
33102-U |
|
5 mL O.D. × height 25 mm × 40 mm
pack of 288 |
33103 |
|
10 mL O.D. × height 25 mm × 50 mm pack
of 36 |
33104 |
|
10 mL O.D. × height 25 mm × 50 mm pack
of 288 |
33105-U |
|
30 mL O.D. × height 36.6 mm × 62.8 mm pack
of 36 |
33106 |
|
30 mL O.D. × height 36.6 mm × 62.8 mm
pack of 288 |
33107 |
|
50 mL O.D. × height 43 mm × 73 mm pack
of 36 |
33108-U |
|
50 mL O.D. × height 43 mm × 73 mm pack
of 272 |
33109-U |
|
100 mL O.D. × height 51.7 mm × 94.5 mm pack
of 36 |
33110-U |
|
100 mL O.D. × height 51.7 mm × 94.5 mm pack
of 144 |
33111-U |